Gene Transcripts

Gene Symbol
PARP4
Entrez Gene ID
143
Gene Name
poly(ADP-ribose) polymerase family member 4
Gene Aliases
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193
Chromosome Location
Chr. 13: 24420930 - 24512993 on Build GRCh38
UniGene ID
Hs.744855
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006437.3 NP_006428.2 21 2609
XM_011534931.1 XP_011533233.1 20 2659
XM_011534932.2 XP_011533234.1 21 2826
GenBank mRNA AF057160.1 - 21 2555
AF158255.1 - 21 2609
AK304399.1 - 21 2577
D79999.2 - 20 2508

View Full Product Details