Gene Transcripts

Gene Symbol
NAP1L4
Entrez Gene ID
4676
Gene Name
nucleosome assembly protein 1 like 4
Gene Aliases
NAP1L4b, NAP2, NAP2L, hNAP2
Chromosome Location
Chr. 11: 2944430 - 2992377 on Build GRCh38
UniGene ID
Hs.731784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005969.3 NP_005960.1 9 864
GenBank mRNA AB500094.1 - 7 723
AK299515.1 - 9 787
AK312599.1 - 9 872
AK316242.1 - 9 781
AK316324.1 - 9 802
AK316548.1 - 10 921
BC022090.2 - 9 774 3029455
U77456.1 - 9 872

View Full Product Details