Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022774.1 | NP_073611.1 | 3 | 878 | |
XM_005271125.3 | XP_005271182.1 | 4 | 1126 | ||
XM_005271126.3 | XP_005271183.1 | 3 | 938 | ||
XM_005271128.3 | XP_005271185.1 | 2 | 875 | ||
XM_011541970.2 | XP_011540272.1 | 2 | 820 | ||
XM_017002099.1 | XP_016857588.1 | 4 | 1550 | ||
XM_017002100.1 | XP_016857589.1 | 4 | 1105 | ||
XM_017002101.1 | XP_016857590.1 | 4 | 1076 | ||
XM_017002102.1 | XP_016857591.1 | 3 | 1478 | ||
XM_017002103.1 | XP_016857592.1 | 3 | 904 | ||
XM_017002104.1 | XP_016857593.1 | 2 | 857 | ||
GenBank mRNA | AK023245.1 | - | 2 | 1705 | |
AK024797.1 | - | 3 | 878 | ||
AK094092.1 | - | 3 | 1700 | ||
BC021969.2 | - | 4 | 1039 | 3952573 |