Gene Transcripts

Gene Symbol
Hexa
Entrez Gene ID
15211
Gene Name
hexosaminidase A
Gene Aliases
Hex-1
Chromosome Location
Chr. 9: 59539545 - 59565109 on Build GRCm38
UniGene ID
Mm.2284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010421.4 NP_034551.2 13 1519
GenBank mRNA AK075895.1 - 13 1495
AK075911.1 - 13 1520
AK080777.1 - 13 1492
AK144168.1 - 13 1647
AK150444.1 - 13 1490
AK151960.1 - 13 1494
AK159091.1 - 14 1490
AK159814.1 - 13 1505
AK159890.1 - 13 1492
AK160112.1 - 13 1479
AK168186.1 - 13 1519
AK170903.1 - 13 1506
BC010755.1 - 13 1488 4225233
U07631.1 - 13 1511
X64331.1 - 13 1462

View Full Product Details