Gene Transcripts

Gene Symbol
Tmem63a
Entrez Gene ID
208795
Gene Name
transmembrane protein 63a
Gene Aliases
BC014795, Tmem64a
Chromosome Location
Chr. 1: 180941225 - 180975106 on Build GRCm38
UniGene ID
Mm.185518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144794.2 NP_659043.1 21 2153
GenBank mRNA AK049360.1 - 21 2150
AK144899.1 - 21 2206
AK170559.1 - 21 2169
AK173027.1 - 21 2146
BC014795.1 - 21 2126 3961992
BC019442.1 - 21 2153 4036849

View Full Product Details