Gene Transcripts

Gene Symbol
Fads1
Entrez Gene ID
76267
Gene Name
fatty acid desaturase 1
Gene Aliases
0710001O03Rik, A930006B21Rik, AI317215, DSD
Chromosome Location
Chr. 19: 10182888 - 10196874 on Build GRCm38
UniGene ID
Mm.30158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146094.2 NP_666206.1 10 1301
GenBank mRNA AB072976.1 - 10 1216
AF061504.1 - 10 1203
AK033308.1 - 8 658
AK083959.1 - 10 1163
AK154367.1 - 10 1251
BC022139.1 - 10 1246 5100199
BC026831.1 - 10 1268 4239695
BC026848.1 - 10 1241 4196030
BC063053.1 - 10 1161 5696722
KF987079.1 - 10 1185

View Full Product Details