Gene Transcripts

Gene Symbol
B9d1
Entrez Gene ID
27078
Gene Name
B9 protein domain 1
Gene Aliases
AW045994, B9, Eppb9
Chromosome Location
Chr. 11: 61505168 - 61512929 on Build GRCm38
UniGene ID
Mm.261226
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013717.2 NP_038745.1 7 550
GenBank mRNA AB030483.1 - 7 550
AK003038.1 - 7 545
AK010355.1 - 7 579
AK131701.1 - 7 541
BC008113.1 - 7 518 3483517

View Full Product Details