Gene Transcripts

Gene Symbol
Hexa
Entrez Gene ID
15211
Gene Name
hexosaminidase A
Gene Aliases
Hex-1
Chromosome Location
Chr. 9: 59539545 - 59565109 on Build GRCm38
UniGene ID
Mm.2284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010421.4 NP_034551.2 6 645
GenBank mRNA AK075895.1 - 6 621
AK075911.1 - 6 646
AK080777.1 - 6 618
AK144168.1 - 6 773
AK150444.1 - 6 616
AK151960.1 - 6 620
AK159091.1 - 6 637
AK159814.1 - 6 631
AK159890.1 - 6 618
AK160112.1 - 6 605
AK167013.1 - 6 631
AK168186.1 - 6 645
AK170903.1 - 6 632
BC010755.1 - 6 614 4225233
U07631.1 - 6 637
X64331.1 - 6 588

View Full Product Details