Gene Transcripts

Gene Symbol
Dnajc25
Entrez Gene ID
72429
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 25
Gene Aliases
2010109C08Rik, 2010203O07Rik
Chromosome Location
Chr. 4: 58994810 - 59023398 on Build GRCm38
UniGene ID
Mm.317801
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033165.3 NP_001028337.2 3 820
GenBank mRNA AK136359.1 - 3 514
BC119039.1 - 3 585 8733788
BC120693.1 - 3 585 40129616
BX512063.1 - 1 110

View Full Product Details