Gene Transcripts

Gene Symbol
Gabrg1
Entrez Gene ID
140674
Gene Name
gamma-aminobutyric acid (GABA) A receptor, gamma 1
Gene Aliases
Chromosome Location
Chr. 14: 39964588 - 40049354 on Build Rnor_6.0
UniGene ID
Rn.10366
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080586.1 NP_542153.1 9 1304
GenBank mRNA X57514.1 - 9 1304

View Full Product Details