Gene Transcripts

Gene Symbol
Polr2f
Entrez Gene ID
83503
Gene Name
polymerase (RNA) II (DNA directed) polypeptide F
Gene Aliases
Chromosome Location
Chr. 7: 120380555 - 120392214 on Build Rnor_6.0
UniGene ID
Rn.28212
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031335.3 NP_112625.1 1, 2 64
GenBank mRNA AB017711.1 - 1, 2 64
BC083552.1 - 1, 2 104 7190444

View Full Product Details