Gene Transcripts

Gene Symbol
SLC30A5
Entrez Gene ID
64924
Gene Name
solute carrier family 30 member 5
Gene Aliases
ZNT5, ZNTL1, ZTL1, ZnT-5
Chromosome Location
Chr. 5: 69093949 - 69131072 on Build GRCh38
UniGene ID
Hs.631975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251969.1 NP_001238898.1 3 1200
NM_022902.4 NP_075053.2 4 654
NM_024055.4 NP_076960.1 4 1323
XM_005248569.3 XP_005248626.1 3 484
XM_006714672.3 XP_006714735.1 4 648
XM_017009749.1 XP_016865238.1 3 484
GenBank mRNA AF461760.1 - 4 548
AK022558.1 - 1 18
AK022818.1 - 2 137
AK292925.1 - 4 518
AY089991.1 - 4 347
AY358505.1 - 4 565
BC000808.1 - 4 1168
BC008198.1 - 1 38
BC017441.1 - 4 1281
BX537394.1 - 4 347

View Full Product Details