Gene Transcripts

Gene Symbol
KLHL23
Entrez Gene ID
151230
Gene Name
kelch like family member 23
Gene Aliases
DITHP
Chromosome Location
Chr. 2: 169733846 - 169751886 on Build GRCh38
UniGene ID
Hs.655150
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144711.5 NP_653312.2 4 1802
GenBank mRNA AK095131.1 - 1378
BC010437.1 - 1636 3354344
BC016950.1 - 1 94
BQ343827.1 - 1 80
R08525.1 - 2 230

Gene Symbol
PHOSPHO2-KLHL23
Entrez Gene ID
100526832
Gene Name
PHOSPHO2-KLHL23 readthrough
Gene Aliases
Chromosome Location
Chr. 2: 169694454 - 169751886 on Build GRCh38
UniGene ID
Hs.655150
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199290.1 NP_001186219.1 6 1933
GenBank mRNA AK090653.1 - 6 1896
AX746559.1 - 6 1896

View Full Product Details