Gene Transcripts
- Gene Symbol
- DOK7
- Entrez Gene ID
- 285489
- Gene Name
- docking protein 7
- Gene Aliases
- C4orf25, CMS10, CMS1B
- Chromosome Location
- Chr. 4: 3463306 - 3501476 on Build GRCh38
- UniGene ID
- Hs.122110
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164673.1 |
NP_001158145.1
|
5, 6 |
709 |
|
NM_001256896.1 |
NP_001243825.1
|
2, 3 |
418 |
|
NM_001301071.1 |
NP_001288000.1
|
5, 6 |
720 |
|
NM_173660.4 |
NP_775931.3
|
5, 6 |
720 |
|
XM_011513435.2 |
XP_011511737.1
|
5, 6 |
720 |
|
XM_011513437.2 |
XP_011511739.1
|
5, 6 |
709 |
|
XM_017008038.1 |
XP_016863527.1
|
3, 4 |
283 |
|
GenBank mRNA |
AB220918.1 |
- |
5, 6 |
650 |
|
AK091037.1 |
- |
5, 6 |
704 |
|
AK096456.1 |
- |
2, 3 |
418 |
|
BC043568.1 |
- |
1, 2 |
57 |
|
BC062369.1 |
- |
1, 2 |
112 |
|
BC131544.1 |
- |
5, 6 |
657 |
40085498 |
BC141852.1 |
- |
5, 6 |
691 |
8860352 |
DA823926.1 |
- |
2, 3 |
419 |
|
View Full Product Details