Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007975.3 | NP_032001.2 | 1 | 105 | |
GenBank mRNA | AF080215.1 | - | 1 | 82 | |
AK036427.1 | - | 1 | 106 | ||
AK153720.1 | - | 1 | 99 | ||
AK156729.1 | - | 1 | 106 | ||
BC119461.2 | - | 1 | 101 | 40045612 |