Gene Transcripts

Gene Symbol
Rps18
Entrez Gene ID
20084
Gene Name
ribosomal protein S18
Gene Aliases
H-2Ke3, H2-Ke3, Ke-3, ke3
Chromosome Location
Chr. 17: 33951999 - 33955641 on Build GRCm38
UniGene ID
Mm.298745
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011296.2 NP_035426.1 3 253
GenBank mRNA AK050626.1 - 3 276
AK151580.1 - 3 276
AK159437.1 - 3 276
AK159821.1 - 3 276
AK169246.1 - 3 276
AK169732.1 - 2 897
BC081458.1 - 2 241 5712669
BC081459.1 - 3 253 6407328
BC093500.1 - 3 254 4207659
BC100604.1 - 3 275 30120583
M76763.1 - 3 254

View Full Product Details