Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011855.4 | NP_035985.2 | 31 | 7125 | |
GenBank mRNA | AB025410.1 | - | 31 | 7127 | |
AK037897.1 | - | 4 | 1814 | ||
BC138860.1 | - | 30 | 7108 | 8861882 | |
BC138861.1 | - | 30 | 7108 | 8861883 |