Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011994.2 | NP_036124.2 | 8 | 2069 | |
GenBank mRNA | AK051445.1 | - | 8 | 2043 | |
AK082588.1 | - | 8 | 2041 | ||
AK087653.1 | - | 8 | 1688 | ||
AK134763.1 | - | 3 | 380 | ||
AK161027.1 | - | 8 | 1730 | ||
BC019187.1 | - | 8 | 2013 | 5027149 | |
Z48670.1 | - | 8 | 2073 |