Gene Transcripts

Gene Symbol
Plgrkt
Entrez Gene ID
67759
Gene Name
plasminogen receptor, C-terminal lysine transmembrane protein
Gene Aliases
1110007H22Rik, 5033414D02Rik, AI852040, Plg-R(KT), Plg-RKT
Chromosome Location
Chr. 19: 29348599 - 29361885 on Build GRCm38
UniGene ID
Mm.275511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026362.2 NP_080638.1 5 498
GenBank mRNA AK017171.1 - 5 523
AK017215.1 - 5 499
AK168454.1 - 5 474
AK170700.1 - 6 572
BC024953.1 - 5 457 4951101

View Full Product Details