Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CPM EBGEX Assays ›
Gene Symbol:
CPM
Gene Name:
carboxypeptidase M
Gene Aliases:
-
Chromosome Location:
Chr.12: 68842197 - 68963469 on Build GRCh38
UniGene ID:
Hs.654387
Species:
Human
Species Specific ID (Flybase ID):
-
Type:
1
Detection Label:
Alexa Fluor 546
ProductType:
ViewRNA Cell Plus Probe Set
Assay ID
VA1-3002138-VCP
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CPM
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_001005502.3
NP_001005502.1
NM_001413387.1
NP_001400316.1
NM_001413388.1
NP_001400317.1
NM_001413389.1
NP_001400318.1
NM_001413390.1
NP_001400319.1
NM_001413391.1
NP_001400320.1
NM_001413392.1
NP_001400321.1
NM_001413393.1
NP_001400322.1
NM_001413394.1
NP_001400323.1
NM_001413395.1
NP_001400324.1
NM_001413396.1
NP_001400325.1
NM_001413397.1
NP_001400326.1
NM_001413398.1
NP_001400327.1
NM_001413399.1
NP_001400328.1
NM_001413400.1
NP_001400329.1
NM_001413401.1
NP_001400330.1
NM_001413402.1
NP_001400331.1
NM_001413403.1
NP_001400332.1
NM_001413404.1
NP_001400333.1
NM_001874.5
NP_001865.1
NM_198320.5
NP_938079.1
NR_182142.1
NR_182143.1
GenBank mRNA
AF368463.1
-
AI524542.1
-
AK127859.1
-
AK130146.1
-
AK313180.1
-
AU106031.1
-
BC016943.1
-
BC022276.1
-
BG677445.1
-
BM049082.1
-
BQ181238.1
-
BQ431065.1
-
BQ706414.1
-
BQ773666.1
-
CB241489.1
-
CB959900.1
-
CB989009.1
-
CB993834.1
-
CD109360.1
-
DA205368.1
-
DA831499.1
-
DA833423.1
-
DB006895.1
-
J04970.2
-
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
peptide metabolic process
anatomical structure morphogenesis
protein processing
carboxypeptidase activity
metallocarboxypeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-6479c7f679-zb4k5:80/100.66.74.142:80.
git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.50.0-Offline