Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VCLQKALNH LREIWELIGI PEDQRLQRTE VVKKHIKELL DMMIAEEESL KERLIKSISV CQKELNTLCS ELHVEPFQEE GETTILQLEK DLRTQVELMR KQKKERKQEL KLLQEQDQEL CEILCMPHYD IDSASVPSLE ELNQFRQHVT TLRETKASRR EEFV (13-175 aa encoded by BC005140)
This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anaphase spindle elongation 1 homolog; MGC1671; MGC3669; PRC1; protein regulating cytokinesis 1; Protein regulator of cytokinesis 1
Gene Aliases: ASE1; D7Ertd348e; PRC1
UniProt ID: (Human) O43663, (Mouse) Q99K43
Entrez Gene ID: (Human) 9055, (Mouse) 233406, (Rat) 308761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support