Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVPVAVTAA VAPVLSINSD FSDLREIKKQ LLLIAGLTRE RGLLHSSKWS AELAFSLPAL PLAELQPPPP ITEEDAQDMD AYTLAKAYFD VKEYDRAAHF LHGCNSKKAY FLYMYSRYLV RAILKCH (1-126 aa encoded by BC005258)
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). Ubr2 (Ubiquitin-protein ligase E3-alpha-2), also known as N-recognin-2, is a 1755 amino acid protein that contains one UBR-type zinc finger and one RING-type zinc finger. Participating in protein modification events within the N-end rule pathway, Ubr2 functions as an E3 ubiquitin-protein ligase that recognizes and binds proteins that contain destabilizing N-terminal residues, thereby leading to their ubiquitination and subsequent degradation. Mice lacking Ubr2 are infertile due to defects in male meiosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anaphase promoting complex subunit 8; Anaphase-promoting complex subunit 8; APC8; CDC23 (cell division cycle 23, yeast, homolog); cell division cycle 23 homolog; Cell division cycle protein 23 homolog; Cyclosome subunit 8
Gene Aliases: 6030435O18; ANAPC8; APC8; CDC23; CUT23; D18Ertd243e
UniProt ID: (Human) Q9UJX2, (Mouse) Q8BGZ4
Entrez Gene ID: (Human) 8697, (Mouse) 52563, (Rat) 291689
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support