Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEKRTCALC PKDVEYNVLY FAQSENIAAH ENCLLYSSGL VECEDQDPLN PDRSFDVESV KKEIQRGRKL KCKFCHKRGA TVGCDLKNCN KNYHFFCAKK DDAVPQSDGV RGIYKLLCQQ HAQFPIIAQS AKFSGVKRKR GRKKPLSGNH VQPPETMKCN TFIRQVKEEH GRHTDATVKV PFLKKCKEAG LLNYLLEEIL DKVHSIPEKL MDETTSESDY EEIGSALFDC RLFEDTFVNF QAAIEKKIHA SQQRWQQLKE EIELLQDLKQ TLCSFQENRD LMSSSTSISS LSY (1-292 aa encoded by BC017212)
PHF11 contains two PHD zinc fingers and probably regulates transcription. Distinctive splice variants were expressed in immune tissues and cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atopic; BRCA1 C-terminus-associated protein; IgE responsiveness; IgE responsiveness (atopic); NY-REN-34 antigen; PHD finger protein 11; PHD finger protein 11 family member; PHD finger protein 11-like; PHD finger protein 11A; PHD finger protein 11D; Renal carcinoma antigen NY-REN-34; RP11-185C18.3
Gene Aliases: 4933417L10Rik; APY; BCAP; D14Ertd668e; IGEL; IGER; IGHER; NY-REN-34; NYREN34; PHF11; Phf11-4; Phf11a; Phf11d; Phf11l
UniProt ID: (Human) Q9UIL8, (Mouse) Q8BVM9, (Mouse) A6H5X4
Entrez Gene ID: (Human) 51131, (Mouse) 219131, (Mouse) 219132
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support