Product References
LncRNA WDR11-AS1 Promotes Extracellular Matrix Synthesis in Osteoarthritis by Directly Interacting with RNA-Binding Protein PABPC1 to Stabilize SOX9 Expression.
International journal of molecular sciences
Huang H,Yan J,Lan X,Guo Y,Sun M,Zhao Y,Zhang F,Sun J,Lu S
Tue Jan 03 00:00:00 EST 2023
LncRNA WDR11-AS1 Promotes Extracellular Matrix Synthesis in Osteoarthritis by Directly Interacting with RNA-Binding Protein PABPC1 to Stabilize SOX9 Expression.
International journal of molecular sciences
Huang H,Yan J,Lan X,Guo Y,Sun M,Zhao Y,Zhang F,Sun J,Lu S
Tue Jan 03 00:00:00 EST 2023
LncRNA BC promotes lung adenocarcinoma progression by modulating IMPAD1 alternative splicing.
Clinical and translational medicine
Chen QW,Cai QQ,Yang Y,Dong S,Liu YY,Chen ZY,Kang CL,Qi B,Dong YW,Wu W,Zhuang LP,Shen YH,Meng ZQ,Wu XZ
Sun Jan 01 00:00:00 EST 2023
CircPTK2/PABPC1/SETDB1 axis promotes EMT-mediated tumor metastasis and gemcitabine resistance in bladder cancer.
Cancer letters
Meng X,Xiao W,Sun J,Li W,Yuan H,Yu T,Zhang X,Dong W
Wed Feb 01 00:00:00 EST 2023
TIAR and FMRP shape pro-survival nascent proteome of leukemia cells in the bone marrow microenvironment.
iScience
Wolczyk M,Serwa R,Kominek A,Klejman A,Milek J,Chwałek M,Turos-Korgul L,Charzyńska A,Dabrowski M,Dziembowska M,Skorski T,Piwocka K,Podszywalow-Bartnicka P
Fri Apr 21 00:00:00 EDT 2023
T-2 toxin and deoxynivalenol (DON) exert distinct effects on stress granule formation depending on altered activity of SIRT1.
Ecotoxicology and environmental safety
Jiang T,Qi X,Lin R,Jiang J,Wen J,Deng Y
Sat Jul 01 00:00:00 EDT 2023
The deubiquitinating enzyme STAMBP is a newly discovered driver of triple-negative breast cancer progression that maintains RAI14 protein stability.
Experimental & molecular medicine
Yang Q,Yan D,Zou C,Xue Q,Lin S,Huang Q,Li X,Tang D,Chen X,Liu J
Tue Nov 01 00:00:00 EDT 2022
Viral proteases activate the CARD8 inflammasome in the human cardiovascular system.
The Journal of experimental medicine
Nadkarni R,Chu WC,Lee CQE,Mohamud Y,Yap L,Toh GA,Beh S,Lim R,Fan YM,Zhang YL,Robinson K,Tryggvason K,Luo H,Zhong F,Ho L
Mon Oct 03 00:00:00 EDT 2022
YTHDF3 modulates hematopoietic stem cells by recognizing RNA m6A modification on Ccnd1.
Haematologica
Zhang X,Cong T,Wei L,Zhong B,Wang X,Sun J,Wang S,Xu MM,Zhu P,Jiang H,Wang J
Sat Oct 01 00:00:00 EDT 2022
YTHDF3 modulates hematopoietic stem cells by recognizing RNA m6A modification on Ccnd1.
Haematologica
Zhang X,Cong T,Wei L,Zhong B,Wang X,Sun J,Wang S,Xu MM,Zhu P,Jiang H,Wang J
Sat Oct 01 00:00:00 EDT 2022
E3 ligase MKRN3 is a tumor suppressor regulating PABPC1 ubiquitination in non-small cell lung cancer.
The Journal of experimental medicine
Li K,Zheng X,Tang H,Zang YS,Zeng C,Liu X,Shen Y,Pang Y,Wang S,Xie F,Lu X,Luo Y,Li Z,Bi W,Jia X,Huang T,Wei R,Huang K,Chen Z,Zhu Q,He Y,Zhang M,Gu Z,Xiao Y,Zhang X,Fletcher JA,Wang Y
Mon Aug 02 00:00:00 EDT 2021
Enhancer-Driven lncRNA BDNF-AS Induces Endocrine Resistance and Malignant Progression of Breast Cancer through the RNH1/TRIM21/mTOR Cascade.
Cell reports
Lin X,Dinglin X,Cao S,Zheng S,Wu C,Chen W,Li Q,Hu Q,Zheng F,Wu Z,Lin DC,Yao Y,Xu X,Xie Z,Liu Q,Yao H,Hu H
Tue Jun 09 00:00:00 EDT 2020
NcPuf1 Is a Key Virulence Factor in Neospora caninum.
Pathogens (Basel, Switzerland)
Wang C,Yang C,Liu J,Liu Q
Wed Dec 02 00:00:00 EST 2020
An oncopeptide regulates m6A recognition by the m6A reader IGF2BP1 and tumorigenesis.
Nature communications
Zhu S,Wang JZ,Chen D,He YT,Meng N,Chen M,Lu RX,Chen XH,Zhang XL,Yan GR
Fri Apr 03 00:00:00 EDT 2020
Purification of cross-linked RNA-protein complexes by phenol-toluol extraction.
Nature communications
Urdaneta EC,Vieira-Vieira CH,Hick T,Wessels HH,Figini D,Moschall R,Medenbach J,Ohler U,Granneman S,Selbach M,Beckmann BM
Fri Mar 01 00:00:00 EST 2019
An atlas of bloodstream-accessible bone marrow proteins for site-directed therapy of acute myeloid leukemia.
Leukemia
Angenendt L,Reuter S,Kentrup D,Benk AS,Neumann F,Hüve J,Martens AC,Schwöppe C,Kessler T,Schmidt LH,Sauer T,Brand C,Mikesch JH,Lenz G,Mesters RM,Müller-Tidow C,Hartmann W,Wardelmann E,Neri D,Berdel WE,Roesli C,Schliemann C
Thu Feb 01 00:00:00 EST 2018
TDP-43 aggregation mirrors TDP-43 knockdown, affecting the expression levels of a common set of proteins.
Scientific reports
Prpar Mihevc S,Baralle M,Buratti E,Rogelj B
Mon Sep 26 00:00:00 EDT 2016
TDP-43 aggregation mirrors TDP-43 knockdown, affecting the expression levels of a common set of proteins.
Scientific reports
Prpar Mihevc S,Baralle M,Buratti E,Rogelj B
Mon Sep 26 00:00:00 EDT 2016
TDP-43 aggregation mirrors TDP-43 knockdown, affecting the expression levels of a common set of proteins.
Scientific reports
Prpar Mihevc S,Baralle M,Buratti E,Rogelj B
Mon Sep 26 00:00:00 EDT 2016
TDP-43 aggregation mirrors TDP-43 knockdown, affecting the expression levels of a common set of proteins.
Scientific reports
Prpar Mihevc S,Baralle M,Buratti E,Rogelj B
Mon Sep 26 00:00:00 EDT 2016