Product References
Engineered Human Dendritic Cell Exosomes as Effective Delivery System for Immune Modulation.
International journal of molecular sciences
Elsayed R,Elashiry M,Tran C,Yang T,Carroll A,Liu Y,Hamrick M,Cutler CW
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Tue Jul 11 00:00:00 EDT 2023
B cell-intrinsic changes with age do not impact antibody-secreting cell formation but delay B cell participation in the germinal centre reaction.
Aging cell
Lee JL,Fra-Bido SC,Burton AR,Innocentin S,Hill DL,Linterman MA
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Thu Sep 01 00:00:00 EDT 2022
Macrophage Membrane-Camouflaged shRNA and Doxorubicin: A pH-Dependent Release System for Melanoma Chemo-Immunotherapy.
Research (Washington, D.C.)
Yang C,Ming Y,Zhou K,Hao Y,Hu D,Chu B,He X,Yang Y,Qian Z
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2022
Response of human macrophages to gamma radiation is mediated via expression of endogenous retroviruses.
PLoS pathogens
Mikhalkevich N,O'Carroll IP,Tkavc R,Lund K,Sukumar G,Dalgard CL,Johnson KR,Li W,Wang T,Nath A,Iordanskiy S
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Different Induction of PD-L1 (CD274) and PD-1 (CD279) Expression in THP-1-Differentiated Types 1 and 2 Macrophages.
Journal of inflammation research
Lai CY,Tseng PC,Chen CL,Satria RD,Wang YT,Lin CF
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Wed Apr 27 00:00:00 EDT 2022
Recombinant human IL-37 inhibited endometriosis development in a mouse model through increasing Th1/Th2 ratio by inducing the maturation of dendritic cells.
Reproductive biology and endocrinology : RB&E
Li L,Liao Z,Ye M,Jiang J
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Tue Aug 24 00:00:00 EDT 2021
Immunological Properties of Neural Crest Cells Derived from Human Induced Pluripotent Stem Cells.
Stem cells and development
Fujii S,Yoshida S,Inagaki E,Hatou S,Tsubota K,Takahashi M,Shimmura S,Sugita S
11-0809 was used in Flow cytometry/Cell sorting to assess the immunological properties of human induced pluripotent stem cells-neural crest cells.
Tue Jan 01 00:00:00 EST 2019
Astragalus polysaccharides (PG2) Enhances the M1 Polarization of Macrophages, Functional Maturation of Dendritic Cells, and T Cell-Mediated Anticancer Immune Responses in Patients with Lung Cancer.
Nutrients
Bamodu OA,Kuo KT,Wang CH,Huang WC,Wu ATH,Tsai JT,Lee KY,Yeh CT,Wang LS
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Fri Sep 20 00:00:00 EDT 2019
Photodynamic therapy reduces the inhibitory effect of osteosarcoma cells on dendritic cells by upregulating HSP70.
Oncology letters
Zhang F,Zhu Y,Fan G,Hu S
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Dexamethasone and Monophosphoryl Lipid A Induce a Distinctive Profile on Monocyte-Derived Dendritic Cells through Transcriptional Modulation of Genes Associated With Essential Processes of the Immune Response.
Frontiers in immunology
García-González PA,Schinnerling K,Sepúlveda-Gutiérrez A,Maggi J,Mehdi AM,Nel HJ,Pesce B,Larrondo ML,Aravena O,Molina MC,Catalán D,Thomas R,Verdugo RA,Aguillón JC
11-0809 was used in Flow cytometry/Cell sorting to study the transcriptional profile of monocyte-derived dendritic cells from healthy subjects to identify molecular regulators and pathways associated with the induction of tolerogenic properties in tolerogenic dendritic cells.
Wed Nov 20 00:00:00 EST 2019
Antigen-Presenting Human γδ T Cells Promote Intestinal CD4+ T Cell Expression of IL-22 and Mucosal Release of Calprotectin.
Journal of immunology (Baltimore, Md. : 1950)
Tyler CJ,McCarthy NE,Lindsay JO,Stagg AJ,Moser B,Eberl M
11-0809 was used in Flow cytometry/Cell sorting to study the role of γδ T cells in the regulation of IL-22 expression.
Mon May 01 00:00:00 EDT 2017
MALP-2, an agonist of TLR6, promotes the immune status without affecting the differentiation capacity of umbilical cord mesenchymal stem cells.
Experimental and therapeutic medicine
Wu X,Xu L,Shen Y,Yu N,Zhang Y,Guo T
11-0809 was used in Flow cytometry/Cell sorting to assess the role of TLR6 in the immune status of umbilical cord mesenchymal stem cells, by stimulation with the TLR6 agonist MALP-2.
Fri Dec 01 00:00:00 EST 2017
Immunoprofiling of Adult-Derived Human Liver Stem/Progenitor Cells: Impact of Hepatogenic Differentiation and Inflammation.
Stem cells international
El-Kehdy H,Sargiacomo C,Fayyad-Kazan M,Fayyad-Kazan H,Lombard C,Lagneaux L,Sokal E,Najar M,Najimi M
11-0809-42 was used in Flow cytometry/Cell sorting to suggest CD200 as a key factor in regulating the immunobiology of differentiated ADHLSCs.
Sat Apr 09 00:00:00 EDT 2022
CD80 down-regulation is associated to aberrant DNA methylation in non-inflammatory colon carcinogenesis.
BMC cancer
Scarpa M,Scarpa M,Castagliuolo I,Erroi F,Basato S,Brun P,Angriman I,Castoro C
11-0809 was used in Flow cytometry/Cell sorting to indicate that the failure of immune surveillance mechanisms in non-inflammatory colon carcinogenesis may affect CD80 expression.
Mon Jul 04 00:00:00 EDT 2016
The role of Toll-like receptor 3 and 4 in regulating the function of mesenchymal stem cells isolated from umbilical cord.
International journal of molecular medicine
Zhang L,Liu D,Pu D,Wang Y,Li L,He Y,Li Y,Li L,Qiu Z,Zhao S,Li W
11-0809 was used in Western Blotting to examine the role of the TLR3 and TLR4 pathways in modulating the immune properties of human umbilical cord-derived MSCs.
Wed Apr 01 00:00:00 EDT 2015
A DNA vaccine encoding mutated HPV58 mE6E7-Fc-GPI fusion antigen and GM-CSF and B7.1.
OncoTargets and therapy
Wang H,Yu J,Li L
Published figure using CD80 (B7-1) monoclonal antibody (Product # 11-0809-42) in Immunofluorescence
Wed Nov 25 00:00:00 EST 2015
HIV-1 and HIV-2 differentially mature plasmacytoid dendritic cells into IFN-producing cells or APCs.
Journal of immunology (Baltimore, Md. : 1950)
Royle CM,Graham DR,Sharma S,Fuchs D,Boasso A
11-0809 was used in Flow cytometry/Cell sorting to study the factors causing the observed differences in pathogenicity between HIV-1 and HIV-2.
Wed Oct 01 00:00:00 EDT 2014
Haemophilus ducreyi-induced interleukin-10 promotes a mixed M1 and M2 activation program in human macrophages.
Infection and immunity
Li W,Katz BP,Spinola SM
11-0809 was used in Flow cytometry/Cell sorting to investigate the contribution of IL-10-modulated macrophage polarisation to H. ducreyi clearance during human infection.
Sat Dec 01 00:00:00 EST 2012
Down-regulation of MHC class II expression through inhibition of CIITA transcription by lytic transactivator Zta during Epstein-Barr virus reactivation.
Journal of immunology (Baltimore, Md. : 1950)
Li D,Qian L,Chen C,Shi M,Yu M,Hu M,Song L,Shen B,Guo N
11-0809 was used in Flow cytometry/Cell sorting to indicate that Zta may act as an inhibitor of the MHC class II pathway, suppressing CIITA transcription.
Sun Feb 15 00:00:00 EST 2009
Renal allograft rejection is prevented by adoptive transfer of anergic T cells in nonhuman primates.
The Journal of clinical investigation
Bashuda H,Kimikawa M,Seino K,Kato Y,Ono F,Shimizu A,Yagita H,Teraoka S,Okumura K
Fri Jul 01 00:00:00 EDT 2005