Product References
TRAF6 controls T cell homeostasis by maintaining the equilibrium of MALT1 scaffolding and protease functions.
Frontiers in immunology
O'Neill TJ,Gewies A,Seeholzer T,Krappmann D
Published figure using CD278 (ICOS) monoclonal antibody (Product # 11-9949-82) in Flow Cytometry
Mon Feb 13 00:00:00 EST 2023
HSF1 promotes CD69+ Treg differentiation to inhibit colitis progression.
Theranostics
Yu L,Zhou B,Zhu Y,Li L,Zhong Y,Zhu L,Wang H,Chen H,Xu J,Guo T,Feng L,Wang X,Cai Z,Wang J,Jin H
Published figure using CD278 (ICOS) monoclonal antibody (Product # 11-9949-82) in Flow Cytometry
Tue Apr 18 00:00:00 EDT 2023
Organoids capture tissue-specific innate lymphoid cell development in mice and humans.
Cell reports
Jowett GM,Read E,Roberts LB,Coman D,Vilà González M,Zabinski T,Niazi U,Reis R,Trieu TJ,Danovi D,Gentleman E,Vallier L,Curtis MA,Lord GM,Neves JF
Published figure using CD278 (ICOS) monoclonal antibody (Product # 11-9949-82) in Flow Cytometry
Tue Aug 30 00:00:00 EDT 2022
Chronic stress physically spares but functionally impairs innate-like invariant T cells.
Cell reports
Rudak PT,Choi J,Parkins KM,Summers KL,Jackson DN,Foster PJ,Skaro AI,Leslie K,McAlister VC,Kuchroo VK,Inoue W,Lantz O,Haeryfar SMM
11-9949-82 was used in Flow Cytometry to report that long-term stress surprisingly abrogates both T helper 1 (TH1)- and TH2-type responses orchestrated by invariant natural killer T (iNKT) cells.
Tue Apr 13 00:00:00 EDT 2021
Dermal Adipocyte Lipolysis and Myofibroblast Conversion Are Required for Efficient Skin Repair.
Cell stem cell
Shook BA,Wasko RR,Mano O,Rutenberg-Schoenberg M,Rudolph MC,Zirak B,Rivera-Gonzalez GC,López-Giráldez F,Zarini S,Rezza A,Clark DA,Rendl M,Rosenblum MD,Gerstein MB,Horsley V
11-9949-82 was used in Flow Cytometry to identify an essential role for adipocyte lipolysis in regulating inflammation and repair after injury in skin.
Thu Jun 04 00:00:00 EDT 2020
Toxin-Triggered Interleukin-1 Receptor Signaling Enables Early-Life Discrimination of Pathogenic versus Commensal Skin Bacteria.
Cell host & microbe
Leech JM,Dhariwala MO,Lowe MM,Chu K,Merana GR,Cornuot C,Weckel A,Ma JM,Leitner EG,Gonzalez JR,Vasquez KS,Diep BA,Scharschmidt TC
11-9949-82 was used in Flow Cytometry to study how skin colonization by commensal Staphylococcus epidermidis facilitates immune tolerance preferentially in neonates via induction of antigen-specific regulatory T cells.
Wed Dec 11 00:00:00 EST 2019
Toxin-Triggered Interleukin-1 Receptor Signaling Enables Early-Life Discrimination of Pathogenic versus Commensal Skin Bacteria.
Cell host & microbe
Leech JM,Dhariwala MO,Lowe MM,Chu K,Merana GR,Cornuot C,Weckel A,Ma JM,Leitner EG,Gonzalez JR,Vasquez KS,Diep BA,Scharschmidt TC
11-9949-82 was used in Flow Cytometry to study how skin colonization by commensal Staphylococcus epidermidis facilitates immune tolerance preferentially in neonates via induction of antigen-specific regulatory T cells.
Wed Dec 11 00:00:00 EST 2019
Pulsatile MEK Inhibition Improves Anti-tumor Immunity and T Cell Function in Murine Kras Mutant Lung Cancer.
Cell reports
Choi H,Deng J,Li S,Silk T,Dong L,Brea EJ,Houghton S,Redmond D,Zhong H,Boiarsky J,Akbay EA,Smith PD,Merghoub T,Wong KK,Wolchok JD
11-9949 was used in Flow cytometry/Cell sorting to set the foundation and show the importance of a combinatorial therapeutic strategy using pulsatile targeted therapy together with immunotherapy to optimally enhance tumor delay and promote long-term anti-tumor immunity.
Tue Apr 16 00:00:00 EDT 2019
Non-conventional Inhibitory CD4+Foxp3-PD-1hi T Cells as a Biomarker of Immune Checkpoint Blockade Activity.
Cancer cell
Zappasodi R,Budhu S,Hellmann MD,Postow MA,Senbabaoglu Y,Manne S,Gasmi B,Liu C,Zhong H,Li Y,Huang AC,Hirschhorn-Cymerman D,Panageas KS,Wherry EJ,Merghoub T,Wolchok JD
11-9949-82 was used in Flow Cytometry to provide evidence that mouse and human circulating and intra-tumor 4PD1hi inhibit T cell functions in a PD-1/PD-L1 dependent fashion and resemble follicular helper T cell (TFH)-like cells.
Mon Jun 11 00:00:00 EDT 2018
The Kinase Complex mTOR Complex 2 Promotes the Follicular Migration and Functional Maturation of Differentiated Follicular Helper CD4+ T Cells During Viral Infection.
Frontiers in immunology
Hao Y,Wang Y,Liu X,Yang X,Wang P,Tian Q,Bai Q,Chen X,Li Z,Wu J,Xie Z,Zhou X,Zhou Y,Yin Z,Wu Y,Ye L
Published figure using CD278 (ICOS) monoclonal antibody (Product # 11-9949-82) in Flow Cytometry
Tue Jul 30 00:00:00 EDT 2019
Differential cell-intrinsic regulations of germinal center B and T cells by miR-146a and miR-146b.
Nature communications
Cho S,Lee HM,Yu IS,Choi YS,Huang HY,Hashemifar SS,Lin LL,Chen MC,Afanasiev ND,Khan AA,Lin SW,Rudensky AY,Crotty S,Lu LF
Published figure using CD278 (ICOS) monoclonal antibody (Product # 11-9949-82) in Flow Cytometry
Mon Jul 16 00:00:00 EDT 2018
MicroRNA-155 Protects Group 2 Innate Lymphoid Cells From Apoptosis to Promote Type-2 Immunity.
Frontiers in immunology
Knolle MD,Chin SB,Rana BMJ,Englezakis A,Nakagawa R,Fallon PG,Git A,McKenzie ANJ
Published figure using CD278 (ICOS) monoclonal antibody (Product # 11-9949-82) in Flow Cytometry
Mon Sep 30 00:00:00 EDT 2019
Regulatory T Cells in Skin Facilitate Epithelial Stem Cell Differentiation.
Cell
Ali N,Zirak B,Rodriguez RS,Pauli ML,Truong HA,Lai K,Ahn R,Corbin K,Lowe MM,Scharschmidt TC,Taravati K,Tan MR,Ricardo-Gonzalez RR,Nosbaum A,Bertolini M,Liao W,Nestle FO,Paus R,Cotsarelis G,Abbas AK,Rosenblum MD
11-9949 was used in Flow cytometry/Cell sorting to study the role of regulatory T cells in hair follicles and hair follicle stem cell biology.
Thu Jun 01 00:00:00 EDT 2017
Commensal Microbes and Hair Follicle Morphogenesis Coordinately Drive Treg Migration into Neonatal Skin.
Cell host & microbe
Scharschmidt TC,Vasquez KS,Pauli ML,Leitner EG,Chu K,Truong HA,Lowe MM,Sanchez Rodriguez R,Ali N,Laszik ZG,Sonnenburg JL,Millar SE,Rosenblum MD
Wed Apr 12 00:00:00 EDT 2017
Interleukin-21 promotes germinal center reaction by skewing the follicular regulatory T cell to follicular helper T cell balance in autoimmune BXD2 mice.
Arthritis & rheumatology (Hoboken, N.J.)
Ding Y,Li J,Yang P,Luo B,Wu Q,Zajac AJ,Wildner O,Hsu HC,Mountz JD
11-9949 was used in Flow cytometry/Cell sorting to indicate that high levels of IL-21 selectively enhance Tfh cell differentiation but inhibit Tfr cell commitment and the suppressive function of Tfr cells on Tfh cells and B cells.
Mon Sep 01 00:00:00 EDT 2014
Cytokine therapy with interleukin-2/anti-interleukin-2 monoclonal antibody complexes expands CD4+CD25+Foxp3+ regulatory T cells and attenuates development and progression of atherosclerosis.
Circulation
Dinh TN,Kyaw TS,Kanellakis P,To K,Tipping P,Toh BH,Bobik A,Agrotis A
11-9949 was used in Flow cytometry/Cell sorting to examine the capacity of a modified IL-2/anti-IL-2 monoclonal antibody treatment to expand Tregs and attenuate atherosclerosis in mice.
Tue Sep 04 00:00:00 EDT 2012
Murine CD4⁺CD25⁻ cells activated in vitro with PMA/ionomycin and anti-CD3 acquire regulatory function and ameliorate experimental colitis in vivo.
BMC gastroenterology
Majowicz A,van der Marel S,te Velde AA,Meijer SL,Petry H,van Deventer SJ,Ferreira V
11-9949 was used in Flow cytometry/Cell sorting to identify a new method to generate in vitro iTreg cells (TregPMA cells) with demonstrated functionality in vivo.
Mon Dec 03 00:00:00 EST 2012
Transient blockade of the inducible costimulator pathway generates long-term tolerance to factor VIII after nonviral gene transfer into hemophilia A mice.
Blood
Peng B,Ye P,Blazar BR,Freeman GJ,Rawlings DJ,Ochs HD,Miao CH
11-9949 was used in Flow cytometry/Cell sorting to study the formation of inhibitory antibodies during treatment of hemophilia, showing that transient blockade of the inducible costimulator pathway generates long-term tolerance to factor VIII.
Mon Sep 01 00:00:00 EDT 2008