Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AEYLRDMEA NLLPGFLRLQ ELADRNVTVL FLSEIVWEKF RPNTGCFEPF VLYFPDYSIG NLQKILSHDH PPEYSADFYA AYINILLGVF YTVCRDLKEL RHLAVLNFPK YCEPVVKGEA SERDTRKLWR NIEPHLKKAM QTVYLREISS SQWEKLQKDD TDPGQLKGLS AHTHVELPYY SKFILIAAYL ASYNPARTDK RFFLKHHGKI KKTNFLKKHE KTSNHLLGPK PFPLDRLLAI LYSIVDSRVA PTANIFSQIT SLVTLQLLTL VGHDDQLDGP KYKCTVSLDF IRAIARTVNF DIIKYLYDFL (127-435 aa encoded by BC023652)
Component of the origin recognition complex (ORC) that binds origins of replication. DNA-binding is ATP-dependent. The specific DNA sequences that define origins of replication have not been identified yet. ORC is required to assemble the pre- replication complex necessary to initiate DNA replication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: origin recognition complex 5; Origin recognition complex subunit 5; origin recognition complex, subunit 5 homolog; origin recognition complex, subunit 5-like; protein phosphatase 1, regulatory subunit 117
Gene Aliases: AL033327; MmORC5; ORC5; ORC5L; ORC5P; ORC5T; PPP1R117
UniProt ID: (Human) O43913, (Mouse) Q9WUV0
Entrez Gene ID: (Human) 5001, (Mouse) 26429, (Rat) 362304
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support