Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ITPVGSLLA LMAHTILFLK LFSYRDVNSW CRRARAKAAS AGKKASSAAA PHTVSYPDNL TYRDLYYFLF APTLCYELNF PRSPRIRKRF LLRRILEMLF FTQLQVGLIQ QWMVPTIQNS MKPFKDMDYS RIIERLLKLA VPNHLIWLIF FYWLFHSCLN AVAELMQFGD REFYRDWWNS ESVTYFWQNW NIPVHKWCIR HFYKPMLRRG SSKWMARTGV FLASAFFHEY LVSVPLRMFR LWAFTGMMAQ IPLAWFVGRF FQGNYGNAAV WLSLIIGQPI AVLMYVHDYY VLNYEAPAAE A (189-488 aa encoded by BC015762)
The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACAT related gene product 1; ACAT-related gene product 1; Acyl coenzyme A; acyl coenzyme A:cholesterol acyltransferase related gene 1; Acyl-CoA retinol O-fatty-acyltransferase; acyl-CoA:diacylglycerol acyltransferase; acyl-coenzyme A:diacylglycerol acyltransferase 1; ARAT; diacylglycerol acyltransferase; diacylglycerol acyltransferase 1; Diacylglycerol O-acyltransferase 1; Diglyceride acyltransferase; EC 2.3.1.20; retinol O-fatty-acyltransferase
Gene Aliases: AGRP1; ARAT; ARGP1; C75990; D15Ertd23e; DGAT; DGAT1; DIAR7
UniProt ID: (Human) O75907, (Rat) Q9ERM3, (Mouse) Q9Z2A7
Entrez Gene ID: (Human) 8694, (Rat) 84497, (Mouse) 13350
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support