Product References
Elevated expression of the rhythm gene NFIL3 promotes the progression of TNBC by activating NF-κB signaling through suppression of NFKBIA transcription.
Journal of experimental & clinical cancer research : CR
Yang W,Li J,Zhang M,Yu H,Zhuang Y,Zhao L,Ren L,Gong J,Bi H,Zeng L,Xue Y,Yang J,Zhao Y,Wang S,Gao S,Fu Z,Li D,Zhang J,Wang T,Shan M,Tang B,Li X
Fri Feb 18 00:00:00 EST 2022
Elevated expression of the rhythm gene NFIL3 promotes the progression of TNBC by activating NF-κB signaling through suppression of NFKBIA transcription.
Journal of experimental & clinical cancer research : CR
Yang W,Li J,Zhang M,Yu H,Zhuang Y,Zhao L,Ren L,Gong J,Bi H,Zeng L,Xue Y,Yang J,Zhao Y,Wang S,Gao S,Fu Z,Li D,Zhang J,Wang T,Shan M,Tang B,Li X
Fri Feb 18 00:00:00 EST 2022
Elevated expression of the rhythm gene NFIL3 promotes the progression of TNBC by activating NF-κB signaling through suppression of NFKBIA transcription.
Journal of experimental & clinical cancer research : CR
Yang W,Li J,Zhang M,Yu H,Zhuang Y,Zhao L,Ren L,Gong J,Bi H,Zeng L,Xue Y,Yang J,Zhao Y,Wang S,Gao S,Fu Z,Li D,Zhang J,Wang T,Shan M,Tang B,Li X
Fri Feb 18 00:00:00 EST 2022
Tangeretin prevents cognitive deficit in delirium through activating RORα/γ-E4BP4 axis in mice.
Biochemical pharmacology
Chen M,Xiao Y,Zhang F,Du J,Zhang L,Li Y,Lu D,Wang Z,Wu B
Tue Nov 01 00:00:00 EDT 2022
NFIL3 Facilitates Neutrophil Autophagy, Neutrophil Extracellular Trap Formation and Inflammation During Gout via REDD1-Dependent mTOR Inactivation.
Frontiers in medicine
Tang H,Tan C,Cao X,Liu Y,Zhao H,Liu Y,Zhao Y
Mon Apr 17 00:00:00 EDT 2023
NFIL3 Facilitates Neutrophil Autophagy, Neutrophil Extracellular Trap Formation and Inflammation During Gout via REDD1-Dependent mTOR Inactivation.
Frontiers in medicine
Tang H,Tan C,Cao X,Liu Y,Zhao H,Liu Y,Zhao Y
Mon Apr 17 00:00:00 EDT 2023
Identification of rhythmic human CYPs and their circadian regulators using synchronized hepatoma cells.
Xenobiotica; the fate of foreign compounds in biological systems
Chen M,Zhou C,Zhang T,Wu B
Tue Sep 01 00:00:00 EDT 2020
The Circadian Clock Gene Bmal1 Controls Intestinal Exporter MRP2 and Drug Disposition.
Theranostics
Yu F,Zhang T,Zhou C,Xu H,Guo L,Chen M,Wu B
Thu Jun 18 00:00:00 EDT 2020
NFIL3 Acts as a Nuclear Factor to Increase Osteosarcoma Progression.
BioMed research international
Xu J,Xu G,Zhang T,Chen T,Zhao W,Wang G
Wed May 13 00:00:00 EDT 2020
The transcription factor E4bp4 regulates the expression and activity of Cyp3a11 in mice.
Biochemical pharmacology
Tong Y,Zeng P,Zhang T,Zhao M,Yu P,Wu B
Wed May 01 00:00:00 EDT 2019
The Molecular Mechanism Regulating Diurnal Rhythm of Flavin-Containing Monooxygenase 5 in Mouse Liver.
Drug metabolism and disposition: the biological fate of chemicals
Chen M,Guan B,Xu H,Yu F,Zhang T,Wu B
Fri Nov 01 00:00:00 EDT 2019
The Molecular Mechanism Regulating Diurnal Rhythm of Flavin-Containing Monooxygenase 5 in Mouse Liver.
Drug metabolism and disposition: the biological fate of chemicals
Chen M,Guan B,Xu H,Yu F,Zhang T,Wu B
Fri Nov 01 00:00:00 EDT 2019
Retigabine ameliorates acute stress-induced impairment of spatial memory retrieval through regulating USP2 signaling pathways in hippocampal CA1 area.
Neuropharmacology
Li C,Zhang J,Xu H,Chang M,Lv C,Xue W,Song Z,Zhang L,Zhang X,Tian X
Fri Jun 01 00:00:00 EDT 2018
Retigabine ameliorates acute stress-induced impairment of spatial memory retrieval through regulating USP2 signaling pathways in hippocampal CA1 area.
Neuropharmacology
Li C,Zhang J,Xu H,Chang M,Lv C,Xue W,Song Z,Zhang L,Zhang X,Tian X
Fri Jun 01 00:00:00 EDT 2018
Mislocated FUS is sufficient for gain-of-toxic-function amyotrophic lateral sclerosis phenotypes in mice.
Brain : a journal of neurology
Shiihashi G,Ito D,Yagi T,Nihei Y,Ebine T,Suzuki N
Thu Sep 01 00:00:00 EDT 2016
Mislocated FUS is sufficient for gain-of-toxic-function amyotrophic lateral sclerosis phenotypes in mice.
Brain : a journal of neurology
Shiihashi G,Ito D,Yagi T,Nihei Y,Ebine T,Suzuki N
Thu Sep 01 00:00:00 EDT 2016