Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRPCIWIHV HLKPPCRLVE LLPFSSALQG LSHLSLGTTL PVILPERNEE QNLQELSHNA DKYQMGDCCK EEIDDSIFY (1-78 aa encoded by BC020692)
May act as a tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TRalphaA1-2
Gene Aliases: BCMS; BCMS1; DLB1; DLEU2; LEU1; LEU2; LINC00021; NCRNA00021; XTP6
Entrez Gene ID: (Human) 10301
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support