Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEAVAKFDF TASGEDELSF HTGDVLKILS NQEEWFKAEL GSQEGYVPKN FIDIQFPKWF HEGLSRHQAE NLLMGKEVGF FIIRASQSSP GDFSISVRHE DDVQHFKVMR DNKGNYFLWT EKFPSLNKLV DYYRTNSISR QKQIFLRDRT REDQGHRGNS LDRRSQGGPH LSGAVGEEIR PSMNRKLSDH PPTLPLQQHQ HQPQPPQYAP APQQLQQPPQ QRYLQHHHFH QERRGGSLDI NDGHCGTGLG SEMNAALMHR RHTDPVQLQA AGRVRWARAL YDFEALEDDE LGFHSGEVVE VLDSSNPSWW TGRLHNKLGL FPANYVAPMT R (1-330 aa encoded by BC026002)
This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adapter protein GRID; GADS protein; GRB-2-like protein; GRB-2-related monocytic adapter protein; GRB2-related adapter protein 2; Grb2-related adaptor downstream of Sch; GRB2-related adaptor protein 2; GRB2-related protein with insert domain; GRB2L; GRBLG; GRBX; Grf-40; Grf40 adapter protein; Growth factor receptor-binding protein; growth factor receptor-bound protein 2-related adaptor protein 2; Hematopoietic cell-associated adapter protein GrpL; Hematopoietic cell-associated adaptor protein GrpL; MONA; monocytic adapter; monocytic adaptor; P38; Protein GADS; RP3-370M22.1; SH3-SH2-SH3 adapter Mona; SH3-SH2-SH3 adaptor molecule
Gene Aliases: GADS; GRAP-2; GRAP2; GRB2L; GRBLG; GrbX; Grf40; GRID; GRPL; Mona; P38
UniProt ID: (Human) O75791, (Mouse) O89100
Entrez Gene ID: (Human) 9402, (Rat) 366962, (Mouse) 17444
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support