Product References
IFNAR blockade synergizes with oncolytic VSV to prevent virus-mediated PD-L1 expression and promote antitumor T cell activity.
Molecular therapy oncolytics
El-Sayes N,Walsh S,Vito A,Reihani A,Ask K,Wan Y,Mossman K
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunohistochemistry
Thu Jun 16 00:00:00 EDT 2022
Modification of PLAC8 by UFM1 affects tumorous proliferation and immune response by impacting PD-L1 levels in triple-negative breast cancer.
Journal for immunotherapy of cancer
Mao M,Chen Y,Yang J,Cheng Y,Xu L,Ji F,Zhou J,Zhang X,Li Z,Chen C,Ju S,Zhang J,Wang L
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
Heterogeneity of tyrosine-based melanin anabolism regulates pulmonary and cerebral organotropic colonization microenvironment of melanoma cells.
Theranostics
Wang X,Chen Y,Lan B,Wang Y,Lin W,Jiang X,Ye J,Shang B,Feng C,Liu J,Zhai J,Xu M,Li Q,Lin L,Hu M,Zheng F,Chen L,Shao C,Wang Y,Shi Y
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Thu Apr 21 00:00:00 EDT 2022
CD4+ CTLs Act as a Key Effector Population for Allograft Rejection of MSCs in a Donor MHC-II Dependent Manner in Injured Liver.
Aging and disease
Shen S,Li Y,Jin M,Fan D,Pan R,Lin A,Chen Y,Xiang L,Zhao RC,Shao J
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
5-Azacytidine-Mediated Modulation of the Immune Microenvironment in Murine Acute Myeloid Leukemia.
Cancers
Ebelt ND,Manuel ER
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Sun Dec 25 00:00:00 EST 2022
Scoring model based on the signature of non-m6A-related neoantigen-coding lncRNAs assists in immune microenvironment analysis and TCR-neoantigen pair selection in gliomas.
Journal of translational medicine
Zhao W,Wu Y,Zhao F,Xue Z,Liu W,Cao Z,Zhao Z,Huang B,Han M,Li X
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunohistochemistry
Sat Oct 29 00:00:00 EDT 2022
Isotope tracing reveals distinct substrate preference in murine melanoma subtypes with differing anti-tumor immunity.
Cancer & metabolism
Zhang X,Halberstam AA,Zhu W,Leitner BP,Thakral D,Bosenberg MW,Perry RJ
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2022
Multiscale imaging of therapeutic anti-PD-L1 antibody localization using molecularly defined imaging agents.
Journal of nanobiotechnology
Hagemans IM,Wierstra PJ,Steuten K,Molkenboer-Kuenen JDM,van Dalen D,Ter Beest M,van der Schoot JMS,Ilina O,Gotthardt M,Figdor CG,Scheeren FA,Heskamp S,Verdoes M
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Wed Feb 02 00:00:00 EST 2022
Radiation-induced abscopal effect and its enhancement by programmed cell death 1 blockade in the hepatocellular carcinoma: A murine model study.
Clinical and molecular hepatology
Yoo GS,Ahn WG,Kim SY,Kang W,Choi C,Park HC
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Fri Jan 01 00:00:00 EST 2021
Oncolytic herpes simplex virus HF10 (canerpaturev) promotes accumulation of CD8+ PD-1- tumor-infiltrating T cells in PD-L1-enriched tumor microenvironment.
International journal of cancer
Eissa IR,Mukoyama N,Abdelmoneim M,Naoe Y,Matsumura S,Bustos-Villalobos I,Ichinose T,Miyajima N,Morimoto D,Tanaka M,Fujimoto Y,Sone M,Kodera Y,Kasuya H
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Thu Jul 01 00:00:00 EDT 2021
Preclinical characterization of bemarituzumab, an anti-FGFR2b antibody for the treatment of cancer.
mAbs
Xiang H,Chan AG,Ahene A,Bellovin DI,Deng R,Hsu AW,Jeffry U,Palencia S,Powers J,Zanghi J,Collins H
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunocytochemistry
Mon Apr 04 00:00:00 EDT 2022
Collagenase-Expressing Salmonella Targets Major Collagens in Pancreatic Cancer Leading to Reductions in Immunosuppressive Subsets and Tumor Growth.
Cancers
Ebelt ND,Zamloot V,Zuniga E,Passi KB,Sobocinski LJ,Young CA,Blazar BR,Manuel ER
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Fri Jul 16 00:00:00 EDT 2021
N-cadherin inhibitor creates a microenvironment that protect TILs from immune checkpoints and Treg cells.
Journal for immunotherapy of cancer
Sun Y,Jing J,Xu H,Xu L,Hu H,Tang C,Liu S,Wei Q,Duan R,Guo J,Yang L
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunohistochemistry
Mon Mar 01 00:00:00 EST 2021
N-cadherin inhibitor creates a microenvironment that protect TILs from immune checkpoints and Treg cells.
Journal for immunotherapy of cancer
Sun Y,Jing J,Xu H,Xu L,Hu H,Tang C,Liu S,Wei Q,Duan R,Guo J,Yang L
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunohistochemistry
Mon Mar 01 00:00:00 EST 2021
Ketogenic diet and ketone bodies enhance the anticancer effects of PD-1 blockade.
JCI insight
Ferrere G,Tidjani Alou M,Liu P,Goubet AG,Fidelle M,Kepp O,Durand S,Iebba V,Fluckiger A,Daillère R,Thelemaque C,Grajeda-Iglesias C,Alves Costa Silva C,Aprahamian F,Lefevre D,Zhao L,Ryffel B,Colomba E,Arnedos M,Drubay D,Rauber C,Raoult D,Asnicar F,Spector T,Segata N,Derosa L,Kroemer G,Zitvogel L
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Mon Jan 25 00:00:00 EST 2021
Progranulin induces immune escape in breast cancer via up-regulating PD-L1 expression on tumor-associated macrophages (TAMs) and promoting CD8+ T cell exclusion.
Journal of experimental & clinical cancer research : CR
Fang W,Zhou T,Shi H,Yao M,Zhang D,Qian H,Zeng Q,Wang Y,Jin F,Chai C,Chen T
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Neutralization
Mon Jan 04 00:00:00 EST 2021
USP12 downregulation orchestrates a protumourigenic microenvironment and enhances lung tumour resistance to PD-1 blockade.
Nature communications
Yang Z,Xu G,Wang B,Liu Y,Zhang L,Jing T,Tang M,Xu X,Jiao K,Xiang L,Fu Y,Tang D,Zhang X,Jin W,Zhuang G,Zhao X,Liu Y
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Wed Aug 11 00:00:00 EDT 2021
Antigen-Capturing Mesoporous Silica Nanoparticles Enhance the Radiation-Induced Abscopal Effect in Murine Hepatocellular Carcinoma Hepa1-6 Models.
Pharmaceutics
Yang K,Choi C,Cho H,Ahn WG,Kim SY,Shin SW,Kim Y,Jang T,Lee N,Park HC
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Fri Oct 29 00:00:00 EDT 2021
Downregulation of PD-L1 via amide analogues of brefelamide: Alternatives to antibody-based cancer immunotherapy.
Experimental and therapeutic medicine
Zhang J,Yamada O,Kida S,Murase S,Hattori T,Oshima Y,Kikuchi H
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
Genetic ablation of adipocyte PD-L1 reduces tumor growth but accentuates obesity-associated inflammation.
Journal for immunotherapy of cancer
Wu B,Chiang HC,Sun X,Yuan B,Mitra P,Hu Y,Curiel TJ,Li R
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2020
Cancer cell-intrinsic expression of MHC II in lung cancer cell lines is actively restricted by MEK/ERK signaling and epigenetic mechanisms.
Journal for immunotherapy of cancer
Neuwelt AJ,Kimball AK,Johnson AM,Arnold BW,Bullock BL,Kaspar RE,Kleczko EK,Kwak JW,Wu MH,Heasley LE,Doebele RC,Li HY,Nemenoff RA,Clambey ET
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2020
CD279 mediates the homeostasis and survival of regulatory T cells by enhancing T cell and macrophage interactions.
FEBS open bio
Liu Y,Gao Y,Hao H,Hou T
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Functional assay
Mon Jun 01 00:00:00 EDT 2020
CD279 mediates the homeostasis and survival of regulatory T cells by enhancing T cell and macrophage interactions.
FEBS open bio
Liu Y,Gao Y,Hao H,Hou T
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Functional assay
Mon Jun 01 00:00:00 EDT 2020
Nuclear imaging-guided PD-L1 blockade therapy increases effectiveness of cancer immunotherapy.
Journal for immunotherapy of cancer
Gao H,Wu Y,Shi J,Zhang X,Liu T,Hu B,Jia B,Wan Y,Liu Z,Wang F
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2020
Epigenetically modulated FOXM1 suppresses dendritic cell maturation in pancreatic cancer and colon cancer.
Molecular oncology
Zhou Z,Chen H,Xie R,Wang H,Li S,Xu Q,Xu N,Cheng Q,Qian Y,Huang R,Shao Z,Xiang M
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Mon Apr 01 00:00:00 EDT 2019
Inhibition of ATM reverses EMT and decreases metastatic potential of cisplatin-resistant lung cancer cells through JAK/STAT3/PD-L1 pathway.
Journal of experimental & clinical cancer research : CR
Shen M,Xu Z,Xu W,Jiang K,Zhang F,Ding Q,Xu Z,Chen Y
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Neutralization
Mon Apr 08 00:00:00 EDT 2019
Persistence of Integrase-Deficient Lentiviral Vectors Correlates with the Induction of STING-Independent CD8+ T Cell Responses.
Cell reports
Cousin C,Oberkampf M,Felix T,Rosenbaum P,Weil R,Fabrega S,Morante V,Negri D,Cara A,Dadaglio G,Leclerc C
13-5982 was used in Flow cytometry/Cell sorting to demonstrate that integrase-deficient lentiviral vectors can trigger persistent adaptive response by the mediation of a weak and transient innate response alongside the persistence of the vector in tissues.
Tue Jan 29 00:00:00 EST 2019
PD-L1 in tumor microenvironment mediates resistance to oncolytic immunotherapy.
The Journal of clinical investigation
Zamarin D,Ricca JM,Sadekova S,Oseledchyk A,Yu Y,Blumenschein WM,Wong J,Gigoux M,Merghoub T,Wolchok JD
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Mon Apr 02 00:00:00 EDT 2018
ATM‑JAK‑PD‑L1 signaling pathway inhibition decreases EMT and metastasis of androgen‑independent prostate cancer.
Molecular medicine reports
Zhang L,Xu LJ,Zhu J,Li J,Xue BX,Gao J,Sun CY,Zang YC,Zhou YB,Yang DR,Shan YX
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunofluorescence
Tue May 01 00:00:00 EDT 2018
Oncofetal gene SALL4 reactivation by hepatitis B virus counteracts miR-200c in PD-L1-induced T cell exhaustion.
Nature communications
Sun C,Lan P,Han Q,Huang M,Zhang Z,Xu G,Song J,Wang J,Wei H,Zhang J,Sun R,Zhang C,Tian Z
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Wed Mar 28 00:00:00 EDT 2018
Oncogenic RAS Signaling Promotes Tumor Immunoresistance by Stabilizing PD-L1 mRNA.
Immunity
Coelho MA,de Carné Trécesson S,Rana S,Zecchin D,Moore C,Molina-Arcas M,East P,Spencer-Dene B,Nye E,Barnouin K,Snijders AP,Lai WS,Blackshear PJ,Downward J
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Tue Dec 19 00:00:00 EST 2017
Neuronal IFN-beta-induced PI3K/Akt-FoxA1 signalling is essential for generation of FoxA1+Treg cells.
Nature communications
Liu Y,Marin A,Ejlerskov P,Rasmussen LM,Prinz M,Issazadeh-Navikas S
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Western Blot
Mon Apr 24 00:00:00 EDT 2017
Spatiotemporally restricted arenavirus replication induces immune surveillance and type I interferon-dependent tumour regression.
Nature communications
Kalkavan H,Sharma P,Kasper S,Helfrich I,Pandyra AA,Gassa A,Virchow I,Flatz L,Brandenburg T,Namineni S,Heikenwalder M,Höchst B,Knolle PA,Wollmann G,von Laer D,Drexler I,Rathbun J,Cannon PM,Scheu S,Bauer J,Chauhan J,Häussinger D,Willimsky G,Löhning M,Schadendorf D,Brandau S,Schuler M,Lang PA,Lang KS
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunofluorescence
Wed Mar 01 00:00:00 EST 2017
Rational combination of oncolytic vaccinia virus and PD-L1 blockade works synergistically to enhance therapeutic efficacy.
Nature communications
Liu Z,Ravindranathan R,Kalinski P,Guo ZS,Bartlett DL
13-5982 was used in Flow cytometry/Cell sorting to demonstrate in colon and ovarian cancer models that an oncolytic vaccinia virus attracts effector T cells and induces PD-L1 expression on both cancer and immune cells in the tumour.
Mon Mar 27 00:00:00 EDT 2017
Maternal-Derived Hepatitis B Virus e Antigen Alters Macrophage Function in Offspring to Drive Viral Persistence after Vertical Transmission.
Immunity
Tian Y,Kuo CF,Akbari O,Ou JH
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Tue May 17 00:00:00 EDT 2016
Preclinical Development of Ipilimumab and Nivolumab Combination Immunotherapy: Mouse Tumor Models, In Vitro Functional Studies, and Cynomolgus Macaque Toxicology.
PloS one
Selby MJ,Engelhardt JJ,Johnston RJ,Lu LS,Han M,Thudium K,Yao D,Quigley M,Valle J,Wang C,Chen B,Cardarelli PM,Blanset D,Korman AJ
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Immunohistochemistry
Fri Aug 25 00:00:00 EDT 2017
Exploring the induction of preproinsulin-specific Foxp3(+) CD4(+) Treg cells that inhibit CD8(+) T cell-mediated autoimmune diabetes by DNA vaccination.
Scientific reports
Stifter K,Schuster C,Schlosser M,Boehm BO,Schirmbeck R
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Neutralization
Mon Jul 11 00:00:00 EDT 2016
Th2 cell-intrinsic hypo-responsiveness determines susceptibility to helminth infection.
PLoS pathogens
van der Werf N,Redpath SA,Azuma M,Yagita H,Taylor MD
13-5982 was used in Flow cytometry/Cell sorting to demonstrate that during chronic Litomosoides sigmodontis infection, CD4(+) Th2 cells are conditioned towards an intrinsically hypo-responsive phenotype.
Fri Mar 01 00:00:00 EST 2013
IL-10 controls dendritic cell-induced T-cell reactivation in the skin to limit contact hypersensitivity.
The Journal of allergy and clinical immunology
Girard-Madoux MJ,Kel JM,Reizis B,Clausen BE
13-5982 was used in Flow cytometry/Cell sorting to demonstrate that IL-10 signaling in dendritic cells is critical in preventing an exaggerated effector T-cell response in the skin.
Sun Jan 01 00:00:00 EST 2012
Ex vivo expanded hematopoietic stem cells overcome the MHC barrier in allogeneic transplantation.
Cell stem cell
Zheng J,Umikawa M,Zhang S,Huynh H,Silvany R,Chen BP,Chen L,Zhang CC
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Neutralization
Fri Aug 05 00:00:00 EDT 2011
Ex vivo expanded hematopoietic stem cells overcome the MHC barrier in allogeneic transplantation.
Cell stem cell
Zheng J,Umikawa M,Zhang S,Huynh H,Silvany R,Chen BP,Chen L,Zhang CC
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Neutralization
Fri Aug 05 00:00:00 EDT 2011
Farnesyltransferase inhibitor FTI-277 reduces mortality of septic mice along with improved bacterial clearance.
The Journal of pharmacology and experimental therapeutics
Yang W,Yamada M,Tamura Y,Chang K,Mao J,Zou L,Feng Y,Kida K,Scherrer-Crosbie M,Chao W,Ichinose F,Yu YM,Fischman AJ,Tompkins RG,Yao S,Kaneki M
Published figure using CD274 (PD-L1, B7-H1) monoclonal antibody (Product # 13-5982-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2011
Pre-existing autoimmunity determines type 1 diabetes outcome after Flt3-ligand treatment.
Journal of autoimmunity
Van Belle TL,Juntti T,Liao J,von Herrath MG
13-5982 was used in Flow cytometry/Cell sorting to investigate how Flt3 stimulation actually accelerates autoimmunity when autoreactive CD8 T cells are detectable in blood prior to treatment.
Tue Jun 01 00:00:00 EDT 2010
A critical role of IFNgamma in priming MSC-mediated suppression of T cell proliferation through up-regulation of B7-H1.
Cell research
Sheng H,Wang Y,Jin Y,Zhang Q,Zhang Y,Wang L,Shen B,Yin S,Liu W,Cui L,Li N
Fri Aug 01 00:00:00 EDT 2008