Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTTEKSLVT EAENSQHQQK EEGEEAINSG QQEPQQEESC QTAAEGDNWC EQKLKASNGD TPTHEDLTKN KERTSESRGL SRLFSSFLKR PKSQVSEEEG KEVESDKEKG EGGQKEIEFG TSLDEEIILK APIAAPEPEL KTDPSLDLHS LSSAETQPAQ EELREDPDFE IKEGEGLEEC SKIEVKEESP QSKAETELKA SQKPIRKHRN MHCKVSLLDD TVYECVVEKH AKGQDLLKRV CEHLNLLEED YFGLAIWDNA TSKTWLDSAK EIKKQVRGVP WNFTFNVKFY PPDPAQLTED (1-299 aa encoded by BC039079)
Elliptocytosis is a hematologic disorder characterized by elliptically shaped erythrocytes and a variable degree of hemolytic anemia. Inherited as an autosomal dominant, elliptocytosis results from mutation in any one of several genes encoding proteins of the red cell membrane skeleton. The form discussed here is the one found in the 1950s to be linked to Rh blood group and more recently shown to be caused by a defect in protein 4. 1. 'Rh-unlinked' forms of elliptocytosis are caused by mutation in the alpha-spectrin gene, the beta-spectrin gene, or the band 3 gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 4.1R; Band 4.1; elliptocytosis 1, RH-linked; EPB4.1; Erythrocyte membrane protein band 4.1; erythrocyte protein band 4.1; erythrocyte surface protein band 4.1; P4.1; Protein 4.1; Protein 4.1R
Gene Aliases: 4.1R; AI415518; D4Ertd442e; E41P; EL1; Elp-1; Elp1; Epb4.1; EPB41; HE; Kiaa4056; mKIAA4056; RGD1564762
UniProt ID: (Human) P11171, (Mouse) P48193
Entrez Gene ID: (Human) 2035, (Mouse) 269587, (Rat) 313052
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support