Product References
Microbiota-Derived Metabolites Suppress Arthritis by Amplifying Aryl-Hydrocarbon Receptor Activation in Regulatory B Cells.
Cell metabolism
Rosser EC,Piper CJM,Matei DE,Blair PA,Rendeiro AF,Orford M,Alber DG,Krausgruber T,Catalan D,Klein N,Manson JJ,Drozdov I,Bock C,Wedderburn LR,Eaton S,Mauri C
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Immunohistochemistry
Tue Apr 07 00:00:00 EDT 2020
Microbiota-Derived Metabolites Suppress Arthritis by Amplifying Aryl-Hydrocarbon Receptor Activation in Regulatory B Cells.
Cell metabolism
Rosser EC,Piper CJM,Matei DE,Blair PA,Rendeiro AF,Orford M,Alber DG,Krausgruber T,Catalan D,Klein N,Manson JJ,Drozdov I,Bock C,Wedderburn LR,Eaton S,Mauri C
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Immunohistochemistry
Tue Apr 07 00:00:00 EDT 2020
αv Integrins regulate germinal center B cell responses through noncanonical autophagy.
The Journal of clinical investigation
Raso F,Sagadiev S,Du S,Gage E,Arkatkar T,Metzler G,Stuart LM,Orr MT,Rawlings DJ,Jackson SW,Lacy-Hulbert A,Acharya M
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Fri Aug 31 00:00:00 EDT 2018
Fas ligand promotes an inducible TLR-dependent model of cutaneous lupus-like inflammation.
The Journal of clinical investigation
Mande P,Zirak B,Ko WC,Taravati K,Bride KL,Brodeur TY,Deng A,Dresser K,Jiang Z,Ettinger R,Fitzgerald KA,Rosenblum MD,Harris JE,Marshak-Rothstein A
14-5902 was used in Flow cytometry/Cell sorting to provide a relevant system for exploring the pathophysiology of cutaneous lupus erythematosus (CLE) as well as the negative regulatory role of TLR9.
Mon Jul 02 00:00:00 EDT 2018
Cleavage of TL1A Differentially Regulates Its Effects on Innate and Adaptive Immune Cells.
Journal of immunology (Baltimore, Md. : 1950)
Ferdinand JR,Richard AC,Meylan F,Al-Shamkhani A,Siegel RM
14-5902 was used in Flow cytometry/Cell sorting to suggest differential roles for membrane and soluble TL1A on adaptive and innate immune cells and have implications for the consequences of blocking these two forms of TL1A.
Thu Feb 15 00:00:00 EST 2018
Anti‑IL‑39 (IL‑23p19/Ebi3) polyclonal antibodies ameliorate autoimmune symptoms in lupus‑like mice.
Molecular medicine reports
Wang X,Zhang Y,Wang Z,Liu X,Zhu G,Han G,Chen G,Hou C,Wang T,Shen B,Li Y,Xiao H,Ma N,Wang R
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Mon Jan 01 00:00:00 EST 2018
B cell activation and plasma cell differentiation are inhibited by de novo DNA methylation.
Nature communications
Barwick BG,Scharer CD,Martinez RJ,Price MJ,Wein AN,Haines RR,Bally APR,Kohlmeier JE,Boss JM
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Immunofluorescence
Tue May 15 00:00:00 EDT 2018
Act1 is a negative regulator in T and B cells via direct inhibition of STAT3.
Nature communications
Zhang CJ,Wang C,Jiang M,Gu C,Xiao J,Chen X,Martin BN,Tang F,Yamamoto E,Xian Y,Wang H,Li F,Sartor RB,Smith H,Husni ME,Shi FD,Gao J,Carman J,Dongre A,McKarns SC,Coppieters K,Jørgensen TN,Leonard WJ,Li X
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Mon Jul 16 00:00:00 EDT 2018
Peli1 negatively regulates noncanonical NF-κB signaling to restrain systemic lupus erythematosus.
Nature communications
Liu J,Huang X,Hao S,Wang Y,Liu M,Xu J,Zhang X,Yu T,Gan S,Dai D,Luo X,Lu Q,Mao C,Zhang Y,Shen N,Li B,Huang M,Zhu X,Jin J,Cheng X,Sun SC,Xiao Y
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Mon Mar 19 00:00:00 EDT 2018
Dissection of SAP-dependent and SAP-independent SLAM family signaling in NKT cell development and humoral immunity.
The Journal of experimental medicine
Chen S,Cai C,Li Z,Liu G,Wang Y,Blonska M,Li D,Du J,Lin X,Yang M,Dong Z
14-5902-82 was used in Flow Cytometry to examine NKT cell development and humoral immunity in mice completely deficient in SFR.
Wed Feb 01 00:00:00 EST 2017
Defective germinal center B-cell response and reduced arthritic pathology in microRNA-29a-deficient mice.
Cellular and molecular life sciences : CMLS
van Nieuwenhuijze A,Dooley J,Humblet-Baron S,Sreenivasan J,Koenders M,Schlenner SM,Linterman M,Liston A
14590282 was used in flow cytometry to evaluate miR-29a in B cells as a potential therapeutic target in arthritis
Thu Jun 01 00:00:00 EDT 2017
Deficiency in Calcium-Binding Protein S100A4 Impairs the Adjuvant Action of Cholera Toxin.
Frontiers in immunology
Sun JB,Holmgren J,Larena M,Terrinoni M,Fang Y,Bresnick AR,Xiang Z
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Immunofluorescence
Wed Nov 20 00:00:00 EST 2019
NCR1-deficiency diminishes the generation of protective murine cytomegalovirus antibodies by limiting follicular helper T-cell maturation.
European journal of immunology
Miletic A,Lenartic M,Popovic B,Brizic I,Trsan T,Miklic K,Mandelboim O,Krmpotic A,Jonjic S
14-5902 was used in Flow cytometry/Cell sorting to provide evidence for the role of NCR1 in antibody response to mouse cytomegalovirus infection.
Fri Sep 01 00:00:00 EDT 2017
The Ox40/Ox40 Ligand Pathway Promotes Pathogenic Th Cell Responses, Plasmablast Accumulation, and Lupus Nephritis in NZB/W F1 Mice.
Journal of immunology (Baltimore, Md. : 1950)
Sitrin J,Suto E,Wuster A,Eastham-Anderson J,Kim JM,Austin CD,Lee WP,Behrens TW
14-5902 was used in Immunohistochemistry-immunofluorescence to support the hypothesis that the Ox40/Ox40L pathway drives cellular and humoral autoimmune responses during lupus nephritis in NZB/W F1.
Tue Aug 15 00:00:00 EDT 2017
Extrafollicular CD4+ T-B interactions are sufficient for inducing autoimmune-like chronic graft-versus-host disease.
Nature communications
Deng R,Hurtz C,Song Q,Yue C,Xiao G,Yu H,Wu X,Muschen M,Forman S,Martin PJ,Zeng D
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Tue Oct 17 00:00:00 EDT 2017
Tetraspanin CD37 protects against the development of B cell lymphoma.
The Journal of clinical investigation
de Winde CM,Veenbergen S,Young KH,Xu-Monette ZY,Wang XX,Xia Y,Jabbar KJ,van den Brand M,van der Schaaf A,Elfrink S,van Houdt IS,Gijbels MJ,van de Loo FA,Bennink MB,Hebeda KM,Groenen PJ,van Krieken JH,Figdor CG,van Spriel AB
14-5902 was used in Immunohistochemistry to show that CD37 deficiency drives B cell lymphomagenesis through constitutive activation of the IL-6 pathway, highlighting a possible therapeutic intervention.
Mon Feb 01 00:00:00 EST 2016
Deficiency of base excision repair enzyme NEIL3 drives increased predisposition to autoimmunity.
The Journal of clinical investigation
Massaad MJ,Zhou J,Tsuchimoto D,Chou J,Jabara H,Janssen E,Glauzy S,Olson BG,Morbach H,Ohsumi TK,Schmitz K,Kyriacos M,Kane J,Torisu K,Nakabeppu Y,Notarangelo LD,Chouery E,Megarbane A,Kang PB,Al-Idrissi E,Aldhekri H,Meffre E,Mizui M,Tsokos GC,Manis JP,Al-Herz W,Wallace SS,Geha RS
14590282 was used in flow cytometry to identify and characterize a Nei endonuclease VIII-like 3 mutation in three siblings from a consanguineous family with autoimmunity
Tue Nov 01 00:00:00 EDT 2016
Limited clonal relatedness between gut IgA plasma cells and memory B cells after oral immunization.
Nature communications
Bemark M,Hazanov H,Strömberg A,Komban R,Holmqvist J,Köster S,Mattsson J,Sikora P,Mehr R,Lycke NY
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Immunofluorescence
Tue Sep 06 00:00:00 EDT 2016
Injectable, spontaneously assembling, inorganic scaffolds modulate immune cells in vivo and increase vaccine efficacy.
Nature biotechnology
Kim J,Li WA,Choi Y,Lewin SA,Verbeke CS,Dranoff G,Mooney DJ
14590282 was used in flow cytometry to investigate the application of intra-dermal mesoporous silica rods in immune modulation.
Thu Jan 01 00:00:00 EST 2015
LEF-1 and TCF-1 orchestrate T(FH) differentiation by regulating differentiation circuits upstream of the transcriptional repressor Bcl6.
Nature immunology
Choi YS,Gullicksrud JA,Xing S,Zeng Z,Shan Q,Li F,Love PE,Peng W,Xue HH,Crotty S
14-5902 was used in Flow cytometry/Cell sorting to report that selective loss of Lef1 or Tcf7 results in T(FH) cell defects.
Tue Sep 01 00:00:00 EDT 2015
Targeting Syk-activated B cells in murine and human chronic graft-versus-host disease.
Blood
Flynn R,Allen JL,Luznik L,MacDonald KP,Paz K,Alexander KA,Vulic A,Du J,Panoskaltsis-Mortari A,Taylor PA,Poe JC,Serody JS,Murphy WJ,Hill GR,Maillard I,Koreth J,Cutler CS,Soiffer RJ,Antin JH,Ritz J,Chao NJ,Clynes RA,Sarantopoulos S,Blazar BR
14-5902 was used in Flow cytometry/Cell sorting to further evaluate the role of the spleen tyrosine kinase in graft-versus-host disease in multiple murine models and human peripheral blood cells.
Thu Jun 25 00:00:00 EDT 2015
IDO1 suppresses inhibitor development in hemophilia A treated with factor VIII.
The Journal of clinical investigation
Matino D,Gargaro M,Santagostino E,Di Minno MN,Castaman G,Morfini M,Rocino A,Mancuso ME,Di Minno G,Coppola A,Talesa VN,Volpi C,Vacca C,Orabona C,Iannitti R,Mazzucconi MG,Santoro C,Tosti A,Chiappalupi S,Sorci G,Tagariello G,Belvini D,Radossi P,Landolfi R,Fuchs D,Boon L,Pirro M,Marchesini E,Grohmann U,Puccetti P,Iorio A,Fallarino F
14-5902 was used in Flow cytometry/Cell sorting to investigate the association between IDO1 competence and inhibitor status in haemophilia A patients.
Thu Oct 01 00:00:00 EDT 2015
Enhanced CD8 T cell responses through GITR-mediated costimulation resolve chronic viral infection.
PLoS pathogens
Pascutti MF,Geerman S,Slot E,van Gisbergen KP,Boon L,Arens R,van Lier RA,Wolkers MC,Nolte MA
14-5902 was used in Flow cytometry/Cell sorting to study resolution of chronic viral infection through GITR-mediated co-stimulation enhancing CD8 T cell responses.
Sun Mar 01 00:00:00 EST 2015
T follicular helper, but not Th1, cell differentiation in the absence of conventional dendritic cells.
Journal of immunology (Baltimore, Md. : 1950)
Dahlgren MW,Gustafsson-Hedberg T,Livingston M,Cucak H,Alsén S,Yrlid U,Johansson-Lindbom B
14-5902 was used in Flow cytometry/Cell sorting to investigate the accessory role of conventional DCs during the concurrent development of Tfh and effector Th1 cells.
Mon Jun 01 00:00:00 EDT 2015
TNF signals are dispensable for the generation of CD23+ CD21/35-high CD1d-high B cells in inflamed lymph nodes.
Cellular immunology
Kuzin II,Bouta EM,Schwarz EM,Bottaro A
14-5902 was used in Flow cytometry/Cell sorting to investigate the role of tumor necrosis factor signals in producing and maintaining B cells in inflamed lymph nodes, and their role in rheumatoid arthritis pathogenesis.
Sat Aug 01 00:00:00 EDT 2015
BILL-cadherin/cadherin-17 contributes to the survival of memory B cells.
PloS one
Funakoshi S,Shimizu T,Numata O,Ato M,Melchers F,Ohnishi K
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Wed Sep 16 00:00:00 EDT 2015
Mutation of POLB causes lupus in mice.
Cell reports
Senejani AG,Liu Y,Kidane D,Maher SE,Zeiss CJ,Park HJ,Kashgarian M,McNiff JM,Zelterman D,Bothwell AL,Sweasy JB
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Flow Cytometry
Thu Jan 16 00:00:00 EST 2014
Plasticity of Th17 cells in Peyer's patches is responsible for the induction of T cell-dependent IgA responses.
Nature immunology
Hirota K,Turner JE,Villa M,Duarte JH,Demengeot J,Steinmetz OM,Stockinger B
14-5902 was used in Flow cytometry/Cell sorting to provide evidence that TH17 cells are the crucial subset required for the production of high-affinity T cell-dependent IgA.
Mon Apr 01 00:00:00 EDT 2013
Natural IgG antibodies provide innate protection against ficolin-opsonized bacteria.
The EMBO journal
Panda S,Zhang J,Tan NS,Ho B,Ding JL
Published figure using GL7 monoclonal antibody (Product # 14-5902-82) in Immunofluorescence
Wed Nov 13 00:00:00 EST 2013
SAP modulates B cell functions in a genetic background-dependent manner.
Immunology letters
Detre C,Yigit B,Keszei M,Castro W,Magelky EM,Terhorst C
14-5902 was used in Flow cytometry/Cell sorting to investigate the mechanism by which SLAM-associated protein modulates B cell functions.
Sat Jun 01 00:00:00 EDT 2013
Peroxisome proliferator-activated receptor γ B cell-specific-deficient mice have an impaired antibody response.
Journal of immunology (Baltimore, Md. : 1950)
Ramon S,Bancos S,Thatcher TH,Murant TI,Moshkani S,Sahler JM,Bottaro A,Sime PJ,Phipps RP
14-5902 was used in Flow cytometry/Cell sorting to study how PPARγ expression in B cells is required for an efficient B cell-mediated immune response by regulating B cell differentiation and Ab production.
Thu Nov 15 00:00:00 EST 2012
The viral latency-associated nuclear antigen augments the B-cell response to antigen in vivo.
Journal of virology
Sin SH,Fakhari FD,Dittmer DP
14-5902 was used in Immunohistochemistry to examine whether the Kaposi sarcoma-associated herpesvirus latency-associated nuclear antigen provides a selective advantage to infected B cells by driving proliferation in response to antigen.
Fri Oct 01 00:00:00 EDT 2010
Human B-lymphocytes express alpha2-6-sialylated 6-sulfo-N-acetyllactosamine serving as a preferred ligand for CD22/Siglec-2.
The Journal of biological chemistry
Kimura N,Ohmori K,Miyazaki K,Izawa M,Matsuzaki Y,Yasuda Y,Takematsu H,Kozutsumi Y,Moriyama A,Kannagi R
Fri Nov 02 00:00:00 EDT 2007
TLR4 on follicular dendritic cells: an activation pathway that promotes accessory activity.
Journal of immunology (Baltimore, Md. : 1950)
El Shikh ME,El Sayed RM,Wu Y,Szakal AK,Tew JG
14-5902-82 was used in Immunohistochemistry-immunofluorescence to report expression of TLR4 on FDCs for the first time and that engagement of FDC-TLR4 activated NF-kappaB, up-regulated expression of molecules important in FDC accessory function, including Fc gammaRIIB, ICAM-1, and VCAM-1, as well as FDC accessory activity in promoting recall IgG responses.
Mon Oct 01 00:00:00 EDT 2007