Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Description: This O1 monoclonal antibody reacts with galactocerebroside and other lipids found on late oligodendrocyte precursors. O1 has been reported to react to all species tested (rat, mouse, human, chicken). Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes.
Applications Reported: This O1 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, ELISA, and immunocytochemistry.
Applications Tested: This O1 antibody has been tested by immunocytochemistry on formaldehyde-fixed, differentiated OLN93 cells and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: O1 Marker
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support