Product References
Novel HLA-B7-restricted human metapneumovirus epitopes enhance viral clearance in mice and are recognized by human CD8+ T cells.
Scientific reports
Miranda-Katz M,Erickson JJ,Lan J,Ecker A,Zhang Y,Joyce S,Williams JV
14-7313-81 was used in ELISA to enable identification of HMPV-specific CD8+ T cells in humans and help to inform future HMPV vaccine design.
Thu Oct 21 00:00:00 EDT 2021
Novel HLA-B7-restricted human metapneumovirus epitopes enhance viral clearance in mice and are recognized by human CD8+ T cells.
Scientific reports
Miranda-Katz M,Erickson JJ,Lan J,Ecker A,Zhang Y,Joyce S,Williams JV
14-7313-81 was used in ELISA to enable identification of HMPV-specific CD8+ T cells in humans and help to inform future HMPV vaccine design.
Thu Oct 21 00:00:00 EDT 2021
T-B Lymphocyte Interactions Promote Type 1 Diabetes Independently of SLAM-Associated Protein.
Journal of immunology (Baltimore, Md. : 1950)
Bonami RH,Nyhoff LE,McNitt DH,Hulbert C,Felton JL,Kendall PL,Thomas JW
14-7313-81 was used in ELISA to understand how fully developed T follicular helper cells contribute to different autoimmune processes, investigating the role of signaling lymphocytic activation molecule-associated protein in type 1 diabetes and autoantibody-mediated arthritis.
Tue Dec 15 00:00:00 EST 2020
Neurokinin-1 Receptor Signaling Is Required for Efficient Ca2+ Flux in T-Cell-Receptor-Activated T Cells.
Cell reports
Morelli AE,Sumpter TL,Rojas-Canales DM,Bandyopadhyay M,Chen Z,Tkacheva O,Shufesky WJ,Wallace CT,Watkins SC,Berger A,Paige CJ,Falo LD,Larregina AT
Published figure using IFN gamma monoclonal antibody (Product # 14-7313-81) in T-Cell Activation
Tue Mar 10 00:00:00 EDT 2020
Rab6-dependent retrograde traffic of LAT controls immune synapse formation and T cell activation.
The Journal of experimental medicine
Carpier JM,Zucchetti AE,Bataille L,Dogniaux S,Shafaq-Zadah M,Bardin S,Lucchino M,Maurin M,Joannas LD,Magalhaes JG,Johannes L,Galli T,Goud B,Hivroz C
14-7313 was used in ELISA to study the retrograde traffic of linker for activation of T cells from the plasma membrane to the Golgi-trans-Golgi network.
Mon Apr 02 00:00:00 EDT 2018
IFNγ PET Imaging as a Predictive Tool for Monitoring Response to Tumor Immunotherapy.
Cancer research
Gibson HM,McKnight BN,Malysa A,Dyson G,Wiesend WN,McCarthy CE,Reyes J,Wei WZ,Viola-Villegas NT
14-7313-81 was used in Radioimmune Assays to present a novel approach to monitor therapeutic outcomes via IFNγ-targeted positron emission tomography.
Mon Oct 01 00:00:00 EDT 2018
Serum Neuroinflammatory Disease-Induced Central Nervous System Proteins Predict Clinical Onset of Experimental Autoimmune Encephalomyelitis.
Frontiers in immunology
Raphael I,Webb J,Gomez-Rivera F,Chase Huizar CA,Gupta R,Arulanandam BP,Wang Y,Haskins WE,Forsthuber TG
14-7313 was used in an ELISpot assay to determine if changes in serum neuroinflammatory disease-induced CNS proteins are detectable prior to clinical onset of experimental autoimmune encephalomyelitis and, therefore, enable prediction of disease onset.
Wed Nov 20 00:00:00 EST 2019
Double conjugation strategy to incorporate lipid adjuvants into multiantigenic vaccines.
Chemical science
Hussein WM,Liu TY,Maruthayanar P,Mukaida S,Moyle PM,Wells JW,Toth I,Skwarczynski M
14-7313 was used in an ELISpot assay to develop a double conjugation strategy for the synthesis of self-adjuvanting, branched, multiantigenic vaccine candidates.
Tue Mar 01 00:00:00 EST 2016
Nonclassical MHC Ib-restricted CD8+ T Cells Recognize Mycobacterium tuberculosis-Derived Protein Antigens and Contribute to Protection Against Infection.
PLoS pathogens
Shang S,Siddiqui S,Bian Y,Zhao J,Wang CR
14-7313 was used in an ELISpot assay to investigate the relative contribution of various MHC Ib-restricted T cell populations to anti-mycobacterial immunity.
Wed Jun 01 00:00:00 EDT 2016
Altered Glycosylation Patterns Increase Immunogenicity of a Subunit Hepatitis C Virus Vaccine, Inducing Neutralizing Antibodies Which Confer Protection in Mice.
Journal of virology
Li D,von Schaewen M,Wang X,Tao W,Zhang Y,Li L,Heller B,Hrebikova G,Deng Q,Ploss A,Zhong J,Huang Z
14-7313 was used in an ELISpot assay to identify a promising broadly protective HCV vaccine candidate that should be considered for further preclinical and clinical development.
Thu Dec 01 00:00:00 EST 2016
Mucosal Langerhans Cells Promote Differentiation of Th17 Cells in a Murine Model of Periodontitis but Are Not Required for Porphyromonas gingivalis-Driven Alveolar Bone Destruction.
Journal of immunology (Baltimore, Md. : 1950)
Bittner-Eddy PD,Fischer LA,Kaplan DH,Thieu K,Costalonga M
14-7313 was used in an ELISpot assay to indicate that both Langerhans cells and Th17 cells are redundant in contributing to alveolar bone destruction in a murine model of periodontitis.
Mon Aug 15 00:00:00 EDT 2016
Targeted Vaccination against Human α-Lactalbumin for Immunotherapy and Primary Immunoprevention of Triple Negative Breast Cancer.
Cancers
Tuohy VK,Jaini R,Johnson JM,Loya MG,Wilk D,Downs-Kelly E,Mazumder S
14-7313 was used in an ELISpot assay to investigate the use of retired self-proteins in the treatment of cancer, showing that vaccination against α-Lactalbumin can be used against triple negative breast cancers.
Thu Jun 16 00:00:00 EDT 2016
Macrophage migration inhibitory factor promotes resistance to glucocorticoid treatment in EAE.
Neurology(R) neuroimmunology & neuroinflammation
Ji N,Kovalovsky A,Fingerle-Rowson G,Guentzel MN,Forsthuber TG
14-7313 was used in an ELISpot assay to establish macrophage migration inhibitory factor as a key molecule underlying resistance to anti-inflammatory treatment for multiple sclerosis.
Thu Oct 01 00:00:00 EDT 2015
Blocking stroke-induced immunodeficiency increases CNS antigen-specific autoreactivity but does not worsen functional outcome after experimental stroke.
The Journal of neuroscience : the official journal of the Society for Neuroscience
Römer C,Engel O,Winek K,Hochmeister S,Zhang T,Royl G,Klehmet J,Dirnagl U,Meisel C,Meisel A
Wed May 20 00:00:00 EDT 2015
Natural killer T cells are essential for the development of contact hypersensitivity in BALB/c mice.
The Journal of investigative dermatology
Shimizuhira C,Otsuka A,Honda T,Kitoh A,Egawa G,Nakajima S,Nakashima C,Watarai H,Miyachi Y,Kabashima K
14-7313 was used in Flow cytometry/Cell sorting to report the pivotal contribution of dendritic and natural killer T cell interactions to hypersensitivity.
Sat Nov 01 00:00:00 EDT 2014
The aryl hydrocarbon receptor promotes IL-10 production by NK cells.
Journal of immunology (Baltimore, Md. : 1950)
Wagage S,John B,Krock BL,Hall AO,Randall LM,Karp CL,Simon MC,Hunter CA
14-7313 was used in an ELISA assay to investigate the source of IL-10 during Toxoplasma gondii infection.
Sat Feb 15 00:00:00 EST 2014
Vaccination with a single CD4 T cell peptide epitope from a Salmonella type III-secreted effector protein provides protection against lethal infection.
Infection and immunity
Kurtz JR,Petersen HE,Frederick DR,Morici LA,McLachlan JB
14-7313 was used in an ELISpot assay to investigate the immune response elicited against Salmonella to gather information for future vaccine design.
Sun Jun 01 00:00:00 EDT 2014
Conventional housing conditions attenuate the development of experimental autoimmune encephalomyelitis.
PloS one
Arndt A,Hoffacker P,Zellmer K,Goecer O,Recks MS,Kuerten S
14-7313 was used in an ELISpot assay to investigate the effect of pathogens on EAE, showing that conventional housing conditions attenuate the development of EAE.
Wed Sep 30 00:00:00 EDT 2015
Small molecule inhibitor of antigen binding and presentation by HLA-DR2b as a therapeutic strategy for the treatment of multiple sclerosis.
Journal of immunology (Baltimore, Md. : 1950)
Ji N,Somanaboeina A,Dixit A,Kawamura K,Hayward NJ,Self C,Olson GL,Forsthuber TG
14-7313 was used in an ELISpot assay to investigate how blocking the HLA-DR2b allele with small molecule inhibitors may offer a promising therapeutic strategy for the treatment of multiple sclerosis.
Fri Nov 15 00:00:00 EST 2013
Regulation of adaptive immunity by the fractalkine receptor during autoimmune inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Garcia JA,Pino PA,Mizutani M,Cardona SM,Charo IF,Ransohoff RM,Forsthuber TG,Cardona AE
14-7313 was used in an ELISpot assay to determine the role of CX3CR1 in the trafficking and function of myeloid cells during experimental autoimmune encephalomyelitis.
Thu Aug 01 00:00:00 EDT 2013
B lymphocyte "original sin" in the bone marrow enhances islet autoreactivity in type 1 diabetes-prone nonobese diabetic mice.
Journal of immunology (Baltimore, Md. : 1950)
Henry-Bonami RA,Williams JM,Rachakonda AB,Karamali M,Kendall PL,Thomas JW
14-7313 was used in Flow cytometry/Cell sorting to better understand the development of anti-insulin B cells by tracking their development in the polyclonal repertoire of VH125Tg mice.
Sat Jun 15 00:00:00 EDT 2013
A T cell epitope-based vaccine protects against chlamydial infection in HLA-DR4 transgenic mice.
Vaccine
Li W,Murthy AK,Lanka GK,Chetty SL,Yu JJ,Chambers JP,Zhong G,Forsthuber TG,Guentzel MN,Arulanandam BP
14-7313 was used in an ELISpot assay to investigate CD4+ T cell reactive chlamydial protease-like activity factor epitopes and characterise activation of T cells following antigen immunisation.
Tue Nov 19 00:00:00 EST 2013
MHC mismatch results in neural progenitor cell rejection following spinal cord transplantation in a model of viral-induced demyelination.
Stem cells (Dayton, Ohio)
Weinger JG,Weist BM,Plaisted WC,Klaus SM,Walsh CM,Lane TE
14-7313 was used in an ELISpot assay to investigate whether allogeneic neural progenitor cells are tolerated or induce immune-mediated rejection under neuroinflammatory demyelinating conditions.
Thu Nov 01 00:00:00 EDT 2012
Dietary n-3 polyunsaturated fatty acids (PUFA) decrease obesity-associated Th17 cell-mediated inflammation during colitis.
PloS one
Monk JM,Hou TY,Turk HF,Weeks B,Wu C,McMurray DN,Chapkin RS
14-7313 was used in Functional assays to monitor changes in immune cell subsets and inflammatory biomarker expression in a mouse model of concurrent obesity and colitis.
Thu Jun 20 00:00:00 EDT 2013
Virus-induced tumor inflammation facilitates effective DC cancer immunotherapy in a Treg-dependent manner in mice.
The Journal of clinical investigation
Woller N,Knocke S,Mundt B,Gürlevik E,Strüver N,Kloos A,Boozari B,Schache P,Manns MP,Malek NP,Sparwasser T,Zender L,Wirth TC,Kubicka S,Kühnel F
14-7313 was used in an ELISpot assay to highlight viruses as attractive tools for eliciting effective antitumour responses upon DC vaccination.
Fri Jul 01 00:00:00 EDT 2011
Inhibition of SOCS1-/- lethal autoinflammatory disease correlated to enhanced peripheral Foxp3+ regulatory T cell homeostasis.
Journal of immunology (Baltimore, Md. : 1950)
Collins EL,Jager LD,Dabelic R,Benitez P,Holdstein K,Lau K,Haider MI,Johnson HM,Larkin J
Published figure using IFN gamma monoclonal antibody (Product # 14-7313-81) in ELISA
Thu Sep 01 00:00:00 EDT 2011
Proteasomes, TAP, and endoplasmic reticulum-associated aminopeptidase associated with antigen processing control CD4+ Th cell responses by regulating indirect presentation of MHC class II-restricted cytoplasmic antigens.
Journal of immunology (Baltimore, Md. : 1950)
Dragovic SM,Hill T,Christianson GJ,Kim S,Elliott T,Scott D,Roopenian DC,Van Kaer L,Joyce S
14-7313 was used in an ELISpot assay to study a novel model in which the cytosolic Ag-processing machinery regulates the quantity of cytoplasmic peptides available for presentation by class II molecules and, hence, modulates Th cell responses.
Wed Jun 15 00:00:00 EDT 2011
In vivo requirement for Atg5 in antigen presentation by dendritic cells.
Immunity
Lee HK,Mattei LM,Steinberg BE,Alberts P,Lee YH,Chervonsky A,Mizushima N,Grinstein S,Iwasaki A
14-7313 was used in an ELISpot assay to investigate how dendritic cells utilise autophagic machinery to optimally present extracellular microbial antigens for MHC II molecules.
Fri Feb 26 00:00:00 EST 2010
CD137 agonist antibody prevents cancer recurrence: contribution of CD137 on both hematopoietic and nonhematopoietic cells.
Blood
Narazaki H,Zhu Y,Luo L,Zhu G,Chen L
14-7313 was used in an ELISpot assay to investigate methods to induce memory T cells to prevent tumour recurrence.
Thu Mar 11 00:00:00 EST 2010
Selectivity of oncolytic viral replication prevents antiviral immune response and toxicity, but does not improve antitumoral immunity.
Molecular therapy : the journal of the American Society of Gene Therapy
Gürlevik E,Woller N,Strüver N,Schache P,Kloos A,Manns MP,Zender L,Kühnel F,Kubicka S
14-7313 was used in an ELISpot assay to investigate how the degree of tumor selectivity of viral replication marginally influences antitumoral immune responses, but is a major determinant of antivector immunity and systemic toxicity.
Mon Nov 01 00:00:00 EDT 2010
CD4+ T cells elicit host immune responses to MHC class II-negative ovarian cancer through CCL5 secretion and CD40-mediated licensing of dendritic cells.
Journal of immunology (Baltimore, Md. : 1950)
Nesbeth YC,Martinez DG,Toraya S,Scarlett UK,Cubillos-Ruiz JR,Rutkowski MR,Conejo-Garcia JR
14-7313 was used in an ELISpot assay to investigate how to incorporate tumor-reactive CD4(+) T cells in adoptive cell transfer immunotherapies against ovarian cancer.
Sat May 15 00:00:00 EDT 2010
CCR7-dependent immunity during acute Toxoplasma gondii infection.
Infection and immunity
Noor S,Habashy AS,Nance JP,Clark RT,Nemati K,Carson MJ,Wilson EH
14-7313 was used in an ELISA assay to investigate the role of CCR7 signaling in generating protective immune responses to the intracellular protozoan parasite Toxoplasma gondii.
Sat May 01 00:00:00 EDT 2010
MHC class II-dependent basophil-CD4+ T cell interactions promote T(H)2 cytokine-dependent immunity.
Nature immunology
Perrigoue JG,Saenz SA,Siracusa MC,Allenspach EJ,Taylor BC,Giacomin PR,Nair MG,Du Y,Zaph C,van Rooijen N,Comeau MR,Pearce EJ,Laufer TM,Artis D
14-7313 was used in an ELISA assay to suggest that major histocompatibility complex class II-dependent interactions between basophils and CD4+ T cells promote T helper type 2 cytokine responses and immunity to helminth infection.
Wed Jul 01 00:00:00 EDT 2009
Human bone marrow-derived mesenchymal stem cells induce Th2-polarized immune response and promote endogenous repair in animal models of multiple sclerosis.
Glia
Bai L,Lennon DP,Eaton V,Maier K,Caplan AI,Miller SD,Miller RH
14-7313 was used in an ELISpot assay to suggest that bone marrow-derived mesenchymal stem cells represent a viable option for therapeutic approaches to multiple sclerosis.
Sat Aug 15 00:00:00 EDT 2009
Encephalitogenicity of complete Freund's adjuvant relative to CpG is linked to induction of Th17 cells.
Journal of immunology (Baltimore, Md. : 1950)
Tigno-Aranjuez JT,Jaini R,Tuohy VK,Lehmann PV,Tary-Lehmann M
14-7313 was used in an ELISPOT assay to compare both complete Freund's adjuvant-induced, and CpG induced models of experimental autoimmune encephalmyelitis, showing that CpG results in fewer Ag-specific IL-17 cells.
Sun Nov 01 00:00:00 EDT 2009
Estradiol-induced vaginal mucus inhibits antigen penetration and CD8(+) T cell priming in response to intravaginal immunization.
Vaccine
Seavey MM,Mosmann TR
14-7313 was used in an ELISpot assay to investigate the effect of estradiol during intravaginal immunisation, showing that estradiol-induced vaginal mucus inhibits antigen penetration and CD8(+) T cell priming.
Tue Apr 14 00:00:00 EDT 2009
IL-10 from CD4CD25Foxp3CD127 adaptive regulatory T cells modulates parasite clearance and pathology during malaria infection.
PLoS pathogens
Couper KN,Blount DG,Wilson MS,Hafalla JC,Belkaid Y,Kamanaka M,Flavell RA,de Souza JB,Riley EM
14-7313 was used in an ELISA assay to investigate the cellular source of IL-10 and TGF-beta during malaria infection, showing that IL-10 from CD4+CD25-Foxp3-CD127- adaptive Tregs modulates parasite clearance and pathology.
Fri Feb 29 00:00:00 EST 2008
Macrophage-mediated but gamma interferon-independent innate immune responses control the primary wave of Plasmodium yoelii parasitemia.
Infection and immunity
Couper KN,Blount DG,Hafalla JC,van Rooijen N,de Souza JB,Riley EM
14-7313 was used in an ELISA assay to discern the factors that control the primary wave of plasmodium yoelii parasitaemia.
Sat Dec 01 00:00:00 EST 2007
Efficient presentation of myelin oligodendrocyte glycoprotein peptides but not protein by astrocytes from HLA-DR2 and HLA-DR4 transgenic mice.
Journal of neuroimmunology
Kort JJ,Kawamura K,Fugger L,Weissert R,Forsthuber TG
Sat Apr 01 00:00:00 EST 2006