Product References
Stretch regulates alveologenesis and homeostasis via mesenchymal Gαq/11-mediated TGFβ2 activation.
Development (Cambridge, England)
Goodwin AT,John AE,Joseph C,Habgood A,Tatler AL,Susztak K,Palmer M,Offermanns S,Henderson NC,Jenkins RG
Mon May 01 00:00:00 EDT 2023
Stretch regulates alveologenesis and homeostasis via mesenchymal Gαq/11-mediated TGFβ2 activation.
Development (Cambridge, England)
Goodwin AT,John AE,Joseph C,Habgood A,Tatler AL,Susztak K,Palmer M,Offermanns S,Henderson NC,Jenkins RG
Mon May 01 00:00:00 EDT 2023
Study on the Mechanism of Jiaotai Pill Intervention on Insomnia Animal Model Based on Gut Microbiome and Metabolomics.
Evidence-based complementary and alternative medicine : eCAM
Yang Y,Liu J,Ou H,Ma X,Li J,Shao B,Jin R,Zhao J
Sun Jun 04 00:00:00 EDT 2023
Dexamethasone Induces Senescence-Associated Changes in Trabecular Meshwork Cells by Increasing ROS Levels Via the TGFβ/Smad3-NOX4 Axis.
Cell transplantation
Li H,Ren J,Cui H,Wang D,Zhao R,Liu X,Tian S,Wang J,Zhang J,Li P,Thorne RF,Duan S
Mon Jun 05 00:00:00 EDT 2023
GSK-3 inhibitor elraglusib enhances tumor-infiltrating immune cell activation in tumor biopsies and synergizes with anti-PD-L1 in a murine model of colorectal cancer.
bioRxiv : the preprint server for biology
Huntington KE,Louie AD,Srinivasan PR,Schorl C,Lu S,Silverberg D,Newhouse D,Wu Z,Zhou L,Borden BA,Giles FJ,Dooner M,Carneiro BA,El-Deiry WS
Tue Feb 07 00:00:00 EST 2023
Role of PCIF1-mediated 5'-cap N6-methyladeonsine mRNA methylation in colorectal cancer and anti-PD-1 immunotherapy.
The EMBO journal
Wang L,Wu L,Zhu Z,Zhang Q,Li W,Gonzalez GM,Wang Y,Rana TM
Mon Jan 16 00:00:00 EST 2023
Bu Shen Yi Sui Capsule Promotes Myelin Repair by Modulating the Transformation of A1/A2 Reactive Astrocytes In Vivo and In Vitro.
Oxidative medicine and cellular longevity
Zha Z,Liu YJ,Liu SS,Zhang N,Li JL,Qi F,Jin LY,Xue B,Yang T,Fan YP,Zhao H,Wang L
Tue Sep 13 00:00:00 EDT 2022
Sodium butyrate alleviates pre-eclampsia in pregnant rats by improving the gut microbiota and short-chain fatty acid metabolites production.
Journal of applied microbiology
Yong W,Zhao Y,Jiang X,Li P
Tue Feb 01 00:00:00 EST 2022
TGFβ2 is a Prognostic Biomarker for Gastric Cancer and is Associated With Methylation and Immunotherapy Responses.
Frontiers in genetics
Han B,Fang T,Wang Y,Zhang Y,Xue Y
Sat Jul 16 00:00:00 EDT 2022
TGFβ2 is a Prognostic Biomarker for Gastric Cancer and is Associated With Methylation and Immunotherapy Responses.
Frontiers in genetics
Han B,Fang T,Wang Y,Zhang Y,Xue Y
Sat Jul 16 00:00:00 EDT 2022
Comparative analyses in transcriptome of human granulosa cells and follicular fluid micro-environment between poor ovarian responders with conventional controlled ovarian or mild ovarian stimulations.
Reproductive biology and endocrinology : RB&E
Liu X,Mai H,Chen P,Zhang Z,Wu T,Chen J,Sun P,Zhou C,Liang X,Huang R
Mon Mar 21 00:00:00 EDT 2022
Nicotinamide N-Methyltransferase Remodeled Cell Metabolism and Aggravated Proinflammatory Responses by Activating STAT3/IL1β/PGE2 Pathway.
ACS omega
Yang C,Wang T,Zhu S,Zong Z,Luo C,Zhao Y,Liu J,Li T,Liu X,Liu C,Deng H
Tue Oct 25 00:00:00 EDT 2022
Platelet Derived Vesicles Enhance the TGF-beta Signaling Pathway of M1 Macrophage.
Frontiers in endocrinology
Song N,Pan K,Chen L,Jin K
Tue Apr 05 00:00:00 EDT 2022
Association of CASC18/miR-20a-3p/TGFB2 ceRNA axis with occult lymph node metastasis in tongue squamous cell carcinoma.
Molecular medicine (Cambridge, Mass.)
Zhou B,Zhou Y,Liu Y,Zhang H,Mao H,Peng M,Xu A,Li Z,Wang H,Tan H,Ren H,Zhou X,Long Y
Fri Aug 06 00:00:00 EDT 2021
LncRNA PART1 promotes cell proliferation and progression in non-small-cell lung cancer cells via sponging miR-17-5p.
Journal of cellular biochemistry
Chen Y,Zhou X,Huang C,Li L,Qin Y,Tian Z,He J,Liu H
Thu Apr 01 00:00:00 EDT 2021
SALL1 regulates commitment of odontoblast lineages by interacting with RUNX2 to remodel open chromatin regions.
Stem cells (Dayton, Ohio)
Lin Y,Xiao Y,Lin C,Zhang Q,Zhang S,Pei F,Liu H,Chen Z
Mon Feb 01 00:00:00 EST 2021
Cadmium promotes apoptosis and inflammation via the circ08409/miR-133a/TGFB2 axis in bovine mammary epithelial cells and mouse mammary gland.
Ecotoxicology and environmental safety
Chen Z,Liang Y,Lu Q,Nazar M,Mao Y,Aboragah A,Yang Z,Loor JJ
Fri Oct 01 00:00:00 EDT 2021
Anti-androgenetic alopecia effect of policosanol from Chinese wax by regulating abnormal hormone levels to suppress premature hair follicle entry into the regression phase.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Wang ZD,Feng Y,Sun L,Gan J,Li X,Ding WF,Chen XM
Thu Apr 01 00:00:00 EDT 2021
USP9X-mediated KDM4C deubiquitination promotes lung cancer radioresistance by epigenetically inducing TGF-β2 transcription.
Cell death and differentiation
Jie X,Fong WP,Zhou R,Zhao Y,Zhao Y,Meng R,Zhang S,Dong X,Zhang T,Yang K,Wu G,Xu S
Thu Jul 01 00:00:00 EDT 2021
tRNA-Derived Fragment tRF-Glu-TTC-027 Regulates the Progression of Gastric Carcinoma via MAPK Signaling Pathway.
Frontiers in oncology
Xu W,Zhou B,Wang J,Tang L,Hu Q,Wang J,Chen H,Zheng J,Yan F,Chen H
Fri Sep 10 00:00:00 EDT 2021
ZEB1 represses biogenesis of circ-DOCK5 to facilitate metastasis in esophageal squamous cell carcinoma via a positive feedback loop with TGF-β.
Cancer letters
Meng L,Zheng Y,Liu S,Ju Y,Ren S,Sang Y,Zhu Y,Gu L,Liu F,Zhao Y,Zhang X,Sang M
Thu Oct 28 00:00:00 EDT 2021
Exosome-transmitted circCOG2 promotes colorectal cancer progression via miR-1305/TGF-β2/SMAD3 pathway.
Cell death discovery
Gao L,Tang X,He Q,Sun G,Wang C,Qu H
Mon Oct 11 00:00:00 EDT 2021
Exosome-transmitted circCOG2 promotes colorectal cancer progression via miR-1305/TGF-β2/SMAD3 pathway.
Cell death discovery
Gao L,Tang X,He Q,Sun G,Wang C,Qu H
Mon Oct 11 00:00:00 EDT 2021
PPARα-Dependent Effects of Palmitoylethanolamide Against Retinal Neovascularization and Fibrosis.
Investigative ophthalmology & visual science
Ye S,Chen Q,Jiang N,Liang X,Li J,Zong R,Huang C,Qiu Y,Ma JX,Liu Z
Thu Apr 09 00:00:00 EDT 2020
Gold Nanoparticles Induce Tumor Vessel Normalization and Impair Metastasis by Inhibiting Endothelial Smad2/3 Signaling.
ACS nano
Huang N,Liu Y,Fang Y,Zheng S,Wu J,Wang M,Zhong W,Shi M,Xing M,Liao W
Tue Jul 28 00:00:00 EDT 2020
Expression of miR-181a and TGF-β2 in lens epithelial cells of patients with cataractous retinal detachment and its clinical significance.
Experimental and therapeutic medicine
Jiang Y,Gong Y,Lin N,Qiu W
Sat Jun 01 00:00:00 EDT 2019
The Smad3-miR-29b/miR-29c axis mediates the protective effect of macrophage migration inhibitory factor against cardiac fibrosis.
Biochimica et biophysica acta. Molecular basis of disease
Liang JN,Zou X,Fang XH,Xu JD,Xiao Z,Zhu JN,Li H,Yang J,Zeng N,Yuan SJ,Pan R,Fu YH,Zhang M,Luo JF,Wang S,Shan ZX
Sun Sep 01 00:00:00 EDT 2019
LncRNA CASC9 interacts with CPSF3 to regulate TGF-β signaling in colorectal cancer.
Journal of experimental & clinical cancer research : CR
Luo K,Geng J,Zhang Q,Xu Y,Zhou X,Huang Z,Shi KQ,Pan C,Wu J
Tue Jun 11 00:00:00 EDT 2019
The role and mechanism of transforming growth factor beta 3 in human myocardial infarction-induced myocardial fibrosis.
Journal of cellular and molecular medicine
Xue K,Zhang J,Li C,Li J,Wang C,Zhang Q,Chen X,Yu X,Sun L,Yu X
Sat Jun 01 00:00:00 EDT 2019
miR-618 Suppresses Metastasis in Gastric Cancer by Downregulating the Expression of TGF-β2.
Anatomical record (Hoboken, N.J. : 2007)
Shi J,Gong L,Chen L,Luo J,Song G,Xu J,Lv Z,Tao H,Xia Y,Ye Z
Sat Jun 01 00:00:00 EDT 2019
Osteoblast-Secreted Factors Mediate Dormancy of Metastatic Prostate Cancer in the Bone via Activation of the TGFβRIII-p38MAPK-pS249/T252RB Pathway.
Cancer research
Yu-Lee LY,Yu G,Lee YC,Lin SC,Pan J,Pan T,Yu KJ,Liu B,Creighton CJ,Rodriguez-Canales J,Villalobos PA,Wistuba II,de Nadal E,Posas F,Gallick GE,Lin SH
Fri Jun 01 00:00:00 EDT 2018
Osteoblast-Secreted Factors Mediate Dormancy of Metastatic Prostate Cancer in the Bone via Activation of the TGFβRIII-p38MAPK-pS249/T252RB Pathway.
Cancer research
Yu-Lee LY,Yu G,Lee YC,Lin SC,Pan J,Pan T,Yu KJ,Liu B,Creighton CJ,Rodriguez-Canales J,Villalobos PA,Wistuba II,de Nadal E,Posas F,Gallick GE,Lin SH
Fri Jun 01 00:00:00 EDT 2018
Osteoblast-Secreted Factors Mediate Dormancy of Metastatic Prostate Cancer in the Bone via Activation of the TGFβRIII-p38MAPK-pS249/T252RB Pathway.
Cancer research
Yu-Lee LY,Yu G,Lee YC,Lin SC,Pan J,Pan T,Yu KJ,Liu B,Creighton CJ,Rodriguez-Canales J,Villalobos PA,Wistuba II,de Nadal E,Posas F,Gallick GE,Lin SH
Fri Jun 01 00:00:00 EDT 2018
Osteoblast-Secreted Factors Mediate Dormancy of Metastatic Prostate Cancer in the Bone via Activation of the TGFβRIII-p38MAPK-pS249/T252RB Pathway.
Cancer research
Yu-Lee LY,Yu G,Lee YC,Lin SC,Pan J,Pan T,Yu KJ,Liu B,Creighton CJ,Rodriguez-Canales J,Villalobos PA,Wistuba II,de Nadal E,Posas F,Gallick GE,Lin SH
Fri Jun 01 00:00:00 EDT 2018
Modulation of TGF‑β activity by latent TGF‑β‑binding protein 1 in human osteoarthritis fibroblast‑like synoviocytes.
Molecular medicine reports
Wang X,Dong C,Li N,Ma Q,Yun Z,Cai C,An M,Ma B
Mon Jan 01 00:00:00 EST 2018
EDIL3 depletion suppress epithelial-mesenchymal transition of lens epithelial cells via transforming growth factor β pathway.
International journal of ophthalmology
Zhang R,Wei YH,Zhao CY,Song HY,Shen N,Cui X,Gao X,Qi ZT,Zhong M,Shen W
Wed Sep 30 00:00:00 EDT 2020
EDIL3 depletion suppress epithelial-mesenchymal transition of lens epithelial cells via transforming growth factor β pathway.
International journal of ophthalmology
Zhang R,Wei YH,Zhao CY,Song HY,Shen N,Cui X,Gao X,Qi ZT,Zhong M,Shen W
Wed Sep 30 00:00:00 EDT 2020
Zoledronate inhibits fibroblasts' proliferation and activation via targeting TGF-β signaling pathway.
Drug design, development and therapy
Zhao Z,Shen W,Zhu H,Lin L,Jiang G,Zhu Y,Song H,Wu L
Mon Dec 24 00:00:00 EST 2018
Zoledronate inhibits fibroblasts' proliferation and activation via targeting TGF-β signaling pathway.
Drug design, development and therapy
Zhao Z,Shen W,Zhu H,Lin L,Jiang G,Zhu Y,Song H,Wu L
Mon Dec 24 00:00:00 EST 2018
Zoledronate inhibits fibroblasts' proliferation and activation via targeting TGF-β signaling pathway.
Drug design, development and therapy
Zhao Z,Shen W,Zhu H,Lin L,Jiang G,Zhu Y,Song H,Wu L
Mon Dec 24 00:00:00 EST 2018
Zoledronate inhibits fibroblasts' proliferation and activation via targeting TGF-β signaling pathway.
Drug design, development and therapy
Zhao Z,Shen W,Zhu H,Lin L,Jiang G,Zhu Y,Song H,Wu L
Mon Dec 24 00:00:00 EST 2018
Endothelial-to-Osteoblast Conversion Generates Osteoblastic Metastasis of Prostate Cancer.
Developmental cell
Lin SC,Lee YC,Yu G,Cheng CJ,Zhou X,Chu K,Murshed M,Le NT,Baseler L,Abe JI,Fujiwara K,deCrombrugghe B,Logothetis CJ,Gallick GE,Yu-Lee LY,Maity SN,Lin SH
Mon Jun 05 00:00:00 EDT 2017
Endothelial-to-Osteoblast Conversion Generates Osteoblastic Metastasis of Prostate Cancer.
Developmental cell
Lin SC,Lee YC,Yu G,Cheng CJ,Zhou X,Chu K,Murshed M,Le NT,Baseler L,Abe JI,Fujiwara K,deCrombrugghe B,Logothetis CJ,Gallick GE,Yu-Lee LY,Maity SN,Lin SH
Mon Jun 05 00:00:00 EDT 2017
High doses of baicalin induces kidney injury and fibrosis through regulating TGF-β/Smad signaling pathway.
Toxicology and applied pharmacology
Cai Y,Ma W,Xiao Y,Wu B,Li X,Liu F,Qiu J,Zhang G
Sun Oct 15 00:00:00 EDT 2017
High doses of baicalin induces kidney injury and fibrosis through regulating TGF-β/Smad signaling pathway.
Toxicology and applied pharmacology
Cai Y,Ma W,Xiao Y,Wu B,Li X,Liu F,Qiu J,Zhang G
Sun Oct 15 00:00:00 EDT 2017
Lnc-ATB contributes to gastric cancer growth through a MiR-141-3p/TGFβ2 feedback loop.
Biochemical and biophysical research communications
Lei K,Liang X,Gao Y,Xu B,Xu Y,Li Y,Tao Y,Shi W,Liu J
Sat Mar 11 00:00:00 EST 2017
Lnc-ATB contributes to gastric cancer growth through a MiR-141-3p/TGFβ2 feedback loop.
Biochemical and biophysical research communications
Lei K,Liang X,Gao Y,Xu B,Xu Y,Li Y,Tao Y,Shi W,Liu J
Sat Mar 11 00:00:00 EST 2017
Gender differences in fibrosis remodeling in patients with long-standing persistent atrial fibrillation.
Oncotarget
Li Z,Wang Z,Yin Z,Zhang Y,Xue X,Han J,Zhu Y,Zhang J,Emmert MY,Wang H
Tue Aug 08 00:00:00 EDT 2017
Gender differences in fibrosis remodeling in patients with long-standing persistent atrial fibrillation.
Oncotarget
Li Z,Wang Z,Yin Z,Zhang Y,Xue X,Han J,Zhu Y,Zhang J,Emmert MY,Wang H
Tue Aug 08 00:00:00 EDT 2017
TGF-β2 is involved in the preservation of the chondrocyte phenotype under hypoxic conditions.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft
Das R,Timur UT,Edip S,Haak E,Wruck C,Weinans H,Jahr H
Sun Mar 01 00:00:00 EST 2015
TGF-β2 is involved in the preservation of the chondrocyte phenotype under hypoxic conditions.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft
Das R,Timur UT,Edip S,Haak E,Wruck C,Weinans H,Jahr H
Sun Mar 01 00:00:00 EST 2015
SIX1 promotes tumor lymphangiogenesis by coordinating TGFβ signals that increase expression of VEGF-C.
Cancer research
Liu D,Li L,Zhang XX,Wan DY,Xi BX,Hu Z,Ding WC,Zhu D,Wang XL,Wang W,Feng ZH,Wang H,Ma D,Gao QL
Wed Oct 01 00:00:00 EDT 2014
SIX1 promotes tumor lymphangiogenesis by coordinating TGFβ signals that increase expression of VEGF-C.
Cancer research
Liu D,Li L,Zhang XX,Wan DY,Xi BX,Hu Z,Ding WC,Zhu D,Wang XL,Wang W,Feng ZH,Wang H,Ma D,Gao QL
Wed Oct 01 00:00:00 EDT 2014
Participation of miR-200a in TGF-β1-mediated hepatic stellate cell activation.
Molecular and cellular biochemistry
Sun X,He Y,Ma TT,Huang C,Zhang L,Li J
Sat Mar 01 00:00:00 EST 2014
Participation of miR-200a in TGF-β1-mediated hepatic stellate cell activation.
Molecular and cellular biochemistry
Sun X,He Y,Ma TT,Huang C,Zhang L,Li J
Sat Mar 01 00:00:00 EST 2014