Product References
The acetyltransferase p300 is recruited in trans to multiple enhancer sites by lncSmad7.
Nucleic acids research
Maldotti M,Lauria A,Anselmi F,Molineris I,Tamburrini A,Meng G,Polignano IL,Scrivano MG,Campestre F,Simon LM,Rapelli S,Morandi E,Incarnato D,Oliviero S
42-0400 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot to find 122 lncRNAs that interacts directly with p300.
Mon Mar 21 00:00:00 EDT 2022
The role of endogenous Smad7 in regulating macrophage phenotype following myocardial infarction.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Li J,Li R,Tuleta I,Hernandez SC,Humeres C,Hanna A,Chen B,Frangogiannis NG
42-0400 was used in Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot to suggest a limited role for macrophage Smad7 in regulation of post-infarction inflammation and repair, and demonstrate that the anti-inflammatory effects of TGF-β in macrophages are not restrained by endogenous Smad7 induction.
Fri Jul 01 00:00:00 EDT 2022
Cancer-associated fibroblasts downregulate type I interferon receptor to stimulate intratumoral stromagenesis.
Oncogene
Cho C,Mukherjee R,Peck AR,Sun Y,McBrearty N,Katlinski KV,Gui J,Govindaraju PK,Puré E,Rui H,Fuchs SY
42-0400 was used in Western Blot, Immunocytochemistry to investigate the importance of IFNAR1 inactivation in the non-immune tumor stroma.
Tue Sep 01 00:00:00 EDT 2020
Cancer-associated fibroblasts downregulate type I interferon receptor to stimulate intratumoral stromagenesis.
Oncogene
Cho C,Mukherjee R,Peck AR,Sun Y,McBrearty N,Katlinski KV,Gui J,Govindaraju PK,Puré E,Rui H,Fuchs SY
42-0400 was used in Western Blot, Immunocytochemistry to investigate the importance of IFNAR1 inactivation in the non-immune tumor stroma.
Tue Sep 01 00:00:00 EDT 2020
HDAC inhibitors promote pancreatic stellate cell apoptosis and relieve pancreatic fibrosis by upregulating miR-15/16 in chronic pancreatitis.
Human cell
Ji T,Feng W,Zhang X,Zang K,Zhu X,Shang F
42-0400 was used in Western Blotting to find that miR-15 and miR-16 decreased significantly in chronic pancreatitis and HDAC inhibition could recover the levels of these two miRNAs.
Thu Oct 01 00:00:00 EDT 2020
HDAC inhibitors promote pancreatic stellate cell apoptosis and relieve pancreatic fibrosis by upregulating miR-15/16 in chronic pancreatitis.
Human cell
Ji T,Feng W,Zhang X,Zang K,Zhu X,Shang F
42-0400 was used in Western Blotting to find that miR-15 and miR-16 decreased significantly in chronic pancreatitis and HDAC inhibition could recover the levels of these two miRNAs.
Thu Oct 01 00:00:00 EDT 2020
Role of sialidase Neu3 and ganglioside GM3 in cardiac fibroblasts activation.
The Biochemical journal
Ghiroldi A,Piccoli M,Creo P,Cirillo F,Rota P,D'Imperio S,Ciconte G,Monasky MM,Micaglio E,Garatti A,Aureli M,Carsana EV,Menicanti L,Pappone C,Anastasia L
42-0400 was used in Western Blot to identify pro-fibrotic pathways in primary cultures of human cardiac fibroblasts as targets for cardiac fibrosis.
Fri Sep 18 00:00:00 EDT 2020
Effects of tacrolimus on the TGF‑β1/SMAD signaling pathway in paraquat‑exposed rat alveolar type II epithelial cells.
Molecular medicine reports
Ren Y,Jian X,Zhang Z,Ning Q,Kan B,Kong L
42-0400 was used in Immunocytochemistry-immunoflourescence to provide evidence that tacrolimus can reduce the levels of TGF‚β1, SMAD3 and CTGF, increase the level of SMAD7 in TGF‚β1 signaling pathway and protect the development of pulmonary fibrosis in paraquat exposed alveolar epithelial cells.
Sun Nov 01 00:00:00 EDT 2020
Effects of tacrolimus on the TGF‑β1/SMAD signaling pathway in paraquat‑exposed rat alveolar type II epithelial cells.
Molecular medicine reports
Ren Y,Jian X,Zhang Z,Ning Q,Kan B,Kong L
42-0400 was used in Immunocytochemistry-immunoflourescence to provide evidence that tacrolimus can reduce the levels of TGF‚β1, SMAD3 and CTGF, increase the level of SMAD7 in TGF‚β1 signaling pathway and protect the development of pulmonary fibrosis in paraquat exposed alveolar epithelial cells.
Sun Nov 01 00:00:00 EDT 2020
Deletion of Akt1 Promotes Kidney Fibrosis in a Murine Model of Unilateral Ureteral Obstruction.
BioMed research international
Kim IY,Lee MY,Park MW,Seong EY,Lee DW,Lee SB,Bae SS,Kim SS,Song SH
42-0400 was used in Western Blotting to show renal fibrosis can be promoted by Akt1 deletion by induction of the TGFβ1/STAT3 pathway.
Fri May 07 00:00:00 EDT 2021
Cryptotanshinone Protects Cartilage against Developing Osteoarthritis through the miR-106a-5p/GLIS3 Axis.
Molecular therapy. Nucleic acids
Ji Q,Qi D,Xu X,Xu Y,Goodman SB,Kang L,Song Q,Fan Z,Maloney WJ,Wang Y
42-0400 was used in Western Blotting to study the role of microRNAs in the therapeutic effect of cryptotanshinone in osteoarthritis.
Fri Jun 01 00:00:00 EDT 2018
SMAD7 regulates proinflammatory and prolabor mediators in amnion and myometrium.
Biology of reproduction
Lim R,Barker G,Lappas M
42-0400 was used in Immunohistochemistry on paraffin embedded tissues to study the effect of spontaneous human labour and prolabour mediators on SMAD7 expression in myometrium and fetal membranes.
Tue Aug 01 00:00:00 EDT 2017
SMAD7 regulates proinflammatory and prolabor mediators in amnion and myometrium.
Biology of reproduction
Lim R,Barker G,Lappas M
42-0400 was used in Immunohistochemistry on paraffin embedded tissues to study the effect of spontaneous human labour and prolabour mediators on SMAD7 expression in myometrium and fetal membranes.
Tue Aug 01 00:00:00 EDT 2017
Cucurbitacin I Attenuates Cardiomyocyte Hypertrophy via Inhibition of Connective Tissue Growth Factor (CCN2) and TGF- β/Smads Signalings.
PloS one
Jeong MH,Kim SJ,Kang H,Park KW,Park WJ,Yang SY,Yang DK
42-0400 was used in western blot to determine the functional role of cucurbitacin I using an in vitro model of cardiac hypertrophy.
Tue May 10 00:00:00 EDT 2016
Neuropilin-2 Is upregulated in lung cancer cells during TGF-β1-induced epithelial-mesenchymal transition.
Cancer research
Nasarre P,Gemmill RM,Potiron VA,Roche J,Lu X,Barón AE,Korch C,Garrett-Mayer E,Lagana A,Howe PH,Drabkin HA
42-0400 was used in western blot to study TGF-beta-driven lung cancer cell epithelial-to-mesenchymal transition and the role played by neuropilin-2.
Sun Dec 01 00:00:00 EST 2013
Kruppel-like factor KLF10 targets transforming growth factor-beta1 to regulate CD4(+)CD25(-) T cells and T regulatory cells.
The Journal of biological chemistry
Cao Z,Wara AK,Icli B,Sun X,Packard RR,Esen F,Stapleton CJ,Subramaniam M,Kretschmer K,Apostolou I,von Boehmer H,Hansson GK,Spelsberg TC,Libby P,Feinberg MW
Fri Sep 11 00:00:00 EDT 2009