Product References
Combination Treatment Targeting mTOR and MAPK Pathways Has Synergistic Activity in Multiple Myeloma.
Cancers
Sun K,Jin L,Karolová J,Vorwerk J,Hailfinger S,Opalka B,Zapukhlyak M,Lenz G,Khandanpour C
44-1100G was used in Western Blotting to provide a scientific rationale for the simultaneous inhibition of mTOR and MEK as a novel strategy for the treatment of MM.
Wed Apr 19 00:00:00 EDT 2023
EIF1AX and RAS Mutations Cooperate to Drive Thyroid Tumorigenesis through ATF4 and c-MYC.
Cancer discovery
Krishnamoorthy GP,Davidson NR,Leach SD,Zhao Z,Lowe SW,Lee G,Landa I,Nagarajah J,Saqcena M,Singh K,Wendel HG,Dogan S,Tamarapu PP,Blenis J,Ghossein RA,Knauf JA,Rätsch G,Fagin JA
44-1100G was used in Western Blotting to describe the identification of the key signaling drivers of transformation by EIF1AX mutants, particularly EIF1AX-A113splice, alone and in the context of RAS, and the therapeutic dependencies they confer.
Fri Feb 01 00:00:00 EST 2019
Integrated Molecular Characterization of Uterine Carcinosarcoma.
Cancer cell
Cherniack AD,Shen H,Walter V,Stewart C,Murray BA,Bowlby R,Hu X,Ling S,Soslow RA,Broaddus RR,Zuna RE,Robertson G,Laird PW,Kucherlapati R,Mills GB,Weinstein JN,Zhang J,Akbani R,Levine DA
441100G was used in reverse phase protein array to characterize the molecular identity of uterine carcinosarcomas.
Mon Mar 13 00:00:00 EDT 2017
Context Specificity in Causal Signaling Networks Revealed by Phosphoprotein Profiling.
Cell systems
Hill SM,Nesser NK,Johnson-Camacho K,Jeffress M,Johnson A,Boniface C,Spencer SE,Lu Y,Heiser LM,Lawrence Y,Pande NT,Korkola JE,Gray JW,Mills GB,Mukherjee S,Spellman PT
441100G was used in western blot to analyze the context specificity of signaling networks within a causal conceptual framework using reverse-phase protein array time-course assays and network analysis approaches
Wed Jan 25 00:00:00 EST 2017
Identification of differential PI3K pathway target dependencies in T-cell acute lymphoblastic leukemia through a large cancer cell panel screen.
Oncotarget
Lynch JT,McEwen R,Crafter C,McDermott U,Garnett MJ,Barry ST,Davies BR
441100G was used in western blot to find tumor types sensitive to phosphoinositide 3-kinase pathway inhibitors
Tue Apr 19 00:00:00 EDT 2016
The Role of PDE3B Phosphorylation in the Inhibition of Lipolysis by Insulin.
Molecular and cellular biology
DiPilato LM,Ahmad F,Harms M,Seale P,Manganiello V,Birnbaum MJ
44-1100G was used in western blot to demonstrate that phosphorylation of PDE3B by Akt is not required for insulin to suppress adipocyte lipolysis.
Sat Aug 01 00:00:00 EDT 2015
Combination of heat shock protein 90 and focal adhesion kinase inhibitors synergistically inhibits the growth of non-small cell lung cancer cells.
Oncoscience
Webber PJ,Park C,Qui M,Ramalingam SS,Khuri FR,Fu H,Du Y
44-1100G was used in western blot to assess the impact of heat shock protein 90 inhibition in combination with focal adhesion kinase inhibitor on the growth of non-small cell lung cancer cells.
Tue Oct 27 00:00:00 EDT 2015
Characterization of the activity of the PI3K/mTOR inhibitor XL765 (SAR245409) in tumor models with diverse genetic alterations affecting the PI3K pathway.
Molecular cancer therapeutics
Yu P,Laird AD,Du X,Wu J,Won KA,Yamaguchi K,Hsu PP,Qian F,Jaeger CT,Zhang W,Buhr CA,Shen P,Abulafia W,Chen J,Young J,Plonowski A,Yakes FM,Chu F,Lee M,Bentzien F,Lam ST,Dale S,Matthews DJ,Lamb P,Foster P
44-1100G was used in western blot to study the anti-tumor effects of a pan class I PI3 kinase/mTOR inhibitor in cells with a genetically diverse range of PI3 kinase pathway modifications.
Thu May 01 00:00:00 EDT 2014
PTEN promoter methylation and activation of the PI3K/Akt/mTOR pathway in pediatric gliomas and influence on clinical outcome.
Neuro-oncology
Mueller S,Phillips J,Onar-Thomas A,Romero E,Zheng S,Wiencke JK,McBride SM,Cowdrey C,Prados MD,Weiss WA,Berger MS,Gupta N,Haas-Kogan DA
Published figure using Phospho-PRAS40 (Thr246) polyclonal antibody (Product # 44-1100G) in Immunohistochemistry
Sat Sep 01 00:00:00 EDT 2012
Phosphorylation of PRAS40 on Thr246 by PKB/AKT facilitates efficient phosphorylation of Ser183 by mTORC1.
Cellular signalling
Nascimento EB,Snel M,Guigas B,van der Zon GC,Kriek J,Maassen JA,Jazet IM,Diamant M,Ouwens DM
Tue Jun 01 00:00:00 EDT 2010
Phosphorylation of PRAS40 on Thr246 by PKB/AKT facilitates efficient phosphorylation of Ser183 by mTORC1.
Cellular signalling
Nascimento EB,Snel M,Guigas B,van der Zon GC,Kriek J,Maassen JA,Jazet IM,Diamant M,Ouwens DM
Tue Jun 01 00:00:00 EDT 2010
Activation of PI3K/mTOR pathway occurs in most adult low-grade gliomas and predicts patient survival.
Journal of neuro-oncology
McBride SM,Perez DA,Polley MY,Vandenberg SR,Smith JS,Zheng S,Lamborn KR,Wiencke JK,Chang SM,Prados MD,Berger MS,Stokoe D,Haas-Kogan DA
44-1100G was used in immunohistochemistry to study the impact of the Akt-mTOR pathway in low-grade gliomas
Mon Mar 01 00:00:00 EST 2010
mTOR complex 2 is required for the development of prostate cancer induced by Pten loss in mice.
Cancer cell
Guertin DA,Stevens DM,Saitoh M,Kinkel S,Crosby K,Sheen JH,Mullholland DJ,Magnuson MA,Wu H,Sabatini DM
Published figure using Phospho-PRAS40 (Thr246) polyclonal antibody (Product # 44-1100G) in Western Blot
Tue Feb 03 00:00:00 EST 2009
mTOR complex 2 is required for the development of prostate cancer induced by Pten loss in mice.
Cancer cell
Guertin DA,Stevens DM,Saitoh M,Kinkel S,Crosby K,Sheen JH,Mullholland DJ,Magnuson MA,Wu H,Sabatini DM
Published figure using Phospho-PRAS40 (Thr246) polyclonal antibody (Product # 44-1100G) in Western Blot
Tue Feb 03 00:00:00 EST 2009
mTOR complex 2 is required for the development of prostate cancer induced by Pten loss in mice.
Cancer cell
Guertin DA,Stevens DM,Saitoh M,Kinkel S,Crosby K,Sheen JH,Mullholland DJ,Magnuson MA,Wu H,Sabatini DM
Published figure using Phospho-PRAS40 (Thr246) polyclonal antibody (Product # 44-1100G) in Western Blot
Tue Feb 03 00:00:00 EST 2009
mTOR complex 2 is required for the development of prostate cancer induced by Pten loss in mice.
Cancer cell
Guertin DA,Stevens DM,Saitoh M,Kinkel S,Crosby K,Sheen JH,Mullholland DJ,Magnuson MA,Wu H,Sabatini DM
Published figure using Phospho-PRAS40 (Thr246) polyclonal antibody (Product # 44-1100G) in Western Blot
Tue Feb 03 00:00:00 EST 2009
Antitumor activity of rapamycin in a Phase I trial for patients with recurrent PTEN-deficient glioblastoma.
PLoS medicine
Cloughesy TF,Yoshimoto K,Nghiemphu P,Brown K,Dang J,Zhu S,Hsueh T,Chen Y,Wang W,Youngkin D,Liau L,Martin N,Becker D,Bergsneider M,Lai A,Green R,Oglesby T,Koleto M,Trent J,Horvath S,Mischel PS,Mellinghoff IK,Sawyers CL
Tue Jan 22 00:00:00 EST 2008
Antitumor activity of rapamycin in a Phase I trial for patients with recurrent PTEN-deficient glioblastoma.
PLoS medicine
Cloughesy TF,Yoshimoto K,Nghiemphu P,Brown K,Dang J,Zhu S,Hsueh T,Chen Y,Wang W,Youngkin D,Liau L,Martin N,Becker D,Bergsneider M,Lai A,Green R,Oglesby T,Koleto M,Trent J,Horvath S,Mischel PS,Mellinghoff IK,Sawyers CL
Tue Jan 22 00:00:00 EST 2008
Insulin signalling to mTOR mediated by the Akt/PKB substrate PRAS40.
Nature cell biology
Vander Haar E,Lee SI,Bandhakavi S,Griffin TJ,Kim DH
Thu Mar 01 00:00:00 EST 2007
Insulin signalling to mTOR mediated by the Akt/PKB substrate PRAS40.
Nature cell biology
Vander Haar E,Lee SI,Bandhakavi S,Griffin TJ,Kim DH
Thu Mar 01 00:00:00 EST 2007
Insulin signalling to mTOR mediated by the Akt/PKB substrate PRAS40.
Nature cell biology
Vander Haar E,Lee SI,Bandhakavi S,Griffin TJ,Kim DH
Thu Mar 01 00:00:00 EST 2007
Insulin signalling to mTOR mediated by the Akt/PKB substrate PRAS40.
Nature cell biology
Vander Haar E,Lee SI,Bandhakavi S,Griffin TJ,Kim DH
Thu Mar 01 00:00:00 EST 2007
Target validation in drug discovery.
Methods in molecular biology (Clifton, N.J.)
Blake RA
Tue Nov 21 00:00:00 EST 2006