Product References
LAL deficiency induced myeloid-derived suppressor cells as targets and biomarkers for lung cancer.
Journal for immunotherapy of cancer
Zhao T,Liu S,Hanna NH,Jalal S,Ding X,Wan J,Yan C,Du H
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Wed Mar 01 00:00:00 EST 2023
Spontaneous Osteoclastogenesis, a risk factor for bone metastasis in advanced luminal A-type breast cancer patients.
Frontiers in oncology
Fernández Vallone V,Borzone FR,Martinez LM,Giorello MB,Choi H,Dimase F,Feldman L,Bordenave RH,Chudzinski-Tavassi AM,Batagelj E,Chasseing NA
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Mar 10 00:00:00 EST 2023
SARS-CoV-2 Delta (B.1.617.2) variant replicates and induces syncytia formation in human induced pluripotent stem cell-derived macrophages.
PeerJ
Thaweerattanasinp T,Wanitchang A,Saenboonrueng J,Srisutthisamphan K,Wanasen N,Sungsuwan S,Jongkaewwattana A,Chailangkarn T
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2023
Heliangin acts as a covalent ligand of RPS2 that disrupts pre-rRNA metabolic processes in NPM1-mutated acute myeloid leukemia.
Acta pharmaceutica Sinica. B
Feng Y,Han Y,Hu A,Qu Y,Hu Y,Wu H,Wang X,He L
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Wed Feb 01 00:00:00 EST 2023
Early macrophage response to obesity encompasses Interferon Regulatory Factor 5 regulated mitochondrial architecture remodelling.
Nature communications
Orliaguet L,Ejlalmanesh T,Humbert A,Ballaire R,Diedisheim M,Julla JB,Chokr D,Cuenco J,Michieletto J,Charbit J,Lindén D,Boucher J,Potier C,Hamimi A,Lemoine S,Blugeon C,Legoix P,Lameiras S,Baudrin LG,Baulande S,Soprani A,Castelli FA,Fenaille F,Riveline JP,Dalmas E,Rieusset J,Gautier JF,Venteclef N,Alzaid F
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Immunocytochemistry
Tue Aug 30 00:00:00 EDT 2022
A Matrix-Assisted Laser Desorption/Ionization Time-of-Flight Assay Identifies Nilotinib as an Inhibitor of Inflammation in Acute Myeloid Leukemia.
Journal of medicinal chemistry
Marín-Rubio JL,Peltier-Heap RE,Dueñas ME,Heunis T,Dannoura A,Inns J,Scott J,Simpson AJ,Blair HJ,Heidenreich O,Allan JM,Watt JE,Martin MP,Saxty B,Trost M
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Sep 22 00:00:00 EDT 2022
Decoding the pathogenesis of Diamond-Blackfan anemia using single-cell RNA-seq.
Cell discovery
Wang B,Wang C,Wan Y,Gao J,Ma Y,Zhang Y,Tong J,Zhang Y,Liu J,Chang L,Xu C,Shen B,Chen Y,Jiang E,Kurita R,Nakamura Y,Lim KC,Engel JD,Zhou J,Cheng T,Zhu X,Zhu P,Shi L
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Tue May 10 00:00:00 EDT 2022
SARS-CoV-2 infection paralyzes cytotoxic and metabolic functions of the immune cells.
Heliyon
Singh Y,Trautwein C,Fendel R,Krickeberg N,Berezhnoy G,Bissinger R,Ossowski S,Salker MS,Casadei N,Riess O
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
SARS-CoV-2 infection paralyzes cytotoxic and metabolic functions of the immune cells.
Heliyon
Singh Y,Trautwein C,Fendel R,Krickeberg N,Berezhnoy G,Bissinger R,Ossowski S,Salker MS,Casadei N,Riess O
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Tue Jun 01 00:00:00 EDT 2021
Exposure of a specific pleioform of multifunctional glyceraldehyde 3-phosphate dehydrogenase initiates CD14-dependent clearance of apoptotic cells.
Cell death & disease
Chaudhary S,Patidar A,Dhiman A,Chaubey GK,Dilawari R,Talukdar S,Modanwal R,Raje M
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Neutralization
Thu Sep 30 00:00:00 EDT 2021
The long non-coding RNA Cancer Susceptibility 15 (CASC15) is induced by isocitrate dehydrogenase (IDH) mutations and maintains an immature phenotype in adult acute myeloid leukemia.
Haematologica
Grasedieck S,Ruess C,Krowiorz K,Lux S,Pochert N,Schwarzer A,Klusmann JH,Jongen-Lavrencic M,Herold T,Bullinger L,Pollack JR,Rouhi A,Kuchenbauer F
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
Gene expression network analyses during infection with virulent and avirulent Trypanosoma cruzi strains unveil a role for fibroblasts in neutrophil recruitment and activation.
PLoS pathogens
Oliveira AER,Pereira MCA,Belew AT,Ferreira LRP,Pereira LMN,Neves EGA,Nunes MDCP,Burleigh BA,Dutra WO,El-Sayed NM,Gazzinelli RT,Teixeira SMR
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Sat Aug 01 00:00:00 EDT 2020
A functional antibody cross-reactive to both human and murine cytotoxic T-lymphocyte-associated protein 4 via binding to an N-glycosylation epitope.
mAbs
Li D,Li J,Chu H,Wang Z
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Feb 12 00:00:00 EST 2021
Differentiation Potential of Early- and Late-Passage Adipose-Derived Mesenchymal Stem Cells Cultured under Hypoxia and Normoxia.
Stem cells international
Zhao AG,Shah K,Freitag J,Cromer B,Sumer H
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Tue Oct 06 00:00:00 EDT 2020
MiR-103 protects from recurrent spontaneous abortion via inhibiting STAT1 mediated M1 macrophage polarization.
International journal of biological sciences
Zhu X,Liu H,Zhang Z,Wei R,Zhou X,Wang Z,Zhao L,Guo Q,Zhang Y,Chu C,Wang L,Li X
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Nov 12 00:00:00 EST 2021
Patient iPSC-Derived Macrophages to Study Inborn Errors of the IFN-γ Responsive Pathway.
Cells
Haake K,Neehus AL,Buchegger T,Kühnel MP,Blank P,Philipp F,Oleaga-Quintas C,Schulz A,Grimley M,Goethe R,Jonigk D,Kalinke U,Boisson-Dupuis S,Casanova JL,Bustamante J,Lachmann N
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Wed Feb 19 00:00:00 EST 2020
Chemerin enhances the adhesion and migration of human endothelial progenitor cells and increases lipid accumulation in mice with atherosclerosis.
Lipids in health and disease
Jia J,Yu F,Xiong Y,Wei W,Ma H,Nisi F,Song X,Yang L,Wang D,Yuan G,Zhou H
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Sun Sep 20 00:00:00 EDT 2020
NLRP3 Inflammasome Activation in Hemodialysis and Hypertensive Patients with Intact Kidney Function.
Toxins
Ulrich C,Wildgrube S,Fiedler R,Seibert E,Kneser L,Fick S,Schäfer C,Markau S,Trojanowicz B,Girndt M
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Oct 26 00:00:00 EDT 2020
LILRB3 (ILT5) is a myeloid cell checkpoint that elicits profound immunomodulation.
JCI insight
Yeboah M,Papagregoriou C,Jones DC,Chan HTC,Hu G,McPartlan JS,Schiött T,Mattson U,Mockridge CI,Tornberg UC,Hambe B,Ljungars A,Mattsson M,Tews I,Glennie MJ,Thirdborough SM,Trowsdale J,Frendeus B,Chen J,Cragg MS,Roghanian A
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2020
NLRP3 inflammasome expression in peripheral blood monocytes of coronary heart disease patients and its modulation by rosuvastatin.
Molecular medicine reports
Zhu J,Wu S,Hu S,Li H,Li M,Geng X,Wang H
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Aug 01 00:00:00 EDT 2019
Scalable high-throughput acoustophoresis in arrayed plastic microchannels.
Biomicrofluidics
Dubay R,Lissandrello C,Swierk P,Moore N,Doty D,Fiering J
56-0149 was used in Flow cytometry/Cell sorting to suggest that acoustophoresis could in the future aid in cell therapy bioprocessing and that further scale-up is possible.
Wed May 01 00:00:00 EDT 2019
Single-Cell Analysis Suggests that Ongoing Affinity Maturation Drives the Emergence of Pemphigus Vulgaris Autoimmune Disease.
Cell reports
Cho A,Caldara AL,Ran NA,Menne Z,Kauffman RC,Affer M,Llovet A,Norwood C,Scanlan A,Mantus G,Bradley B,Zimmer S,Schmidt T,Hertl M,Payne AS,Feldman R,Kowalczyk AP,Wrammert J
56-0149-42 was used in Flow cytometry/Cell sorting to find that Dsg3-specific MBCs have an activated phenotype and show signs of ongoing affinity maturation and clonal selection.
Tue Jul 23 00:00:00 EDT 2019
Characterizing the Role of Monocytes in T Cell Cancer Immunotherapy Using a 3D Microfluidic Model.
Frontiers in immunology
Lee SWL,Adriani G,Ceccarello E,Pavesi A,Tan AT,Bertoletti A,Kamm RD,Wong SC
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Jun 24 00:00:00 EDT 2019
AURKA Suppresses Leukemic THP-1 Cell Differentiation through Inhibition of the KDM6B Pathway.
Molecules and cells
Park JW,Cho H,Oh H,Kim JY,Seo SB
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu May 31 00:00:00 EDT 2018
Butyrate upregulates the TLR4 expression and the phosphorylation of MAPKs and NK-κB in colon cancer cell in vitro.
Oncology letters
Xiao T,Wu S,Yan C,Zhao C,Jin H,Yan N,Xu J,Wu Y,Li C,Shao Q,Xia S
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Oct 01 00:00:00 EDT 2018
Impaired efferocytosis by monocytes in multiple myeloma.
Oncology letters
Liang YY,Schwarzinger I,Simonitsch-Klupp I,Agis H,Oehler R
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Sun Jul 01 00:00:00 EDT 2018
Human umbilical cord-derived mesenchymal stem cells ameliorate the enteropathy of food allergies in mice.
Experimental and therapeutic medicine
Yan N,Xu J,Zhao C,Wu Y,Gao F,Li C,Zhou W,Xiao T,Zhou X,Shao Q,Xia S
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Sat Dec 01 00:00:00 EST 2018
Comprehensive characterization of chorionic villi-derived mesenchymal stromal cells from human placenta.
Stem cell research & therapy
Ventura Ferreira MS,Bienert M,Müller K,Rath B,Goecke T,Opländer C,Braunschweig T,Mela P,Brümmendorf TH,Beier F,Neuss S
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Feb 05 00:00:00 EST 2018
Cellular metabolism constrains innate immune responses in early human ontogeny.
Nature communications
Kan B,Michalski C,Fu H,Au HHT,Lee K,Marchant EA,Cheng MF,Anderson-Baucum E,Aharoni-Simon M,Tilley P,Mirmira RG,Ross CJ,Luciani DS,Jan E,Lavoie PM
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Nov 16 00:00:00 EST 2018
The pro-inflammatory phenotype of the human non-classical monocyte subset is attributed to senescence.
Cell death & disease
Ong SM,Hadadi E,Dang TM,Yeap WH,Tan CT,Ng TP,Larbi A,Wong SC
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Feb 15 00:00:00 EST 2018
A therapeutic T cell receptor mimic antibody targets tumor-associated PRAME peptide/HLA-I antigens.
The Journal of clinical investigation
Chang AY,Dao T,Gejman RS,Jarvis CA,Scott A,Dubrovsky L,Mathias MD,Korontsvit T,Zakhaleva V,Curcio M,Hendrickson RC,Liu C,Scheinberg DA
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Jun 30 00:00:00 EDT 2017
Toll-like receptor 3 activation selectively reverses HIV latency in microglial cells.
Retrovirology
Alvarez-Carbonell D,Garcia-Mesa Y,Milne S,Das B,Dobrowolski C,Rojas R,Karn J
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Immunofluorescence
Mon Feb 06 00:00:00 EST 2017
Human umbilical cord mesenchymal stem cells improve the reserve function of perimenopausal ovary via a paracrine mechanism.
Stem cell research & therapy
Li J,Mao Q,He J,She H,Zhang Z,Yin C
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Mar 09 00:00:00 EST 2017
Pulmonary sarcoidosis is associated with high-level inducible co-stimulator (ICOS) expression on lung regulatory T cells--possible implications for the ICOS/ICOS-ligand axis in disease course and resolution.
Clinical and experimental immunology
Sakthivel P,Grunewald J,Eklund A,Bruder D,Wahlström J
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2016
Antithymocyte Globulin at Clinically Relevant Concentrations Kills Leukemic Blasts.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Dabas R,Lee R,Servito MT,Dharmani-Khan P,Modi M,van Slyke T,Luider J,Durand C,Larratt L,Brandwein J,Morris D,Daly A,Khan FM,Storek J
Sun May 01 00:00:00 EDT 2016
Thrombomodulin regulates monocye differentiation via PKCδ and ERK1/2 pathway in vitro and in atherosclerotic artery.
Scientific reports
Tsai CS,Lin YW,Huang CY,Shih CM,Tsai YT,Tsao NW,Lin CS,Shih CC,Jeng H,Lin FY
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Dec 02 00:00:00 EST 2016
Thrombomodulin regulates monocye differentiation via PKCδ and ERK1/2 pathway in vitro and in atherosclerotic artery.
Scientific reports
Tsai CS,Lin YW,Huang CY,Shih CM,Tsai YT,Tsao NW,Lin CS,Shih CC,Jeng H,Lin FY
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri Dec 02 00:00:00 EST 2016
STK4 regulates TLR pathways and protects against chronic inflammation-related hepatocellular carcinoma.
The Journal of clinical investigation
Li W,Xiao J,Zhou X,Xu M,Hu C,Xu X,Lu Y,Liu C,Xue S,Nie L,Zhang H,Li Z,Zhang Y,Ji F,Hui L,Tao W,Wei B,Wang H
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Nov 02 00:00:00 EST 2015
Liposomal Glutathione Supplementation Restores TH1 Cytokine Response to Mycobacterium tuberculosis Infection in HIV-Infected Individuals.
Journal of interferon & cytokine research : the official journal of the International Society for Interferon and Cytokine Research
Ly J,Lagman M,Saing T,Singh MK,Tudela EV,Morris D,Anderson J,Daliva J,Ochoa C,Patel N,Pearce D,Venketaraman V
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Sun Nov 01 00:00:00 EDT 2015
Investigating the causes for decreased levels of glutathione in individuals with type II diabetes.
PloS one
Lagman M,Ly J,Saing T,Kaur Singh M,Vera Tudela E,Morris D,Chi PT,Ochoa C,Sathananthan A,Venketaraman V
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Wed Dec 30 00:00:00 EST 2015
A novel antibody-drug conjugate targeting SAIL for the treatment of hematologic malignancies.
Blood cancer journal
Kim SY,Theunissen JW,Balibalos J,Liao-Chan S,Babcock MC,Wong T,Cairns B,Gonzalez D,van der Horst EH,Perez M,Levashova Z,Chinn L,D'Alessio JA,Flory M,Bermudez A,Jackson DY,Ha E,Monteon J,Bruhns MF,Chen G,Migone TS
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Fri May 29 00:00:00 EDT 2015
The role of transforming growth factor β signaling in fibroblast-like synoviocytes from patients with oligoarticular juvenile idiopathic arthritis: dysregulation of transforming growth factor β signaling, including overexpression of bone morphogenetic protein 4, may lead to a chondrocyte phenotype and may contribute to bony hypertrophy.
Arthritis & rheumatology (Hoboken, N.J.)
Brescia AC,Simonds MM,McCahan SM,Fawcett PT,Rose CD
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Immunofluorescence
Thu May 01 00:00:00 EDT 2014
Human mesenchymal stem cells possess different biological characteristics but do not change their therapeutic potential when cultured in serum free medium.
Stem cell research & therapy
Wang Y,Wu H,Yang Z,Chi Y,Meng L,Mao A,Yan S,Hu S,Zhang J,Zhang Y,Yu W,Ma Y,Li T,Cheng Y,Wang Y,Wang S,Liu J,Han J,Li C,Liu L,Xu J,Han ZB,Han ZC
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Dec 04 00:00:00 EST 2014
Biologically active polymers from spontaneous carotenoid oxidation: a new frontier in carotenoid activity.
PloS one
Johnston JB,Nickerson JG,Daroszewski J,Mogg TJ,Burton GW
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Immunocytochemistry
Thu Jun 25 00:00:00 EDT 2015
Biologically active polymers from spontaneous carotenoid oxidation: a new frontier in carotenoid activity.
PloS one
Johnston JB,Nickerson JG,Daroszewski J,Mogg TJ,Burton GW
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Immunocytochemistry
Thu Jun 25 00:00:00 EDT 2015
Noncanonical dendritic cell differentiation and survival driven by a bacteremic pathogen.
Journal of leukocyte biology
Miles B,Scisci E,Carrion J,Sabino GJ,Genco CA,Cutler CW
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Aug 01 00:00:00 EDT 2013
Induction of autophagy is essential for monocyte-macrophage differentiation.
Blood
Zhang Y,Morgan MJ,Chen K,Choksi S,Liu ZG
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Thu Mar 22 00:00:00 EDT 2012
Negative regulation of JAK2 by H3K9 methyltransferase G9a in leukemia.
Molecular and cellular biology
Son HJ,Kim JY,Hahn Y,Seo SB
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2012
Ulcerative colitis impairs the acylethanolamide-based anti-inflammatory system reversal by 5-aminosalicylic acid and glucocorticoids.
PloS one
Suárez J,Romero-Zerbo Y,Márquez L,Rivera P,Iglesias M,Bermúdez-Silva FJ,Andreu M,Rodríguez de Fonseca F
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Immunofluorescence
Thu Dec 13 00:00:00 EST 2012
Upregulation of programmed death-1 on T cells and programmed death ligand-1 on monocytes in septic shock patients.
Critical care (London, England)
Zhang Y,Li J,Lou J,Zhou Y,Bo L,Zhu J,Zhu K,Wan X,Cai Z,Deng X
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Wed Dec 07 00:00:00 EST 2011
Ro60-associated single-stranded RNA links inflammation with fetal cardiac fibrosis via ligation of TLRs: a novel pathway to autoimmune-associated heart block.
Journal of immunology (Baltimore, Md. : 1950)
Clancy RM,Alvarez D,Komissarova E,Barrat FJ,Swartz J,Buyon JP
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Feb 15 00:00:00 EST 2010
B cells and monocytes from patients with active multiple sclerosis exhibit increased surface expression of both HERV-H Env and HERV-W Env, accompanied by increased seroreactivity.
Retrovirology
Brudek T,Christensen T,Aagaard L,Petersen T,Hansen HJ,Møller-Larsen A
Published figure using CD14 monoclonal antibody (Product # 56-0149-42) in Flow Cytometry
Mon Nov 16 00:00:00 EST 2009