Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAMDVDT PSGTNSGAGK KRFEVKKWNA VALWAWDIVV DNCAICRNHI MDLCIECQAN QASATSEECT VAWGVCNHAF HFHCISRWLK TRQVCPLDNR EWEFQKYGH (1-108 aa encoded by BC001466 )
This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase RBX1; E3 ubiquitin-protein transferase RBX1; Protein ZYP; Rbx1; Regulator of cullins 1; RING box protein 1; RING finger protein 75; ring-box 1, E3 ubiquitin protein ligase; RING-box protein 1; ROC1; ZYP protein
Gene Aliases: 1500002P15Rik; AA517855; BA554C12.1; RBX1; RNF75; ROC1
UniProt ID: (Human) P62877, (Mouse) P62878
Entrez Gene ID: (Human) 9978, (Mouse) 56438, (Rat) 300084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support