Product References
Transient expansion and myofibroblast conversion of adipogenic lineage precursors mediate bone marrow repair after radiation.
JCI insight
Zhong L,Yao L,Holdreith N,Yu W,Gui T,Miao Z,Elkaim Y,Li M,Gong Y,Pacifici M,Maity A,Busch TM,Joeng KS,Cengel K,Seale P,Tong W,Qin L
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Apr 08 00:00:00 EDT 2022
Lenvatinib for effectively treating antiangiogenic drug-resistant nasopharyngeal carcinoma.
Cell death & disease
Sun Q,Wang Y,Ji H,Sun X,Xie S,Chen L,Li S,Zeng W,Chen R,Tang Q,Zuo J,Hou L,Hosaka K,Lu Y,Liu Y,Ye Y,Yang Y
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Aug 19 00:00:00 EDT 2022
Tet-mediated DNA demethylation regulates specification of hematopoietic stem and progenitor cells during mammalian embryogenesis.
Science advances
Ma L,Tang Q,Gao X,Lee J,Lei R,Suzuki M,Zheng D,Ito K,Frenette PS,Dawlaty MM
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Mar 04 00:00:00 EST 2022
Sphk2 deletion is involved in structural abnormalities and Th17 response but does not aggravate colon inflammation induced by sub-chronic stress.
Scientific reports
Martín-Hernández D,Gutiérrez IL,González-Prieto M,MacDowell KS,Robledo-Montaña J,Tendilla-Beltrán H,Calleja-Rodríguez N,Bris ÁG,Ulecia-Morón C,Moreno B,Caso JR,García-Bueno B,Rodrigues-Mascarenhas S,Marín-Jiménez I,Leza JC,Menchén L
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Mar 08 00:00:00 EST 2022
Aged hematopoietic stem cells are refractory to bloodborne systemic rejuvenation interventions.
The Journal of experimental medicine
Ho TT,Dellorusso PV,Verovskaya EV,Bakker ST,Flach J,Smith LK,Ventura PB,Lansinger OM,Hérault A,Zhang SY,Kang YA,Mitchell CA,Villeda SA,Passegué E
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Mon Jul 05 00:00:00 EDT 2021
A rapid CRISPR competitive assay for in vitro and in vivo discovery of potential drug targets affecting the hematopoietic system.
Computational and structural biotechnology journal
Shen Y,Jiang L,Iyer VS,Raposo B,Dubnovitsky A,Boddul SV,Kasza Z,Wermeling F
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Wed Nov 10 00:00:00 EST 2021
Defining the Mechanistic Correlates of Protection Conferred by Whole-Cell Vaccination against Pseudomonas aeruginosa Acute Murine Pneumonia.
Infection and immunity
Sen-Kilic E,Blackwood CB,Huckaby AB,Horspool AM,Weaver KL,Malkowski AC,Witt WT,Bevere JR,Damron FH,Barbier M
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Jan 19 00:00:00 EST 2021
Polyvinyl alcohol hydrolysis rate and molecular weight influence human and murine HSC activity ex vivo.
Stem cell research
Sudo K,Yamazaki S,Wilkinson AC,Nakauchi H,Nakamura Y
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
Impaired HA-specific T follicular helper cell and antibody responses to influenza vaccination are linked to inflammation in humans.
eLife
Hill DL,Whyte CE,Innocentin S,Lee JL,Dooley J,Wang J,James EA,Lee JC,Kwok WW,Zand MS,Liston A,Carr EJ,Linterman MA
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Nov 02 00:00:00 EDT 2021
Metabolic preconditioning in CD4+ T cells restores inducible immune tolerance in lupus-prone mice.
JCI insight
Wilson CS,Stocks BT,Hoopes EM,Rhoads JP,McNew KL,Major AS,Moore DJ
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Oct 08 00:00:00 EDT 2021
The folate cycle enzyme MTHFD2 induces cancer immune evasion through PD-L1 up-regulation.
Nature communications
Shang M,Yang H,Yang R,Chen T,Fu Y,Li Y,Fang X,Zhang K,Zhang J,Li H,Cao X,Gu J,Xiao J,Zhang Q,Liu X,Yu Q,Wang T
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Mon Mar 29 00:00:00 EDT 2021
Type 2 diabetic mice enter a state of spontaneous hibernation-like suspended animation following accumulation of uric acid.
The Journal of biological chemistry
Zhao Y,Cheng R,Zhao Y,Ge W,Yang Y,Ding Z,Xu X,Wang Z,Wu Z,Zhang J
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Oct 01 00:00:00 EDT 2021
SARS-CoV-2 mRNA Vaccines Foster Potent Antigen-Specific Germinal Center Responses Associated with Neutralizing Antibody Generation.
Immunity
Lederer K,Castaño D,Gómez Atria D,Oguin TH,Wang S,Manzoni TB,Muramatsu H,Hogan MJ,Amanat F,Cherubin P,Lundgreen KA,Tam YK,Fan SHY,Eisenlohr LC,Maillard I,Weissman D,Bates P,Krammer F,Sempowski GD,Pardi N,Locci M
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Dec 15 00:00:00 EST 2020
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Sep 17 00:00:00 EDT 2020
Myeloid Cell CK2 Regulates Inflammation and Resistance to Bacterial Infection.
Frontiers in immunology
Larson SR,Bortell N,Illies A,Crisler WJ,Matsuda JL,Lenz LL
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Jul 27 00:00:00 EDT 2021
Shp1 Loss Enhances Macrophage Effector Function and Promotes Anti-Tumor Immunity.
Frontiers in immunology
Myers DR,Abram CL,Wildes D,Belwafa A,Welsh AMN,Schulze CJ,Choy TJ,Nguyen T,Omaque N,Hu Y,Singh M,Hansen R,Goldsmith MA,Quintana E,Smith JAM,Lowell CA
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Mon Jun 14 00:00:00 EDT 2021
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
Tpl2 Protects Against Fulminant Hepatitis Through Mobilization of Myeloid-Derived Suppressor Cells.
Frontiers in immunology
Xu J,Pei S,Wang Y,Liu J,Qian Y,Huang M,Zhang Y,Xiao Y
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Wed Oct 14 00:00:00 EDT 2020
Bacteroides fragilis polysaccharide A induces IL-10 secreting B and T cells that prevent viral encephalitis.
Nature communications
Ramakrishna C,Kujawski M,Chu H,Li L,Mazmanian SK,Cantin EM
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue May 14 00:00:00 EDT 2019
The Hematopoietic Oxidase NOX2 Regulates Self-Renewal of Leukemic Stem Cells.
Cell reports
Adane B,Ye H,Khan N,Pei S,Minhajuddin M,Stevens BM,Jones CL,D'Alessandro A,Reisz JA,Zaberezhnyy V,Gasparetto M,Ho TC,Kelly KK,Myers JR,Ashton JM,Siegenthaler J,Kume T,Campbell EL,Pollyea DA,Becker MW,Jordan CT
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Apr 02 00:00:00 EDT 2019
Adrenergic nerve degeneration in bone marrow drives aging of the hematopoietic stem cell niche.
Nature medicine
Maryanovich M,Zahalka AH,Pierce H,Pinho S,Nakahara F,Asada N,Wei Q,Wang X,Ciero P,Xu J,Leftin A,Frenette PS
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Jun 01 00:00:00 EDT 2018
Mitophagy in Intestinal Epithelial Cells Triggers Adaptive Immunity during Tumorigenesis.
Cell
Ziegler PK,Bollrath J,Pallangyo CK,Matsutani T,Canli Ö,De Oliveira T,Diamanti MA,Müller N,Gamrekelashvili J,Putoczki T,Horst D,Mankan AK,Öner MG,Müller S,Müller-Höcker J,Kirchner T,Slotta-Huspenina J,Taketo MM,Reinheckel T,Dröse S,Larner AC,Wels WS,Ernst M,Greten TF,Arkan MC,Korn T,Wirth D,Greten FR
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Jun 28 00:00:00 EDT 2018
Proapoptotic BIM Impacts B Lymphoid Homeostasis by Limiting the Survival of Mature B Cells in a Cell-Autonomous Manner.
Frontiers in immunology
Liu R,King A,Bouillet P,Tarlinton DM,Strasser A,Heierhorst J
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Wed May 01 00:00:00 EDT 2019
Host Tumor Suppressor p18INK4c Functions as a Potent Cell-Intrinsic Inhibitor of Murine Gammaherpesvirus 68 Reactivation and Pathogenesis.
Journal of virology
Niemeyer BF,Oko LM,Medina EM,Oldenburg DG,White DW,Cool CD,Clambey ET,van Dyk LF
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Mar 15 00:00:00 EDT 2018
Inactivation of mTORC1 Signaling in Osterix-Expressing Cells Impairs B-cell Differentiation.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research
Wang Y,Xiao M,Tao C,Chen J,Wang Z,Yang J,Chen Z,Zou Z,Liu A,Cai D,Jiang Y,Ding C,Li M,Bai X
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Sun Apr 01 00:00:00 EDT 2018
Tanshinone IIA and Astragaloside IV promote the angiogenesis of mesenchymal stem cell-derived endothelial cell-like cells via upregulation of Cx37, Cx40 and Cx43.
Experimental and therapeutic medicine
Li Z,Zhang S,Cao L,Li W,Ye YC,Shi ZX,Wang ZR,Sun LX,Wang JW,Jia LT,Wang W
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Feb 01 00:00:00 EST 2018
miR-143/145 differentially regulate hematopoietic stem and progenitor activity through suppression of canonical TGFβ signaling.
Nature communications
Lam J,van den Bosch M,Wegrzyn J,Parker J,Ibrahim R,Slowski K,Chang L,Martinez-Høyer S,Condorelli G,Boldin M,Deng Y,Umlandt P,Fuller M,Karsan A
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Wed Jun 20 00:00:00 EDT 2018
Dysregulated TRAF3 and BCL2 Expression Promotes Multiple Classes of Mature Non-hodgkin B Cell Lymphoma in Mice.
Frontiers in immunology
Perez-Chacon G,Adrados M,Vallejo-Cremades MT,Lefebvre S,Reed JC,Zapata JM
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Oct 25 00:00:00 EDT 2019
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
EZH2 enables germinal centre formation through epigenetic silencing of CDKN1A and an Rb-E2F1 feedback loop.
Nature communications
Béguelin W,Rivas MA,Calvo Fernández MT,Teater M,Purwada A,Redmond D,Shen H,Challman MF,Elemento O,Singh A,Melnick AM
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Oct 12 00:00:00 EDT 2017
DNA-binding of the Tet-transactivator curtails antigen-induced lymphocyte activation in mice.
Nature communications
Ottina E,Peperzak V,Schoeler K,Carrington E,Sgonc R,Pellegrini M,Preston S,Herold MJ,Strasser A,Villunger A
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Wed Oct 18 00:00:00 EDT 2017
The Interleukin (IL)-1R1 pathway is a critical negative regulator of PyMT-mediated mammary tumorigenesis and pulmonary metastasis.
Oncoimmunology
Dagenais M,Dupaul-Chicoine J,Douglas T,Champagne C,Morizot A,Saleh M
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Jan 12 00:00:00 EST 2021
Irgm1 coordinately regulates autoimmunity and host defense at select mucosal surfaces.
JCI insight
Azzam KM,Madenspacher JH,Cain DW,Lai L,Gowdy KM,Rai P,Janardhan K,Clayton N,Cunningham W,Jensen H,Patel PS,Kearney JF,Taylor GA,Fessler MB
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017
Multiple functional therapeutic effects of TnP: A small stable synthetic peptide derived from fish venom in a mouse model of multiple sclerosis.
PloS one
Komegae EN,Souza TA,Grund LZ,Lima C,Lopes-Ferreira M
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Aug 17 00:00:00 EDT 2017
Murine iPSC-Derived Macrophages as a Tool for Disease Modeling of Hereditary Pulmonary Alveolar Proteinosis due to Csf2rb Deficiency.
Stem cell reports
Mucci A,Kunkiel J,Suzuki T,Brennig S,Glage S,Kühnel MP,Ackermann M,Happle C,Kuhn A,Schambach A,Trapnell BC,Hansen G,Moritz T,Lachmann N
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Aug 09 00:00:00 EDT 2016
Mitochondrial ATP transporter Ant2 depletion impairs erythropoiesis and B lymphopoiesis.
Cell death and differentiation
Cho J,Seo J,Lim CH,Yang L,Shiratsuchi T,Lee MH,Chowdhury RR,Kasahara H,Kim JS,Oh SP,Lee YJ,Terada N
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Sep 01 00:00:00 EDT 2015
Disruption of p21-activated kinase 1 gene diminishes atherosclerosis in apolipoprotein E-deficient mice.
Nature communications
Singh NK,Kotla S,Dyukova E,Traylor JG,Orr AW,Chernoff J,Marion TN,Rao GN
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Wed Jun 24 00:00:00 EDT 2015
IκBε is a key regulator of B cell expansion by providing negative feedback on cRel and RelA in a stimulus-specific manner.
Journal of immunology (Baltimore, Md. : 1950)
Alves BN,Tsui R,Almaden J,Shokhirev MN,Davis-Turak J,Fujimoto J,Birnbaum H,Ponomarenko J,Hoffmann A
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Tue Apr 01 00:00:00 EDT 2014
Reciprocal effects of rab7 deletion in activated and neglected T cells.
Autophagy
Roy SG,Stevens MW,So L,Edinger AL
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Mon Jul 01 00:00:00 EDT 2013
TDP2-dependent non-homologous end-joining protects against topoisomerase II-induced DNA breaks and genome instability in cells and in vivo.
PLoS genetics
Gómez-Herreros F,Romero-Granados R,Zeng Z,Alvarez-Quilón A,Quintero C,Ju L,Umans L,Vermeire L,Huylebroeck D,Caldecott KW,Cortés-Ledesma F
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Thu Jun 13 00:00:00 EDT 2013
Activation-induced cytidine deaminase-initiated off-target DNA breaks are detected and resolved during S phase.
Journal of immunology (Baltimore, Md. : 1950)
Hasham MG,Snow KJ,Donghia NM,Branca JA,Lessard MD,Stavnezer J,Shopland LS,Mills KD
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2012
Essential role of phospholipase C gamma 2 in early B-cell development and Myc-mediated lymphomagenesis.
Molecular and cellular biology
Wen R,Chen Y,Bai L,Fu G,Schuman J,Dai X,Zeng H,Yang C,Stephan RP,Cleveland JL,Wang D
Published figure using CD45R (B220) monoclonal antibody (Product # 83-0452-42) in Flow Cytometry
Fri Dec 01 00:00:00 EST 2006
A role for brain-derived neurotrophic factor in B cell development.
Journal of neuroimmunology
Schuhmann B,Dietrich A,Sel S,Hahn C,Klingenspor M,Lommatzsch M,Gudermann T,Braun A,Renz H,Nockher WA
Wed Jun 01 00:00:00 EDT 2005
A role for brain-derived neurotrophic factor in B cell development.
Journal of neuroimmunology
Schuhmann B,Dietrich A,Sel S,Hahn C,Klingenspor M,Lommatzsch M,Gudermann T,Braun A,Renz H,Nockher WA
Wed Jun 01 00:00:00 EDT 2005