Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2610509L04Rik; 4E-T; CD40 ligand-activated specific transcript 4; eIF4E transporter; eIF4E-transporter; Eukaryotic translation initiation factor 4E nuclear import factor 1; Eukaryotic translation initiation factor 4E transporter; FLJ21601; FLJ26551; OTTHUMP00000063276
Gene Aliases: 2610509L04Rik; 4E-T; A930019J01Rik; AA410001; AU021239; Clast4; D11Ertd166e; EIF4ENIF1
UniProt ID: (Human) Q9NRA8, (Mouse) Q9EST3
Entrez Gene ID: (Human) 56478, (Mouse) 74203
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support