Product References
Melanoma-derived gangliosides impair migratory and antigen-presenting function of human epidermal Langerhans cells and induce their apoptosis.
International immunology
Bennaceur K,Popa I,Portoukalian J,Berthier-Vergnes O,Péguet-Navarro J
AHS4002 was used in flow cytometry to investigate the effects of gangliosides purified from human melanoma tumors on human epidermal Langerhans cells
Thu Jun 01 00:00:00 EDT 2006
Activation of mesangial cells by platelets in systemic lupus erythematosus via a CD154-dependent induction of CD40.
Kidney international
Delmas Y,Viallard JF,Solanilla A,Villeneuve J,Pasquet JM,Belloc F,Dubus I,Déchanet-Merville J,Merville P,Blanco P,Pellegrin JL,Nurden AT,Combe C,Ripoche J
AHS4002 was used in flow cytometry to test if platelet-associated CD154 activates mesangial cells
Tue Nov 01 00:00:00 EST 2005
UVA radiation impairs phenotypic and functional maturation of human dermal dendritic cells.
The Journal of investigative dermatology
Furio L,Berthier-Vergnes O,Ducarre B,Schmitt D,Peguet-Navarro J
AHS4002 was used in flow cytometry to determine the effects of ultraviolet A radiation on dermal dendritic cells
Tue Nov 01 00:00:00 EST 2005
Adenosine-activated mast cells induce IgE synthesis by B lymphocytes: an A2B-mediated process involving Th2 cytokines IL-4 and IL-13 with implications for asthma.
Journal of immunology (Baltimore, Md. : 1950)
Ryzhov S,Goldstein AE,Matafonov A,Zeng D,Biaggioni I,Feoktistov I
AHS4002 was used in ELISA to test if adenosine up-regulates Th2 cytokines in mast cells.
Tue Jun 15 00:00:00 EDT 2004
Evidence for modulation of human epidermal differentiation and remodelling by CD40.
The British journal of dermatology
Concha M,Vidal MA,Moreno I,Salem C,Figueroa CD,Schmitt D,Péguet-Navarro J
AHS4002 was used in blocking or activating experiment to elucidate the significance of CD40 expression on normal human keratinocytes
Sun Jun 01 00:00:00 EDT 2003
Effects of low concentrations of cadmium on immunoglobulin E production by human B lymphocytes in vitro.
Toxicology
Jelovcan S,Gutschi A,Kleinhappl B,Sedlmayr P,Barth S,Marth E
AHS4002 was used in blocking or activating experiment to examine the consequences of cadmium exposure on human immunoglobulin E synthesis
Tue Jun 03 00:00:00 EDT 2003
Disruption of CD40/CD40 ligand interaction with cleavage of CD40 on human gingival fibroblasts by human leukocyte elastase resulting in down-regulation of chemokine production.
Journal of leukocyte biology
Nemoto E,Tada H,Shimauchi H
AHS4002 was used in flow cytometry to propose that human leukocyte elastase inhibits CD40-dependent cell activation.
Sun Sep 01 00:00:00 EDT 2002
Maturation of human dendritic cells as sulfasalazine target.
Croatian medical journal
Matasić R,Dietz AB,Vuk-Pavlović S
AHS4002 was used in blocking assay to examine the effects of sulfasalazine and its metabolites on human dendritic cells.
Wed Aug 01 00:00:00 EDT 2001
Dendritic cells transduced by multiply deleted HIV-1 vectors exhibit normal phenotypes and functions and elicit an HIV-specific cytotoxic T-lymphocyte response in vitro.
Blood
Gruber A,Kan-Mitchell J,Kuhen KL,Mukai T,Wong-Staal F
AHS4002 was used in flow cytometry to investigate using dendritic cells as physiologic adjuvants.
Tue Aug 15 00:00:00 EDT 2000
Cyclooxygenase-independent inhibition of dendritic cell maturation by aspirin.
Immunology
Matasic R,Dietz AB,Vuk-Pavlovic S
AHS4002 was used in blocking assay to elucidate how aspirin inhibits dendritic cell function.
Fri Sep 01 00:00:00 EDT 2000
Effect of stimulation of human macrophages on intracellular survival of Mycobacterium bovis Bacillus Calmette-Guerin. Evaluation with a mycobacterial reporter strain.
American journal of respiratory and critical care medicine
Bonay M,Bouchonnet F,Pelicic V,Lagier B,Grandsaigne M,Lecossier D,Grodet A,Vokurka M,Gicquel B,Hance AJ
AHS4002 was used in blocking assay to study mycobacterial survival in human monocyte-derived macrophages.
Sat May 01 00:00:00 EDT 1999
Productive infection of dendritic cells by HIV-1 and their ability to capture virus are mediated through separate pathways.
The Journal of clinical investigation
Blauvelt A,Asada H,Saville MW,Klaus-Kovtun V,Altman DJ,Yarchoan R,Katz SI
Wed Oct 15 00:00:00 EDT 1997
CD40 is a functional activation antigen and B7-independent T cell costimulatory molecule on normal human lung fibroblasts.
Journal of immunology (Baltimore, Md. : 1950)
Sempowski GD,Chess PR,Phipps RP
AHS4002 was used in flow cytometry to elucidate role of CD40 in fibroblasts.
Thu May 15 00:00:00 EDT 1997
CD40-mediated signals inhibit the binding of TNF receptor-associated factor 2 to the CD40 cytoplasmic domain.
Journal of immunology (Baltimore, Md. : 1950)
Chaudhuri A,Orme S,Eilam S,Cherayil BJ
AHS4002 was used in blocking assay and immunoprecipitation to elucidate the mechanisms that regulate the inhibition of the CD40-TRAF2 interaction by CD40 signals.
Sat Nov 01 00:00:00 EST 1997
CD40 expression by gingival fibroblasts: correlation of phenotype with function.
International immunology
Dongari-Bagtzoglou AI,Warren WD,Berton MT,Ebersole JL
AHS4002 was used in flow cytometry to report a functional relationship between CD40 expression and IL-6 or IL-8 secretion by gingival fibroblast.
Mon Sep 01 00:00:00 EDT 1997
Activated mast cells produce interleukin 13.
The Journal of experimental medicine
Burd PR,Thompson WC,Max EE,Mills FC
AHS4002 was used in blocking assay to study IL-13 secretion by mast cells.
Sat Apr 01 00:00:00 EST 1995