Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AQP-8; Aquaporin-8; Aquaporin8; membrane water channel; MGC93492; water channel
Gene Aliases: AI255744; AQP-8; AQP8
UniProt ID: (Human) O94778, (Rat) P56405, (Mouse) P56404
Entrez Gene ID: (Human) 343, (Sheep) 443183, (Bovine) 450206, (Rat) 29172, (Mouse) 11833
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support