Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: VAP-1 (TK8-14) is a mouse monoclonal antibody raised against affinity purifiedVAP-1 from tonsil stroma of human origin.
Lymphocyte binding to vascular endothelium is a prerequisite for the move-ment of immune cells from the blood into lymphoid tissues and into sites ofinflammation. Under inflammatory conditions, cell surface expression ofVAP-1 (vascular adhesion protein-1) which is an endothelial sialoglycoprotein,is induced. VAP-1 is a type II transmembrane protein of 84.6 kDa with a singletransmembrane domain and N and O-glycosylation sites in the extracellulardomain. In vivo, VAP-1 exists predominantly as a 180 kDa homodimer andfunctions both as an enzyme (monoamine oxidase) and an adhesion moleculefor lymphocytes.
Applications Tested: ELISA, Flow Cytometry, Immunofluorescence, Western Blotting.
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been discribed and is located approximately 9-kb downstream.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amine oxidase [copper-containing] 3; Amine oxidase copper-containing 3; amine oxidase, copper containing 3 (vascular adhesion protein 1); Copper amine oxidase; HPAO; placenta copper monamine oxidase; Semicarbazide-sensitive amine oxidase; SSAO; VAP-1; Vascular adhesion protein 1
Gene Aliases: AOC3; HPAO; SSAO; VAP-1; VAP1
UniProt ID: (Human) Q16853
Entrez Gene ID: (Human) 8639
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support