Product References
Nuclear speckle integrity and function require TAO2 kinase.
Proceedings of the National Academy of Sciences of the United States of America
Gao S,Esparza M,Dehghan I,Aksenova V,Zhang K,Batten K,Ferretti MB,Begg BE,Cagatay T,Shay JW,García-Sastre A,Goldsmith EJ,Chen ZJ,Dasso M,Lynch KW,Cobb MH,Fontoura BMA
MA1-082 was used in Western Blotting to find that a nuclear pool of TAO2 is localized at nuclear speckles and interacts with nuclear speckle factors involved in RNA splicing and nuclear export, including SRSF1 and Aly/Ref.
Tue Jun 21 00:00:00 EDT 2022
Expression of influenza A virus-derived peptides on a rotavirus VP6-based delivery platform.
Archives of virology
Gröhn S,Heinimäki S,Tamminen K,Blazevic V
MA1-082 was used in Western Blot to explore different rotavirus medium-layer VP6 protein insertion sites for the presentation of two influenza A virus peptides toward potential vaccination strategies.
Fri Jan 01 00:00:00 EST 2021
Thapsigargin Is a Broad-Spectrum Inhibitor of Major Human Respiratory Viruses: Coronavirus, Respiratory Syncytial Virus and Influenza A Virus.
Viruses
Al-Beltagi S,Preda CA,Goulding LV,James J,Pu J,Skinner P,Jiang Z,Wang BL,Yang J,Banyard AC,Mellits KH,Gershkovich P,Hayes CJ,Nguyen-Van-Tam J,Brown IH,Liu J,Chang KC
Published figure using Influenza A M2 monoclonal antibody (Product # MA1-082) in Western Blot
Wed Feb 03 00:00:00 EST 2021
Thapsigargin Is a Broad-Spectrum Inhibitor of Major Human Respiratory Viruses: Coronavirus, Respiratory Syncytial Virus and Influenza A Virus.
Viruses
Al-Beltagi S,Preda CA,Goulding LV,James J,Pu J,Skinner P,Jiang Z,Wang BL,Yang J,Banyard AC,Mellits KH,Gershkovich P,Hayes CJ,Nguyen-Van-Tam J,Brown IH,Liu J,Chang KC
Published figure using Influenza A M2 monoclonal antibody (Product # MA1-082) in Western Blot
Wed Feb 03 00:00:00 EST 2021
Influenza A virus co-opts ERI1 exonuclease bound to histone mRNA to promote viral transcription.
Nucleic acids research
Declercq M,Biquand E,Karim M,Pietrosemoli N,Jacob Y,Demeret C,Barbezange C,van der Werf S
Published figure using Influenza A M2 monoclonal antibody (Product # MA1-082) in Western Blot
Fri Oct 09 00:00:00 EDT 2020
Influenza A virus co-opts ERI1 exonuclease bound to histone mRNA to promote viral transcription.
Nucleic acids research
Declercq M,Biquand E,Karim M,Pietrosemoli N,Jacob Y,Demeret C,Barbezange C,van der Werf S
Published figure using Influenza A M2 monoclonal antibody (Product # MA1-082) in Western Blot
Fri Oct 09 00:00:00 EDT 2020
Double-Layered M2e-NA Protein Nanoparticle Immunization Induces Broad Cross-Protection against Different Influenza Viruses in Mice.
Advanced healthcare materials
Wang Y,Deng L,Gonzalez GX,Luthra L,Dong C,Ma Y,Zou J,Kang SM,Wang BZ
MA1-082 was used in Western Blotting to investigate if double-layered protein nanoparticles containing structure-stabilized M2e and NA can be developed into a universal influenza vaccine or a synergistic component of such vaccines.
Wed Jan 01 00:00:00 EST 2020
Chemical intervention of influenza virus mRNA nuclear export.
PLoS pathogens
Esparza M,Mor A,Niederstrasser H,White K,White A,Zhang K,Gao S,Wang J,Liang J,Sho S,Sakthivel R,Sathe AA,Xing C,Muñoz-Moreno R,Shay JW,García-Sastre A,Ready J,Posner B,Fontoura BMA
Published figure using Influenza A M2 monoclonal antibody (Product # MA1-082) in Western Blot
Wed Apr 01 00:00:00 EDT 2020
A non-death function of the mitochondrial apoptosis apparatus in immunity.
The EMBO journal
Brokatzky D,Dörflinger B,Haimovici A,Weber A,Kirschnek S,Vier J,Metz A,Henschel J,Steinfeldt T,Gentle IE,Häcker G
MA1-082 was used in Immunocytochemistry-immunoflourescence to report that such sub-lethal engagement of mitochondrial apoptosis signaling causes the secretion of cytokines from human epithelial cells in a process controlled by the Bcl-2 family of proteins. As well as showing that sub-lethal signaling of the mitochondrial apoptosis pathway is initiated by infections with all tested viral, bacterial, and protozoan pathogens and causes damage to the genomic DNA.
Mon Jun 03 00:00:00 EDT 2019
Double-layered protein nanoparticles induce broad protection against divergent influenza A viruses.
Nature communications
Deng L,Mohan T,Chang TZ,Gonzalez GX,Wang Y,Kwon YM,Kang SM,Compans RW,Champion JA,Wang BZ
MA1-082 was used in Western Blotting to report a methodology to generate double-layered protein nanoparticles as a universal influenza vaccine.
Wed Jan 24 00:00:00 EST 2018
Influenza virus differentially activates mTORC1 and mTORC2 signaling to maximize late stage replication.
PLoS pathogens
Kuss-Duerkop SK,Wang J,Mena I,White K,Metreveli G,Sakthivel R,Mata MA,Muñoz-Moreno R,Chen X,Krammer F,Diamond MS,Chen ZJ,García-Sastre A,Fontoura BMA
Published figure using Influenza A M2 monoclonal antibody (Product # MA1-082) in Western Blot
Fri Sep 01 00:00:00 EDT 2017
Recruitment of RED-SMU1 complex by Influenza A Virus RNA polymerase to control Viral mRNA splicing.
PLoS pathogens
Fournier G,Chiang C,Munier S,Tomoiu A,Demeret C,Vidalain PO,Jacob Y,Naffakh N
MA1-082 was used in western blot to determine viral mRNA splicing controlled by the recruitment of RED-SMU1 complex by Influenza A Virus RNA polymerase
Sun Jun 01 00:00:00 EDT 2014
Kinetics of proton transport into influenza virions by the viral M2 channel.
PloS one
Ivanovic T,Rozendaal R,Floyd DL,Popovic M,van Oijen AM,Harrison SC
MA1-082 was used in Western Blotting to study the kinetics of proton transport into influenza virions by the viral M2 channel.
Thu Aug 02 00:00:00 EDT 2012
Increased immunogenicity and protective efficacy of influenza M2e fused to a tetramerizing protein.
PloS one
Andersson AM,Håkansson KO,Jensen BA,Christensen D,Andersen P,Thomsen AR,Christensen JP
MA1-082 was used in flow cytometry to generate a tetrameric form of the influenza A M2e protein ectodomain with increased immunogenicity and protective efficacy
Fri Feb 22 00:00:00 EST 2013
Characterization of a fully human monoclonal antibody against extracellular domain of matrix protein 2 of influenza A virus.
Antiviral research
Ozawa T,Jin A,Tajiri K,Takemoto M,Okuda T,Shiraki K,Kishi H,Muraguchi A
MA1-082 was used in western blot to isolate and characterize a human antibody to influenza virus matrix protein 2 (M2e)
Thu Sep 01 00:00:00 EDT 2011
Matrix protein 2 of influenza A virus blocks autophagosome fusion with lysosomes.
Cell host & microbe
Gannagé M,Dormann D,Albrecht R,Dengjel J,Torossi T,Rämer PC,Lee M,Strowig T,Arrey F,Conenello G,Pypaert M,Andersen J,García-Sastre A,Münz C
MA1-082 was used in western blot to study the role of matrix protein 2 of influenza A virus in blocking autophagosome fusion with lysosomes
Thu Oct 22 00:00:00 EDT 2009