Product References
Intermittent hypoxia during recovery from neonatal hyperoxic lung injury causes long-term impairment of alveolar development: A new rat model of BPD.
American journal of physiology. Lung cellular and molecular physiology
Mankouski A,Kantores C,Wong MJ,Ivanovska J,Jain A,Benner EJ,Mason SN,Tanswell AK,Auten RL,Jankov RP
MA112772 was used in immunohistochemistry - paraffin section to refine the rat model of bronchopulmonary dysplasia
Wed Feb 01 00:00:00 EST 2017
Simvastatin prevents and reverses chronic pulmonary hypertension in newborn rats via pleiotropic inhibition of RhoA signaling.
American journal of physiology. Lung cellular and molecular physiology
Wong MJ,Kantores C,Ivanovska J,Jain A,Jankov RP
MA112772 was used in immunohistochemistry - paraffin section to propose that simvastatin inhibits RhoA/ROCK activity in the chronic hypoxia-exposed lung
Tue Nov 01 00:00:00 EDT 2016
Human pathological basis of blood vessels and stromal tissue for nanotechnology.
Advanced drug delivery reviews
Nishihara H
MA1-12772 was used in immunocytochemistry to review the pathophysiology of nanotechnology
Tue Jul 01 00:00:00 EDT 2014
Tight skin 2 mice exhibit a novel time line of events leading to increased extracellular matrix deposition and dermal fibrosis.
Matrix biology : journal of the International Society for Matrix Biology
Long KB,Artlett CM,Blankenhorn EP
MA1-12772 was used in immunohistochemistry to study the time-course of extracellular matrix deposition and dermal fibrogenesis in the murine Tsk2 model of systemic sclerosis
Mon Sep 01 00:00:00 EDT 2014
Long-term B cell depletion in murine lupus eliminates autoantibody-secreting cells and is associated with alterations in the kidney plasma cell niche.
Journal of immunology (Baltimore, Md. : 1950)
Wang W,Rangel-Moreno J,Owen T,Barnard J,Nevarez S,Ichikawa HT,Anolik JH
MA1-12772 was used in immunohistochemistry to study the effects on auto-antibody secreting cells and the kidney plasma cell niche of long-term rituximab-mediated B cell depletion in a murine lupus model
Tue Apr 01 00:00:00 EDT 2014
Human adipose tissue possesses a unique population of pluripotent stem cells with nontumorigenic and low telomerase activities: potential implications in regenerative medicine.
Stem cells and development
Ogura F,Wakao S,Kuroda Y,Tsuchiyama K,Bagheri M,Heneidi S,Chazenbalk G,Aiba S,Dezawa M
MA1-12772 was used in immunohistochemistry to study the implications for tissue engineering and regenerative medicine of the identification of a population of human adipose tissue stem cells with low telomerase that lack teratogenic activity
Tue Apr 01 00:00:00 EDT 2014
The time-dependent expression of α7nAChR during skeletal muscle wound healing in rats.
International journal of legal medicine
Fan YY,Zhang ST,Yu LS,Ye GH,Lin KZ,Wu SZ,Dong MW,Han JG,Feng XP,Li XB
MA1-12772 was used in immunohistochemistry to study the time-course and cellular localization of alpha7nAChR expression in rat skeletal muscle following contusion injury
Mon Sep 01 00:00:00 EDT 2014
Use of platelet-rich plasma solution applied with composite chondrocutaneous graft technique: an experimental study in rabbit model.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons
Sevim KZ,Yazar M,Irmak F,Tekkeşin MS,Yildiz K,Sirvan SS
MA1-12772 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of using platelet-rich plasma in composite chondrocutaneous graft procedures
Tue Jul 01 00:00:00 EDT 2014
Dynamic intersection of the longitudinal muscle and external anal sphincter in the layered structure of the anal canal posterior wall.
Surgical and radiologic anatomy : SRA
Muro S,Yamaguchi K,Nakajima Y,Watanabe K,Harada M,Nimura A,Akita K
MA1-12772 was used in immunohistochemistry to study the detailed structure of the posterior anal canal
Fri Aug 01 00:00:00 EDT 2014
Impaired cornea wound healing in a tenascin C-deficient mouse model.
Laboratory investigation; a journal of technical methods and pathology
Sumioka T,Kitano A,Flanders KC,Okada Y,Yamanaka O,Fujita N,Iwanishi H,Kao WW,Saika S
MA1-12772 was used in immunohistochemistry to study the stromal healing of injured corneas in mice ablated for tenascin C
Fri Feb 01 00:00:00 EST 2013
Thy-1 knockdown retards wound repair in mouse skin.
Journal of dermatological science
Lee MJ,Shin JO,Jung HS
MA1-12772 was used in immunohistochemistry to study the role of Thy-1 in the repair of murine skin wounds using siRNA-mediated knockdown
Fri Feb 01 00:00:00 EST 2013
The effect of platelet-rich plasma on flap survival in random extension of an axial pattern flap in rabbits.
Plastic and reconstructive surgery
Kim HY,Park JH,Han YS,Kim H
MA1-12772 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of autologous platelet-rich plasma on axial pattern flap procedures in a rabbit model
Mon Jul 01 00:00:00 EDT 2013
Cyclic adenosine monophosphate response-element binding protein activation by mitogen-activated protein kinase-activated protein kinase 3 and four-and-a-half LIM domains 5 plays a key role for vein graft intimal hyperplasia.
Journal of vascular surgery
Nakanishi K,Saito Y,Azuma N,Sasajima T
MA1-12772 was used in immunohistochemistry to study the intimal hyperplasia of vein grafts and the roles played by CREB, MAPKAPK3 and FHL5
Tue Jan 01 00:00:00 EST 2013
Cannabinoid receptor 1 suppresses transient receptor potential vanilloid 1-induced inflammatory responses to corneal injury.
Cellular signalling
Yang Y,Yang H,Wang Z,Varadaraj K,Kumari SS,Mergler S,Okada Y,Saika S,Kingsley PJ,Marnett LJ,Reinach PS
MA1-12772 was used in immunohistochemistry to study the mechanism by which injury-induced activativation of CB1 receptors leads to downregulation of corneal TRPV1-induced inflammation
Fri Feb 01 00:00:00 EST 2013
Malignant transformation of adenomyoepithelioma of the breast by a monophasic population: a report of two cases and review of literature.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica
Marian C,Boila A,Soanca D,Malau M,Podeanu DM,Resetkova E,Stolnicu S
MA1-12772 was used in immunohistochemistry to report on two cases of malignant transformation of adenomyoepithelioma of the breast by a monophasic population
Mon Apr 01 00:00:00 EDT 2013
Contribution of DOG1 expression to the diagnosis of gastrointestinal stromal tumors.
Pathology, research and practice
Kara T,Serinsoz E,Arpaci RB,Gubur O,Orekici G,Ata A,Colak T,Arican A
MA1-12772 was used in immunohistochemistry to study the diagnostic value of the immunohistochemical expression of DOG1 in gastrointestinal stromal tumors
Mon Jul 01 00:00:00 EDT 2013
Time-dependent expression and distribution of Egr-1 during skeletal muscle wound healing in rats.
Journal of molecular histology
Fan YY,Ye GH,Lin KZ,Yu LS,Wu SZ,Dong MW,Han JG,Feng XP,Li XB
MA1-12772 was used in immunohistochemistry to study the temporal and cellular expression of Egr-1 during wound repair in rat skeletal muscle
Fri Feb 01 00:00:00 EST 2013
DNA microarray and quantitative analysis reveal enhanced myocardial VEGF expression with stunted angiogenesis in human tetralogy of Fallot.
Cell biochemistry and biophysics
Peters TH,Sharma V,Yilmaz E,Mooi WJ,Bogers AJ,Sharma HS
MA1-12772 was used in immunohistochemistry to study the role of elevated myocardial expression of VEGF in the pathology of human tetralogy of Fallot
Fri Nov 01 00:00:00 EDT 2013
The effects of hyperbaric oxygen application against cholestatic oxidative stress and hepatic damage after bile duct ligation in rats.
The Journal of surgical research
Ayvaz S,Kanter M,Aksu B,Sahin SH,Uzun H,Erboga M,Pul M
MA1-12772 was used in immunohistochemistry to study the mechanisms underlying the beneficial effects of hyperbaric oxygen against oxidative and cholestatic hepatic damage caused by bile duct ligation in rats
Mon Jul 01 00:00:00 EDT 2013
Stromal-epithelial crosstalk provides a suitable microenvironment for the progression of ovarian cancer cells in vitro.
Cancer investigation
Fu S,Dong L,Sun W,Xu Y,Gao L,Miao Y
MA1-12772 was used in immunocytochemistry to study the in vivo progression of ovarian cancer cells and the role of stromal-epithelial interactions in creating an appropriate microenvironment
Fri Nov 01 00:00:00 EDT 2013
Protective effect of Urtica dioica on liver damage induced by biliary obstruction in rats.
Toxicology and industrial health
Oguz S,Kanter M,Erboga M,Ibis C
MA1-12772 was used in immunohistochemistry to study the ability of Urtica dioica to protect against liver damage in a rat bile duct ligation model
Tue Oct 01 00:00:00 EDT 2013
Effect of mesenchymal stem cells on anti-Thy1,1 induced kidney injury in albino rats.
Asian Pacific journal of tropical biomedicine
Sakr S,Rashed L,Zarouk W,El-Shamy R
MA1-12772 was used in immunohistochemistry to study the beneficial effects in a rat model of nephritis of therapy with bone marrow-derived mesenchymal stem cells
Fri Mar 01 00:00:00 EST 2013
Mesenchymal-to-endothelial transition in Kaposi sarcoma: a histogenetic hypothesis based on a case series and literature review.
PloS one
Gurzu S,Ciortea D,Munteanu T,Kezdi-Zaharia I,Jung I
MA1-12772 was used in immunohistochemistry to develop a hypothesis of Kaposi sarcoma histogenesis based on a case and literature review
Tue Apr 01 00:00:00 EDT 2014
Keratocyte apoptosis and not myofibroblast differentiation mark the graft/host interface at early time-points post-DSAEK in a cat model.
PloS one
Weis AJ,Huxlin KR,Callan CL,DeMagistris MA,Hindman HB
MA1-12772 was used in immunohistochemistry to study keratinocyte apoptosis as a graft-host marker in a feline model of Descemet's stripping automated endothelial keratoplasty
Mon May 12 00:00:00 EDT 2014
Diet induced mild hypercholesterolemia in pigs: local and systemic inflammation, effects on vascular injury - rescue by high-dose statin treatment.
PloS one
Busnelli M,Manzini S,Froio A,Vargiolu A,Cerrito MG,Smolenski RT,Giunti M,Cinti A,Zannoni A,Leone BE,Forni M,Bacci ML,Biasi GM,Giovannoni R,Lavitrano M
MA1-12772 was used in immunohistochemistry to study the effects of high-dose statin therapy on inflammation and vascular injury in a porcine model of mild hypercholesterolemia
Tue Aug 26 00:00:00 EDT 2014
Attenuation of CCl4-induced hepatic fibrosis in mice by vaccinating against TGF-β1.
PloS one
Fan X,Zhang Q,Li S,Lv Y,Su H,Jiang H,Hao Z
MA1-12772 was used in western blot to study the ability of vaccination against TGF-beta1 to protect against hepatic fibrosis in a liver injury model
Sun Oct 12 00:00:00 EDT 2014
Oral administration of recombinant adeno-associated virus-mediated bone morphogenetic protein-7 suppresses CCl(4)-induced hepatic fibrosis in mice.
Molecular therapy : the journal of the American Society of Gene Therapy
Hao ZM,Cai M,Lv YF,Huang YH,Li HH
MA1-12772 was used in immunohistochemistry to study the ability of an orally administered recombinant AAV-BMP-7 virus to protect against experimentally induced hepatic fibrosis in a murine model
Thu Nov 01 00:00:00 EDT 2012
Transgenic expression of interferon-γ in mouse stomach leads to inflammation, metaplasia, and dysplasia.
The American journal of pathology
Syu LJ,El-Zaatari M,Eaton KA,Liu Z,Tetarbe M,Keeley TM,Pero J,Ferris J,Wilbert D,Kaatz A,Zheng X,Qiao X,Grachtchouk M,Gumucio DL,Merchant JL,Samuelson LC,Dlugosz AA
MA1-12772 was used in immunohistochemistry to study the induction of inflammation, metaplasia and displasia following transgenic expression of IFN-gamma in the murine stomach
Sat Dec 01 00:00:00 EST 2012
Loss of semaphorin-neuropilin-1 signaling causes dysmorphic vascularization reminiscent of alveolar capillary dysplasia.
The American journal of pathology
Joza S,Wang J,Fox E,Hillman V,Ackerley C,Post M
MA1-12772 was used in immunohistochemistry to study the role of SEMA3-neuropilin-1 signaling in the vascular development of the fetal lung and the effects of signaling deficiency
Sat Dec 01 00:00:00 EST 2012
The pulmonary mesenchymal tissue layer is defective in an in vitro recombinant model of nitrofen-induced lung hypoplasia.
The American journal of pathology
van Loenhout RB,Tseu I,Fox EK,Huang Z,Tibboel D,Post M,Keijzer R
MA1-12772 was used in immunohistochemistry to develop a recombinant in vitro cellular model of lung hypoplasia
Sun Jan 01 00:00:00 EST 2012
Multipotent nestin-expressing stem cells capable of forming neurons are located in the upper, middle and lower part of the vibrissa hair follicle.
Cell cycle (Georgetown, Tex.)
Amoh Y,Mii S,Aki R,Hamada Y,Kawahara K,Hoffman RM,Katsuoka K
MA1-12772 was used in immunohistochemistry to study hair follicle multipotent stem cells that express nestin and are capable of differentiating into neurons
Sat Sep 15 00:00:00 EDT 2012
Asbestos exposure induces alveolar epithelial cell plasticity through MAPK/Erk signaling.
Journal of cellular biochemistry
Tamminen JA,Myllärniemi M,Hyytiäinen M,Keski-Oja J,Koli K
MA1-12772 was used in western blot to study the role of MAPK/Erk signaling in the mechanism by which asbestos exposure induces plasticity in alveolar epithelial cells
Sun Jul 01 00:00:00 EDT 2012
Vaccination with platelet-derived growth factor B kinoids inhibits CCl₄-induced hepatic fibrosis in mice.
The Journal of pharmacology and experimental therapeutics
Hao ZM,Fan XB,Li S,Lv YF,Su HQ,Jiang HP,Li HH
MA1-12772 was used in western blot to study the ability of vaccination against PDGF-D to protect against hepatic fibrosis in a rat model of acute liver injury
Sat Sep 01 00:00:00 EDT 2012
Regulation of TGF-β storage and activation in the human idiopathic pulmonary fibrosis lung.
Cell and tissue research
Leppäranta O,Sens C,Salmenkivi K,Kinnula VL,Keski-Oja J,Myllärniemi M,Koli K
MA1-12772 was used in immunohistochemistry to study the role of LTBP-1 in modulating TGF-beta levels in the lungs of patients with idiopathic pulmonary fibrosis
Fri Jun 01 00:00:00 EDT 2012
Protective effects of thymoquinone against cholestatic oxidative stress and hepatic damage after biliary obstruction in rats.
Journal of molecular histology
Oguz S,Kanter M,Erboga M,Erenoglu C
MA1-12772 was used in immunohistochemistry to study the mechanisms underlying the protective and therapeutic effects of thymoquinone in a rat model of cholestatic liver disease
Sun Apr 01 00:00:00 EDT 2012
Fstl1 antagonizes BMP signaling and regulates ureter development.
PloS one
Xu J,Qi X,Gong J,Yu M,Zhang F,Sha H,Gao X
MA1-12772 was used in immunocytochemistry to study the antagonization of BMP signaling by Fstl1 and the significance for normal development of the ureter
Fri Jul 27 00:00:00 EDT 2012
Accreta complicating complete placenta previa is characterized by reduced systemic levels of vascular endothelial growth factor and by epithelial-to-mesenchymal transition of the invasive trophoblast.
American journal of obstetrics and gynecology
Wehrum MJ,Buhimschi IA,Salafia C,Thung S,Bahtiyar MO,Werner EF,Campbell KH,Laky C,Sfakianaki AK,Zhao G,Funai EF,Buhimschi CS
MA1-12772 was used in immunohistochemistry to study the serum angiogenic factor profile of women with complete placenta previa
Sun May 01 00:00:00 EDT 2011
Myometrial wound healing post-Cesarean delivery in the MRL/MpJ mouse model of uterine scarring.
The American journal of pathology
Buhimschi CS,Zhao G,Sora N,Madri JA,Buhimschi IA
MA1-12772 was used in immunohistochemistry to investigate myometrial wound healing process after cesarean delivery
Thu Jul 01 00:00:00 EDT 2010
Purification and characterization of mouse lacrimal gland epithelial cells and reconstruction of an acinarlike structure in three-dimensional culture.
Investigative ophthalmology & visual science
Ueda Y,Karasawa Y,Satoh Y,Nishikawa S,Imaki J,Ito M
MA1-12772 was used in immunocytochemistry to study which factors are involved in postnatal lacrimal gland development
Fri May 01 00:00:00 EDT 2009
Chick pulmonary Wnt5a directs airway and vascular tubulogenesis.
Development (Cambridge, England)
Loscertales M,Mikels AJ,Hu JK,Donahoe PK,Roberts DJ
MA1-12772 was used in immunohistochemistry to study the role of non-cannonical Wnt5a in modulating pulmonary airway and vascular tubulogenesis
Tue Apr 01 00:00:00 EDT 2008
Epithelioid leiomyosarcoma with rhabdoid features.
Wiener klinische Wochenschrift
Yorulmaz G,Erdogan G,Pestereli HE,Savas B,Karaveli FS
MA1-12772 was used in immunohistochemistry to report on a case of epithelioid leiomyosarcoma with rhabdoid features
Wed Jan 30 00:00:00 EST 2008
Renoprotective properties of angiotensin receptor blockers beyond blood pressure lowering.
Journal of the American Society of Nephrology : JASN
Izuhara Y,Nangaku M,Inagi R,Tominaga N,Aizawa T,Kurokawa K,van Ypersele de Strihou C,Miyata T
MA1-12772 was used in immunohistochemistry to study the ability of angiotensin receptor blockers to protect against kidney damage independently of blood pressure lowering effects
Thu Dec 01 00:00:00 EST 2005
Ginkgo biloba extract reverses CCl4-induced liver fibrosis in rats.
World journal of gastroenterology
Luo YJ,Yu JP,Shi ZH,Wang L
MA1-12772 was used in immunohistochemistry to study the potential mechanisms by which gonko biloba extract reverses CCl4-induced liver injury in a mouse model
Thu Apr 01 00:00:00 EST 2004
Synchronous primary adenocarcinoma and gastrointestinal stromal tumor in the stomach: a report of two cases.
The Turkish journal of gastroenterology : the official journal of Turkish Society of Gastroenterology
Bircan S,Candir O,Aydin S,Başpinar S,Bülbül M,Kapucuoğlu N,Karahan N,Ciriş M
MA1-12772 was used in immunohistochemistry to report on two patients with synchronous primary adenocarcinoma and gastrointestinal stromal tumor in the stomach
Wed Sep 01 00:00:00 EDT 2004