Product References
The Cortisol Response of Male and Female Choroidal Endothelial Cells: Implications for Central Serous Chorioretinopathy.
The Journal of clinical endocrinology and metabolism
Brinks J,van Dijk EHC,Kiełbasa SM,Mei H,van der Veen I,Peters HAB,Sips HCM,Notenboom RGE,Quax PHA,Boon CJF,Meijer OC
MA1-150 was used in Immunohistochemistry to show that the marked response of particular target genes in endothelial cells to cortisol, such as ZBTB16, warrants further investigation into their potential role in the pathophysiology of CSC and other vascular conditions.
Tue Jan 18 00:00:00 EST 2022
Adverse effect of rheumatoid arthritis on male Wistar rat's fertility: protective role of Costus extract.
Environmental science and pollution research international
Kamel S,Tag HM,Ebeid H,Khaled HE,Almallah AA,El-Naggar MS
MA1-150 was used in Immunohistochemistry to assess the protective effect of Costus extract on the fertility of male rats with Freund's adjuvant-induced rheumatoid arthritis.
Sat Jan 01 00:00:00 EST 2022
Nobiletin protects against diabetes-induced testicular injury via hypophysis-gonadal axis upregulation and amelioration of oxidative stress.
Molecular biology reports
Salah M,Ismail KA,Khadrawy SM
MA1-150 was used in Immunohistochemistry to compare the effects of two different doses of nobiletin and analyze its mechanisms of action against diabetes-induced testicular impairment in rats.
Sat Jan 01 00:00:00 EST 2022
Chlorella vulgaris or Spirulina platensis mitigate lead acetate-induced testicular oxidative stress and apoptosis with regard to androgen receptor expression in rats.
Environmental science and pollution research international
Ibrahim IA,Shalaby AA,Abd Elaziz RT,Bahr HI
MA1-150 was used in Immunohistochemistry to conclude that further clinical trials are needed to test S. platensis and C. vulgaris as a promising candidate in treating male infertility.
Sun Aug 01 00:00:00 EDT 2021
A novel immunocompetent model of metastatic prostate cancer-induced bone pain.
The Prostate
Liu Z,Murphy SF,Huang J,Zhao L,Hall CC,Schaeffer AJ,Schaeffer EM,Thumbikat P
MA1-150 was used in Immunohistochemistry to describe a prostate cancer cell-originated bone pain model and demonstrate pain responses to be associated with increased expressions of calcitonin gene-related peptide (CGRP) and neuronal p75NTR in primary afferent neurons and enhanced expression of nerve growth factor (NGF) and TrkA in a population of neurons in the superficial dorsal horn.
Wed Jul 01 00:00:00 EDT 2020
The Impact of Hippocampal Sex Hormones Receptors in Modulation of Depressive-Like Behavior Following Chronic Anabolic Androgenic Steroids and Exercise Protocols in Rats.
Frontiers in behavioral neuroscience
Selakovic D,Joksimovic J,Jovicic N,Mitrovic S,Mihailovic V,Katanic J,Milovanovic D,Pantovic S,Mijailovic N,Rosic G
MA1-150 was used in Immunohistochemistry to evaluate alterations in depressive-like behaviours in rats following chronic administration of a supraphysiological dose of anabolic androgenic steroids, as well as exposure to a prolonged exercise protocol.
Tue Sep 29 00:00:00 EDT 2020
Ameliorating effects of green tea extract on cadmium induced reproductive injury in male Wistar rats with respect to androgen receptors and caspase- 3.
Reproductive biology
Abdelrazek HM,Helmy SA,Elsayed DH,Ebaid HM,Mohamed RM
MA1150 was used in immunohistochemistry - paraffin section to study the protective role of green tea extract on cadmium chloride-induced reproductive toxicities in rats
Thu Dec 01 00:00:00 EST 2016
Effects of lead acetate on testicular function and caspase-3 expression with respect to the protective effect of cinnamon in albino rats.
Toxicology reports
Elgawish RAR,Abdelrazek HMA
MA1-150 was used in Immunohistochemistry to investigate the protective effects of cinnamon on lead acetate induced reproductive toxicities in rats.
Sun Nov 12 00:00:00 EST 2023
Differential regulation of gonadotropin-releasing hormone secretion and gene expression by androgen: membrane versus nuclear receptor activation.
Molecular endocrinology (Baltimore, Md.)
Shakil T,Hoque AN,Husain M,Belsham DD
MA1-150 was used in western blot to investigate the effect of androgen on gonadotropin-releasing hormone secretion and gene expression.
Fri Nov 01 00:00:00 EST 2002
Cyclin E as a coactivator of the androgen receptor.
The Journal of cell biology
Yamamoto A,Hashimoto Y,Kohri K,Ogata E,Kato S,Ikeda K,Nakanishi M
MA1-150 was used in western blot to study the relationship between cyclin E ad androgen receptor.
Mon Aug 21 00:00:00 EDT 2000
Expression of maspin in prostate cells is regulated by a positive ets element and a negative hormonal responsive element site recognized by androgen receptor.
Proceedings of the National Academy of Sciences of the United States of America
Zhang M,Magit D,Sager R
MA1-150 was used in EMSA to study the transcriptional regulation of maspin in the prostate.
Tue May 27 00:00:00 EDT 1997
Atypical androgen receptor in the human melanoma cell line IIB-MEL-J.
Pigment cell research
Morvillo V,Luthy IA,Bravo AI,Capurro MI,Donaldson M,Quintans C,Calandra RS,Mordoh J
MA1-150 was used in immunohistochemistry to investigate the expression of androgen receptor in IIB-MEL-J cells
Thu Jun 01 00:00:00 EDT 1995
Human androgen receptor binding to the androgen response element of prostate specific antigen.
Journal of andrology
Luke MC,Coffey DS
MA1-150 was used in EMSA to study the in vitro binding of the human androgen receptor to the putative androgen response element of prostate specific antigen
Thu Jun 23 00:00:00 EDT 1994
Immunohistochemistry and biochemistry in detection of androgen, progesterone, and estrogen receptors in benign and malignant human prostatic tissue.
The Prostate
Brolin J,Skoog L,Ekman P
MA1-150 was used in immunohistochemistry to investigate the distribution of AR, ER and PR in human prostatic tumor cell lines
Thu Jul 23 00:00:00 EDT 1992
Evidence for the presence of androgen receptors in the synovial tissue of rheumatoid arthritis patients and healthy controls.
Arthritis and rheumatism
Cutolo M,Accardo S,Villaggio B,Clerico P,Indiveri F,Carruba G,Fecarotta E,Castagnetta L
MA1-150 was used in immunohistochemistry to investigate the expression of androgen receptor in the synovial tissue from rheumatoid arthritic patients
Tue Sep 01 00:00:00 EDT 1992
Androgen receptor and 5 alpha-reductase activity in the ductuli efferentes and epididymis of adult rhesus macaques.
Biology of reproduction
Roselli CE,West NB,Brenner RM
MA1-150 was used in immunocytochemistry to study the immunocytochemical distribution of AR in comparison with biochemical quantification of AR and 5 alpha-reductase activity in the ductuli efferentes and epididymides of adult male rhesus monkeys
Mon Apr 01 00:00:00 EST 1991
Prostate androgen receptor: immunohistological localization and mRNA characterization.
Journal of steroid biochemistry
Chang C,Chodak G,Sarac E,Takeda H,Liao S
MA1-150 was used in immunohistochemistry to investigate the distribution of androgen receptor in vivo
Fri Mar 30 00:00:00 EST 1990