Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Suggested positive control: antigen standard for NKX3 (transient overexpression lysate), LNCAP cell extracts, mouse testis protein, mouse testis tissue lysate.
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Drosophila NK-3 transcription factor, locus 1; Homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK-3 transcription factor, locus 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NKX3-1 antibody; prostate-specific ag
Gene Aliases: bagpipe; BAPX2; Bax; Nkx-3.1; NKX3; NKX3-1; NKX3.1; NKX3A
UniProt ID: (Human) Q99801, (Mouse) P97436
Entrez Gene ID: (Human) 4824, (Mouse) 18095, (Rat) 305999
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support